Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 128572785-128573330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC01565
Entrez Gene ID
23434
Gene Name
long intergenic non-protein coding RNA 1565
Gene Aliases
C3orf27, GR6
UniGene
Hs.194283
Gene Chromosome Location
Chr.3: 128572000-128576086 on Build GRCh38
Transcripts
NR_125802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
445532 c.435C>T p.F145F Chr.3: 128573295-128573295 on Build GRCh38
6334573 c.423C>A p.S141R Chr.3: 128573307-128573307 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs879936826 Chr.3: 128573110-128573110 on Build GRCh38
rs149766819 Chr.3: 128572970-128572970 on Build GRCh38
rs373592252 Chr.3: 128573058-128573058 on Build GRCh38
rs766174371 Chr.3: 128573266-128573267 on Build GRCh38
rs771928293 Chr.3: 128573245-128573245 on Build GRCh38
rs2734044 Chr.3: 128572897-128572897 on Build GRCh38
rs181491347 Chr.3: 128572836-128572836 on Build GRCh38
rs568856571 Chr.3: 128573160-128573160 on Build GRCh38
rs774952677 Chr.3: 128573288-128573288 on Build GRCh38
rs920318403 Chr.3: 128572936-128572936 on Build GRCh38
rs533909068 Chr.3: 128573301-128573301 on Build GRCh38
rs369311056 Chr.3: 128573240-128573240 on Build GRCh38
rs760981637 Chr.3: 128573096-128573096 on Build GRCh38
rs977166264 Chr.3: 128572850-128572850 on Build GRCh38
rs763757882 Chr.3: 128573097-128573097 on Build GRCh38
rs374799271 Chr.3: 128573283-128573283 on Build GRCh38
rs778895323 Chr.3: 128572841-128572841 on Build GRCh38
rs1037948081 Chr.3: 128572900-128572900 on Build GRCh38
rs539335515 Chr.3: 128573216-128573216 on Build GRCh38
rs1051936864 Chr.3: 128573263-128573263 on Build GRCh38
rs571372814 Chr.3: 128573048-128573048 on Build GRCh38
rs1020804089 Chr.3: 128573116-128573116 on Build GRCh38
rs750855434 Chr.3: 128573307-128573307 on Build GRCh38
rs370018358 Chr.3: 128572978-128572978 on Build GRCh38
rs551609666 Chr.3: 128573044-128573044 on Build GRCh38
rs902316012 Chr.3: 128573128-128573128 on Build GRCh38
rs773043502 Chr.3: 128573258-128573258 on Build GRCh38
rs1019252770 Chr.3: 128573189-128573189 on Build GRCh38
rs566322692 Chr.3: 128573270-128573270 on Build GRCh38
rs928377693 Chr.3: 128572854-128572855 on Build GRCh38
rs2734043 Chr.3: 128572887-128572887 on Build GRCh38
rs200851555 Chr.3: 128573159-128573164 on Build GRCh38
rs966617496 Chr.3: 128572861-128572861 on Build GRCh38
rs938413074 Chr.3: 128572854-128572854 on Build GRCh38
rs187017507 Chr.3: 128572869-128572869 on Build GRCh38
rs375624455 Chr.3: 128573266-128573266 on Build GRCh38
rs200346912 Chr.3: 128573220-128573220 on Build GRCh38
rs945881855 Chr.3: 128572844-128572844 on Build GRCh38
rs764437243 Chr.3: 128573282-128573282 on Build GRCh38
rs771172822 Chr.3: 128572860-128572860 on Build GRCh38
rs578072730 Chr.3: 128572856-128572856 on Build GRCh38
rs549324029 Chr.3: 128572875-128572875 on Build GRCh38
rs930855167 Chr.3: 128572937-128572937 on Build GRCh38
rs879487088 Chr.3: 128572938-128572938 on Build GRCh38
rs958057981 Chr.3: 128573025-128573025 on Build GRCh38
rs991218288 Chr.3: 128573209-128573209 on Build GRCh38
rs893205571 Chr.3: 128573067-128573067 on Build GRCh38
rs928446257 Chr.3: 128573122-128573122 on Build GRCh38
rs770633777 Chr.3: 128573279-128573279 on Build GRCh38
rs762034250 Chr.3: 128573299-128573299 on Build GRCh38
rs942283606 Chr.3: 128572899-128572899 on Build GRCh38
rs751216443 Chr.3: 128573294-128573295 on Build GRCh38
rs974536464 Chr.3: 128572929-128572929 on Build GRCh38
rs929520480 Chr.3: 128573246-128573246 on Build GRCh38
rs990513881 Chr.3: 128573045-128573045 on Build GRCh38
rs1001640698 Chr.3: 128572829-128572829 on Build GRCh38
rs759096435 Chr.3: 128573281-128573281 on Build GRCh38
rs145728397 Chr.3: 128572976-128572976 on Build GRCh38
rs2734045 Chr.3: 128573217-128573217 on Build GRCh38
rs116311182 Chr.3: 128572986-128572986 on Build GRCh38
rs754816351 Chr.3: 128573234-128573234 on Build GRCh38
rs908172965 Chr.3: 128572949-128572949 on Build GRCh38
rs1033489344 Chr.3: 128572832-128572832 on Build GRCh38
rs527823165 Chr.3: 128572923-128572923 on Build GRCh38

View Full Product Details