Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 128884404-128884905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAD9
Entrez Gene ID
28976
Gene Name
acyl-CoA dehydrogenase family member 9
Gene Aliases
NPD002
UniGene
Hs.567482
Gene Chromosome Location
Chr.3: 128879490-128916075 on Build GRCh38
Transcripts
NM_014049.4 NR_033426.1 XM_017006264.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4113285 c.238G>A p.E80K Chr.3: 128884740-128884740 on Build GRCh38
7076475 c.241G>A p.E81K Chr.3: 128884743-128884743 on Build GRCh38
1038008 c.220G>A p.V74M Chr.3: 128884722-128884722 on Build GRCh38
278569 c.231C>A p.F77L Chr.3: 128884733-128884733 on Build GRCh38
7200588 c.158delT p.F54fs*48 Chr.3: 128884660-128884660 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs863223877 Chr.3: 128884677-128884677 on Build GRCh38
rs75748817 Chr.3: 128884453-128884453 on Build GRCh38
rs765257311 Chr.3: 128884624-128884625 on Build GRCh38
rs149538775 Chr.3: 128884599-128884599 on Build GRCh38
rs765060373 Chr.3: 128884707-128884707 on Build GRCh38
rs996004696 Chr.3: 128884689-128884689 on Build GRCh38
rs147206100 Chr.3: 128884773-128884773 on Build GRCh38
rs768744842 Chr.3: 128884567-128884567 on Build GRCh38
rs1002233668 Chr.3: 128884774-128884774 on Build GRCh38
rs914323328 Chr.3: 128884826-128884826 on Build GRCh38
rs765422380 Chr.3: 128884779-128884779 on Build GRCh38
rs745405919 Chr.3: 128884721-128884721 on Build GRCh38
rs762859894 Chr.3: 128884651-128884654 on Build GRCh38
rs149931573 Chr.3: 128884654-128884654 on Build GRCh38
rs762959806 Chr.3: 128884786-128884786 on Build GRCh38
rs752716687 Chr.3: 128884708-128884708 on Build GRCh38
rs863225058 Chr.3: 128884651-128884651 on Build GRCh38
rs980368033 Chr.3: 128884446-128884446 on Build GRCh38
rs981229614 Chr.3: 128884462-128884462 on Build GRCh38
rs1054180935 Chr.3: 128884504-128884504 on Build GRCh38
rs1027899962 Chr.3: 128884797-128884797 on Build GRCh38
rs965739037 Chr.3: 128884865-128884866 on Build GRCh38
rs892800284 Chr.3: 128884533-128884533 on Build GRCh38
rs773302706 Chr.3: 128884641-128884641 on Build GRCh38
rs766026673 Chr.3: 128884651-128884652 on Build GRCh38
rs904560266 Chr.3: 128884646-128884646 on Build GRCh38
rs949924138 Chr.3: 128884459-128884459 on Build GRCh38
rs773015796 Chr.3: 128884631-128884632 on Build GRCh38
rs140606723 Chr.3: 128884798-128884798 on Build GRCh38
rs779258809 Chr.3: 128884722-128884722 on Build GRCh38
rs748758547 Chr.3: 128884733-128884733 on Build GRCh38
rs771313668 Chr.3: 128884766-128884766 on Build GRCh38
rs773642310 Chr.3: 128884754-128884754 on Build GRCh38
rs761039361 Chr.3: 128884653-128884653 on Build GRCh38
rs927067006 Chr.3: 128884467-128884467 on Build GRCh38
rs983038590 Chr.3: 128884816-128884816 on Build GRCh38
rs772314192 Chr.3: 128884745-128884745 on Build GRCh38
rs564971522 Chr.3: 128884579-128884579 on Build GRCh38
rs752560901 Chr.3: 128884783-128884783 on Build GRCh38
rs945736228 Chr.3: 128884846-128884846 on Build GRCh38
rs760918247 Chr.3: 128884755-128884755 on Build GRCh38
rs144978857 Chr.3: 128884697-128884697 on Build GRCh38
rs566886864 Chr.3: 128884487-128884487 on Build GRCh38
rs529115687 Chr.3: 128884810-128884810 on Build GRCh38
rs758120770 Chr.3: 128884712-128884712 on Build GRCh38
rs767966274 Chr.3: 128884625-128884625 on Build GRCh38
rs373584581 Chr.3: 128884769-128884769 on Build GRCh38
rs567195433 Chr.3: 128884486-128884486 on Build GRCh38
rs983197522 Chr.3: 128884484-128884484 on Build GRCh38
rs557401373 Chr.3: 128884719-128884719 on Build GRCh38
rs371244300 Chr.3: 128884715-128884715 on Build GRCh38
rs762062780 Chr.3: 128884613-128884613 on Build GRCh38
rs777302134 Chr.3: 128884598-128884598 on Build GRCh38
rs774729637 Chr.3: 128884610-128884610 on Build GRCh38
rs770395658 Chr.3: 128884706-128884706 on Build GRCh38
rs777679870 Chr.3: 128884713-128884713 on Build GRCh38
rs138999688 Chr.3: 128884440-128884440 on Build GRCh38
rs377484856 Chr.3: 128884667-128884667 on Build GRCh38
rs796334086 Chr.3: 128884565-128884565 on Build GRCh38
rs907595638 Chr.3: 128884536-128884536 on Build GRCh38

View Full Product Details