Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 128904295-128904794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAD9
Entrez Gene ID
28976
Gene Name
acyl-CoA dehydrogenase family member 9
Gene Aliases
NPD002
UniGene
Hs.567482
Gene Chromosome Location
Chr.3: 128879490-128916075 on Build GRCh38
Transcripts
NM_014049.4 NR_033426.1 XM_017006264.1 XM_017006265.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
581464 c.1063G>A p.V355I Chr.3: 128904419-128904419 on Build GRCh38
6591665 c.1134G>A p.E378E Chr.3: 128904490-128904490 on Build GRCh38
4422829 c.1136C>T p.A379V Chr.3: 128904492-128904492 on Build GRCh38
233856 c.1149+6C>T p.? Chr.3: 128904511-128904511 on Build GRCh38
7426670 c.1045A>T p.M349L Chr.3: 128904401-128904401 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977474706 Chr.3: 128904560-128904560 on Build GRCh38
rs375363759 Chr.3: 128904488-128904488 on Build GRCh38
rs1010789570 Chr.3: 128904738-128904739 on Build GRCh38
rs978405425 Chr.3: 128904744-128904744 on Build GRCh38
rs967381006 Chr.3: 128904767-128904767 on Build GRCh38
rs1016201968 Chr.3: 128904328-128904328 on Build GRCh38
rs532422170 Chr.3: 128904534-128904534 on Build GRCh38
rs942292299 Chr.3: 128904597-128904597 on Build GRCh38
rs775279600 Chr.3: 128904693-128904693 on Build GRCh38
rs748724880 Chr.3: 128904369-128904369 on Build GRCh38
rs768270636 Chr.3: 128904382-128904382 on Build GRCh38
rs993673297 Chr.3: 128904646-128904646 on Build GRCh38
rs777000518 Chr.3: 128904398-128904398 on Build GRCh38
rs778498182 Chr.3: 128904521-128904521 on Build GRCh38
rs769809288 Chr.3: 128904407-128904407 on Build GRCh38
rs916198389 Chr.3: 128904586-128904586 on Build GRCh38
rs751398822 Chr.3: 128904476-128904476 on Build GRCh38
rs1007381928 Chr.3: 128904685-128904685 on Build GRCh38
rs758166180 Chr.3: 128904379-128904379 on Build GRCh38
rs143332279 Chr.3: 128904624-128904624 on Build GRCh38
rs73210609 Chr.3: 128904562-128904562 on Build GRCh38
rs758407530 Chr.3: 128904444-128904444 on Build GRCh38
rs370866774 Chr.3: 128904533-128904533 on Build GRCh38
rs766670497 Chr.3: 128904392-128904392 on Build GRCh38
rs771347620 Chr.3: 128904657-128904657 on Build GRCh38
rs751735658 Chr.3: 128904636-128904637 on Build GRCh38
rs938853431 Chr.3: 128904563-128904563 on Build GRCh38
rs968369816 Chr.3: 128904491-128904491 on Build GRCh38
rs183349576 Chr.3: 128904735-128904735 on Build GRCh38
rs756022677 Chr.3: 128904496-128904496 on Build GRCh38
rs1017503875 Chr.3: 128904490-128904490 on Build GRCh38
rs367985938 Chr.3: 128904510-128904510 on Build GRCh38
rs778849943 Chr.3: 128904333-128904333 on Build GRCh38
rs761173018 Chr.3: 128904391-128904391 on Build GRCh38
rs770264599 Chr.3: 128904557-128904557 on Build GRCh38
rs897743749 Chr.3: 128904640-128904640 on Build GRCh38
rs759711229 Chr.3: 128904412-128904412 on Build GRCh38
rs1022688332 Chr.3: 128904726-128904726 on Build GRCh38
rs960091334 Chr.3: 128904414-128904414 on Build GRCh38
rs888503390 Chr.3: 128904582-128904582 on Build GRCh38
rs771983419 Chr.3: 128904691-128904691 on Build GRCh38
rs765411526 Chr.3: 128904425-128904425 on Build GRCh38
rs954523101 Chr.3: 128904338-128904338 on Build GRCh38
rs1035970845 Chr.3: 128904663-128904663 on Build GRCh38
rs762968752 Chr.3: 128904627-128904627 on Build GRCh38
rs781096699 Chr.3: 128904487-128904487 on Build GRCh38
rs886057957 Chr.3: 128904418-128904418 on Build GRCh38
rs777159838 Chr.3: 128904551-128904551 on Build GRCh38
rs569363607 Chr.3: 128904692-128904692 on Build GRCh38
rs139008397 Chr.3: 128904556-128904556 on Build GRCh38
rs537254892 Chr.3: 128904713-128904713 on Build GRCh38
rs764240652 Chr.3: 128904456-128904456 on Build GRCh38
rs1006170291 Chr.3: 128904326-128904326 on Build GRCh38
rs570831726 Chr.3: 128904746-128904746 on Build GRCh38
rs779559482 Chr.3: 128904497-128904497 on Build GRCh38
rs991194840 Chr.3: 128904475-128904475 on Build GRCh38
rs35316785 Chr.3: 128904451-128904452 on Build GRCh38
rs959881222 Chr.3: 128904681-128904681 on Build GRCh38
rs773586510 Chr.3: 128904385-128904385 on Build GRCh38
rs745683893 Chr.3: 128904347-128904347 on Build GRCh38
rs890191113 Chr.3: 128904670-128904670 on Build GRCh38
rs753025393 Chr.3: 128904435-128904435 on Build GRCh38
rs749036538 Chr.3: 128904502-128904502 on Build GRCh38
rs745470957 Chr.3: 128904345-128904345 on Build GRCh38
rs1005573332 Chr.3: 128904608-128904608 on Build GRCh38
rs190101630 Chr.3: 128904581-128904581 on Build GRCh38
rs961964320 Chr.3: 128904335-128904335 on Build GRCh38
rs932752961 Chr.3: 128904579-128904579 on Build GRCh38

View Full Product Details