Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129167687-129168189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNBP
Entrez Gene ID
7555
Gene Name
CCHC-type zinc finger nucleic acid binding protein
Gene Aliases
CNBP1, DM2, PROMM, RNF163, ZCCHC22, ZNF9
UniGene
Hs.518249
Gene Chromosome Location
Chr.3: 129167815-129183967 on Build GRCh38
Transcripts
NM_001127192.1 NM_001127193.1 NM_001127194.1 NM_001127195.1 NM_001127196.1 NM_003418.4

SNP Detail

SNP ID Chromosome Location
rs567026320 Chr.3: 129167986-129167986 on Build GRCh38
rs746829442 Chr.3: 129167811-129167811 on Build GRCh38
rs1010621077 Chr.3: 129167984-129167984 on Build GRCh38
rs979851989 Chr.3: 129167857-129167858 on Build GRCh38
rs1021134838 Chr.3: 129168047-129168047 on Build GRCh38
rs776358032 Chr.3: 129168028-129168029 on Build GRCh38
rs984195975 Chr.3: 129168160-129168160 on Build GRCh38
rs975817653 Chr.3: 129168142-129168142 on Build GRCh38
rs145251732 Chr.3: 129168153-129168153 on Build GRCh38
rs770963173 Chr.3: 129167962-129167962 on Build GRCh38
rs565429431 Chr.3: 129168159-129168159 on Build GRCh38
rs557809721 Chr.3: 129168105-129168105 on Build GRCh38
rs944002055 Chr.3: 129167783-129167783 on Build GRCh38
rs966541870 Chr.3: 129167774-129167774 on Build GRCh38
rs774458329 Chr.3: 129168028-129168028 on Build GRCh38
rs904830612 Chr.3: 129167765-129167765 on Build GRCh38
rs531678707 Chr.3: 129167887-129167887 on Build GRCh38
rs779926793 Chr.3: 129167772-129167772 on Build GRCh38
rs906821597 Chr.3: 129167893-129167893 on Build GRCh38
rs1033832872 Chr.3: 129167744-129167744 on Build GRCh38
rs556480605 Chr.3: 129167940-129167940 on Build GRCh38
rs540459900 Chr.3: 129168122-129168122 on Build GRCh38
rs149706654 Chr.3: 129168076-129168076 on Build GRCh38
rs1004566723 Chr.3: 129168016-129168016 on Build GRCh38
rs949543584 Chr.3: 129167938-129167938 on Build GRCh38
rs982632910 Chr.3: 129167783-129167785 on Build GRCh38
rs1004353078 Chr.3: 129167869-129167869 on Build GRCh38
rs562572797 Chr.3: 129167819-129167819 on Build GRCh38
rs956962949 Chr.3: 129168158-129168159 on Build GRCh38
rs749510564 Chr.3: 129167908-129167908 on Build GRCh38
rs777327279 Chr.3: 129167844-129167846 on Build GRCh38
rs545118050 Chr.3: 129168141-129168141 on Build GRCh38
rs577867734 Chr.3: 129168080-129168080 on Build GRCh38
rs996640530 Chr.3: 129168112-129168112 on Build GRCh38
rs150014526 Chr.3: 129167764-129167764 on Build GRCh38
rs995148724 Chr.3: 129168008-129168008 on Build GRCh38
rs987326230 Chr.3: 129168113-129168113 on Build GRCh38
rs953342693 Chr.3: 129167848-129167848 on Build GRCh38
rs58395398 Chr.3: 129168031-129168032 on Build GRCh38
rs573636612 Chr.3: 129167929-129167929 on Build GRCh38
rs763905667 Chr.3: 129167994-129167994 on Build GRCh38
rs115036251 Chr.3: 129167932-129167932 on Build GRCh38
rs760171976 Chr.3: 129168128-129168129 on Build GRCh38
rs571556654 Chr.3: 129168036-129168036 on Build GRCh38
rs1048575686 Chr.3: 129168010-129168010 on Build GRCh38
rs887245110 Chr.3: 129167858-129167858 on Build GRCh38
rs118160527 Chr.3: 129167781-129167781 on Build GRCh38
rs768384094 Chr.3: 129167817-129167817 on Build GRCh38
rs777863307 Chr.3: 129167846-129167846 on Build GRCh38
rs913696175 Chr.3: 129167771-129167771 on Build GRCh38
rs1044812323 Chr.3: 129167943-129167943 on Build GRCh38
rs921341180 Chr.3: 129167786-129167786 on Build GRCh38
rs78922466 Chr.3: 129167712-129167712 on Build GRCh38
rs957843993 Chr.3: 129167753-129167753 on Build GRCh38
rs1029002355 Chr.3: 129168131-129168131 on Build GRCh38
rs887013776 Chr.3: 129168011-129168011 on Build GRCh38
rs976111018 Chr.3: 129167845-129167845 on Build GRCh38
rs989714892 Chr.3: 129167769-129167769 on Build GRCh38
rs911829346 Chr.3: 129167782-129167782 on Build GRCh38
rs988768621 Chr.3: 129168159-129168159 on Build GRCh38
rs139580350 Chr.3: 129167881-129167881 on Build GRCh38
rs904945058 Chr.3: 129167968-129167968 on Build GRCh38
rs918117018 Chr.3: 129167935-129167935 on Build GRCh38
rs186990300 Chr.3: 129167856-129167856 on Build GRCh38
rs192379625 Chr.3: 129167928-129167928 on Build GRCh38
rs1039738006 Chr.3: 129167785-129167785 on Build GRCh38
rs1133046 Chr.3: 129167850-129167850 on Build GRCh38
rs573060132 Chr.3: 129168118-129168118 on Build GRCh38
rs542611107 Chr.3: 129167775-129167775 on Build GRCh38

View Full Product Details