Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129168178-129168683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNBP
Entrez Gene ID
7555
Gene Name
CCHC-type zinc finger nucleic acid binding protein
Gene Aliases
CNBP1, DM2, PROMM, RNF163, ZCCHC22, ZNF9
UniGene
Hs.518249
Gene Chromosome Location
Chr.3: 129167815-129183967 on Build GRCh38
Transcripts
NM_001127192.1 NM_001127193.1 NM_001127194.1 NM_001127195.1 NM_001127196.1 NM_003418.4

SNP Detail

SNP ID Chromosome Location
rs956896845 Chr.3: 129168355-129168355 on Build GRCh38
rs901092510 Chr.3: 129168543-129168543 on Build GRCh38
rs764698140 Chr.3: 129168491-129168491 on Build GRCh38
rs545673318 Chr.3: 129168283-129168283 on Build GRCh38
rs1055792562 Chr.3: 129168243-129168243 on Build GRCh38
rs1048047174 Chr.3: 129168230-129168235 on Build GRCh38
rs375592071 Chr.3: 129168527-129168527 on Build GRCh38
rs57560229 Chr.3: 129168629-129168629 on Build GRCh38
rs190011929 Chr.3: 129168552-129168552 on Build GRCh38
rs761506718 Chr.3: 129168455-129168455 on Build GRCh38
rs764876598 Chr.3: 129168497-129168497 on Build GRCh38
rs982618313 Chr.3: 129168532-129168532 on Build GRCh38
rs772724600 Chr.3: 129168219-129168219 on Build GRCh38
rs749984795 Chr.3: 129168495-129168495 on Build GRCh38
rs566744187 Chr.3: 129168632-129168632 on Build GRCh38
rs931079489 Chr.3: 129168206-129168206 on Build GRCh38
rs550056735 Chr.3: 129168533-129168533 on Build GRCh38
rs1001384790 Chr.3: 129168324-129168326 on Build GRCh38
rs938236146 Chr.3: 129168242-129168242 on Build GRCh38
rs1020697528 Chr.3: 129168339-129168339 on Build GRCh38
rs200193915 Chr.3: 129168645-129168645 on Build GRCh38
rs34121423 Chr.3: 129168547-129168547 on Build GRCh38
rs34453888 Chr.3: 129168541-129168541 on Build GRCh38
rs764546386 Chr.3: 129168391-129168391 on Build GRCh38
rs185205817 Chr.3: 129168545-129168545 on Build GRCh38
rs988160809 Chr.3: 129168356-129168356 on Build GRCh38
rs149175255 Chr.3: 129168207-129168207 on Build GRCh38
rs886747476 Chr.3: 129168244-129168244 on Build GRCh38
rs9880375 Chr.3: 129168304-129168304 on Build GRCh38
rs56763410 Chr.3: 129168204-129168204 on Build GRCh38
rs980917581 Chr.3: 129168393-129168393 on Build GRCh38
rs1003902542 Chr.3: 129168247-129168247 on Build GRCh38
rs528001881 Chr.3: 129168305-129168305 on Build GRCh38
rs112606287 Chr.3: 129168595-129168595 on Build GRCh38
rs35661727 Chr.3: 129168520-129168520 on Build GRCh38
rs1025197731 Chr.3: 129168376-129168376 on Build GRCh38
rs1040723460 Chr.3: 129168539-129168539 on Build GRCh38
rs1008727561 Chr.3: 129168407-129168407 on Build GRCh38
rs939650919 Chr.3: 129168534-129168534 on Build GRCh38
rs901108651 Chr.3: 129168280-129168280 on Build GRCh38
rs1032439791 Chr.3: 129168350-129168350 on Build GRCh38
rs145051505 Chr.3: 129168438-129168438 on Build GRCh38
rs1040901934 Chr.3: 129168273-129168273 on Build GRCh38
rs1028149959 Chr.3: 129168286-129168286 on Build GRCh38
rs1046579928 Chr.3: 129168205-129168205 on Build GRCh38
rs1018830744 Chr.3: 129168428-129168428 on Build GRCh38

View Full Product Details