Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 131555416-131555910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE4
Entrez Gene ID
131034
Gene Name
copine 4
Gene Aliases
COPN4, CPN4
UniGene
Hs.199877
Gene Chromosome Location
Chr.3: 131533560-132285702 on Build GRCh38
Transcripts
NM_001289112.1 NM_130808.2 NM_153429.1 XM_011512406.2 XM_011512407.2 XM_011512408.2 XM_017005693.1 XM_017005694.1 XM_017005695.1 XM_017005696.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5918040 c.1062-8G>A p.? Chr.3: 131555559-131555559 on Build GRCh38
3587521 c.1075C>T p.P359S Chr.3: 131555538-131555538 on Build GRCh38
3587519 c.1106C>T p.P369L Chr.3: 131555507-131555507 on Build GRCh38
7413502 c.1090G>T p.G364C Chr.3: 131555523-131555523 on Build GRCh38
3587517 c.1116+1G>A p.? Chr.3: 131555496-131555496 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891073806 Chr.3: 131555822-131555822 on Build GRCh38
rs569164515 Chr.3: 131555723-131555723 on Build GRCh38
rs370033923 Chr.3: 131555520-131555520 on Build GRCh38
rs905135407 Chr.3: 131555740-131555740 on Build GRCh38
rs752813662 Chr.3: 131555506-131555506 on Build GRCh38
rs769510328 Chr.3: 131555456-131555456 on Build GRCh38
rs776792296 Chr.3: 131555497-131555497 on Build GRCh38
rs141966606 Chr.3: 131555557-131555557 on Build GRCh38
rs1046476219 Chr.3: 131555731-131555731 on Build GRCh38
rs758663472 Chr.3: 131555766-131555766 on Build GRCh38
rs539754047 Chr.3: 131555773-131555773 on Build GRCh38
rs942145695 Chr.3: 131555716-131555716 on Build GRCh38
rs773694748 Chr.3: 131555578-131555578 on Build GRCh38
rs115245241 Chr.3: 131555461-131555461 on Build GRCh38
rs765542131 Chr.3: 131555509-131555509 on Build GRCh38
rs748852334 Chr.3: 131555574-131555574 on Build GRCh38
rs745650287 Chr.3: 131555469-131555471 on Build GRCh38
rs529063926 Chr.3: 131555675-131555675 on Build GRCh38
rs773746499 Chr.3: 131555473-131555473 on Build GRCh38
rs369567326 Chr.3: 131555558-131555558 on Build GRCh38
rs759813686 Chr.3: 131555499-131555499 on Build GRCh38
rs1046766912 Chr.3: 131555627-131555627 on Build GRCh38
rs533619402 Chr.3: 131555878-131555878 on Build GRCh38
rs937486000 Chr.3: 131555657-131555657 on Build GRCh38
rs758451266 Chr.3: 131555508-131555508 on Build GRCh38
rs750867772 Chr.3: 131555575-131555575 on Build GRCh38
rs374112978 Chr.3: 131555521-131555521 on Build GRCh38
rs146317922 Chr.3: 131555632-131555632 on Build GRCh38
rs79264071 Chr.3: 131555821-131555821 on Build GRCh38
rs375925943 Chr.3: 131555485-131555485 on Build GRCh38
rs897815805 Chr.3: 131555770-131555770 on Build GRCh38
rs575822238 Chr.3: 131555733-131555734 on Build GRCh38
rs1021090183 Chr.3: 131555847-131555847 on Build GRCh38
rs528518281 Chr.3: 131555583-131555583 on Build GRCh38
rs779816948 Chr.3: 131555573-131555573 on Build GRCh38
rs965318571 Chr.3: 131555563-131555563 on Build GRCh38
rs781226530 Chr.3: 131555578-131555580 on Build GRCh38
rs757078596 Chr.3: 131555548-131555548 on Build GRCh38
rs189523375 Chr.3: 131555708-131555708 on Build GRCh38
rs1002511834 Chr.3: 131555743-131555743 on Build GRCh38
rs370639886 Chr.3: 131555595-131555595 on Build GRCh38
rs57687010 Chr.3: 131555785-131555785 on Build GRCh38
rs111311927 Chr.3: 131555785-131555785 on Build GRCh38
rs1036742908 Chr.3: 131555730-131555730 on Build GRCh38
rs990201782 Chr.3: 131555564-131555564 on Build GRCh38
rs945723905 Chr.3: 131555587-131555587 on Build GRCh38
rs887819250 Chr.3: 131555713-131555713 on Build GRCh38
rs369001089 Chr.3: 131555455-131555455 on Build GRCh38
rs386665905 Chr.3: 131555461-131555463 on Build GRCh38
rs775194936 Chr.3: 131555460-131555460 on Build GRCh38
rs761038808 Chr.3: 131555480-131555480 on Build GRCh38
rs755895556 Chr.3: 131555562-131555562 on Build GRCh38
rs771283890 Chr.3: 131555599-131555599 on Build GRCh38
rs954951942 Chr.3: 131555854-131555854 on Build GRCh38
rs765442416 Chr.3: 131555500-131555500 on Build GRCh38
rs957148283 Chr.3: 131555528-131555528 on Build GRCh38
rs755251491 Chr.3: 131555568-131555568 on Build GRCh38
rs574940932 Chr.3: 131555664-131555664 on Build GRCh38
rs185151792 Chr.3: 131555476-131555476 on Build GRCh38

View Full Product Details