Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 132599795-132600319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAD11
Entrez Gene ID
84129
Gene Name
acyl-CoA dehydrogenase family member 11
Gene Aliases
ACAD-11
UniGene
Hs.441378
Gene Chromosome Location
Chr.3: 132558138-132660131 on Build GRCh38
Transcripts
NM_032169.4 NR_132426.1 NR_132427.1 NR_132428.1 NM_016557.3 NM_178445.2 NR_037804.1

Gene Symbol
ACKR4
Entrez Gene ID
51554
Gene Name
atypical chemokine receptor 4
Gene Aliases
CC-CKR-11, CCBP2, CCR-11, CCR10, CCR11, CCRL1, CCX CKR, CCX-CKR, CKR-11, PPR1, VSHK1
UniGene
Hs.310512
Gene Chromosome Location
Chr.3: 132597237-132602644 on Build GRCh38
Transcripts
NM_032169.4 NR_132426.1 NR_132427.1 NR_132428.1 NM_016557.3 NM_178445.2 NR_037804.1

Gene Symbol
NPHP3-ACAD11
Entrez Gene ID
100532724
Gene Name
NPHP3-ACAD11 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.511991
Gene Chromosome Location
Chr.3: 132558138-132722459 on Build GRCh38
Transcripts
NM_032169.4 NR_132426.1 NR_132427.1 NR_132428.1 NM_016557.3 NM_178445.2 NR_037804.1

SNP Detail

SNP ID Chromosome Location
rs963789045 Chr.3: 132600080-132600080 on Build GRCh38
rs940711885 Chr.3: 132599890-132599890 on Build GRCh38
rs114413101 Chr.3: 132599839-132599839 on Build GRCh38
rs1050511878 Chr.3: 132600006-132600006 on Build GRCh38
rs1002305250 Chr.3: 132600254-132600254 on Build GRCh38
rs937477387 Chr.3: 132599953-132599953 on Build GRCh38
rs554643810 Chr.3: 132600075-132600075 on Build GRCh38
rs565084078 Chr.3: 132600295-132600295 on Build GRCh38
rs902823449 Chr.3: 132599997-132599997 on Build GRCh38
rs892464810 Chr.3: 132600074-132600074 on Build GRCh38
rs572292281 Chr.3: 132600030-132600030 on Build GRCh38
rs543904253 Chr.3: 132600294-132600294 on Build GRCh38
rs187380315 Chr.3: 132599988-132599988 on Build GRCh38
rs767602608 Chr.3: 132600045-132600045 on Build GRCh38
rs574430457 Chr.3: 132600227-132600227 on Build GRCh38
rs908473851 Chr.3: 132600271-132600271 on Build GRCh38
rs543271492 Chr.3: 132599977-132599982 on Build GRCh38
rs768129807 Chr.3: 132600185-132600185 on Build GRCh38
rs756318686 Chr.3: 132600088-132600088 on Build GRCh38
rs1011769941 Chr.3: 132599985-132599985 on Build GRCh38
rs1027091624 Chr.3: 132600257-132600257 on Build GRCh38
rs755113190 Chr.3: 132600023-132600026 on Build GRCh38
rs907294849 Chr.3: 132599938-132599938 on Build GRCh38
rs1045794898 Chr.3: 132599907-132599907 on Build GRCh38
rs903914019 Chr.3: 132600043-132600043 on Build GRCh38
rs982702990 Chr.3: 132600269-132600269 on Build GRCh38
rs181930157 Chr.3: 132599954-132599954 on Build GRCh38
rs996640286 Chr.3: 132600093-132600093 on Build GRCh38
rs982733213 Chr.3: 132599827-132599827 on Build GRCh38
rs3804636 Chr.3: 132600053-132600053 on Build GRCh38
rs908243151 Chr.3: 132600282-132600282 on Build GRCh38
rs893339561 Chr.3: 132599965-132599965 on Build GRCh38
rs1026812655 Chr.3: 132600221-132600221 on Build GRCh38
rs1025192148 Chr.3: 132600146-132600146 on Build GRCh38
rs1014258563 Chr.3: 132600129-132600129 on Build GRCh38
rs908188685 Chr.3: 132599847-132599847 on Build GRCh38

View Full Product Details