Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 134477576-134478107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC13
Entrez Gene ID
25847
Gene Name
anaphase promoting complex subunit 13
Gene Aliases
APC13, SWM1
UniGene
Hs.106909
Gene Chromosome Location
Chr.3: 134477704-134486023 on Build GRCh38
Transcripts
NM_001242374.1 NM_001242375.1 NM_015391.3

SNP Detail

SNP ID Chromosome Location
rs3181005 Chr.3: 134477705-134477705 on Build GRCh38
rs563874669 Chr.3: 134478071-134478071 on Build GRCh38
rs991370687 Chr.3: 134477806-134477806 on Build GRCh38
rs1053389777 Chr.3: 134477723-134477723 on Build GRCh38
rs917436587 Chr.3: 134477601-134477601 on Build GRCh38
rs773884422 Chr.3: 134477926-134477926 on Build GRCh38
rs1048818619 Chr.3: 134477871-134477871 on Build GRCh38
rs776984222 Chr.3: 134478006-134478006 on Build GRCh38
rs755553613 Chr.3: 134478026-134478026 on Build GRCh38
rs977597537 Chr.3: 134477616-134477616 on Build GRCh38
rs1051980647 Chr.3: 134477783-134477783 on Build GRCh38
rs5016172 Chr.3: 134477905-134477905 on Build GRCh38
rs931748058 Chr.3: 134477760-134477760 on Build GRCh38
rs374109861 Chr.3: 134477604-134477604 on Build GRCh38
rs554203460 Chr.3: 134477726-134477726 on Build GRCh38
rs1036191035 Chr.3: 134477816-134477816 on Build GRCh38
rs1024832899 Chr.3: 134477939-134477939 on Build GRCh38
rs936247509 Chr.3: 134477668-134477668 on Build GRCh38
rs978025022 Chr.3: 134477969-134477969 on Build GRCh38
rs529707287 Chr.3: 134477915-134477915 on Build GRCh38
rs574289504 Chr.3: 134477749-134477749 on Build GRCh38
rs977239267 Chr.3: 134477755-134477755 on Build GRCh38
rs1021402510 Chr.3: 134478020-134478022 on Build GRCh38
rs772425982 Chr.3: 134477896-134477896 on Build GRCh38
rs758415712 Chr.3: 134477753-134477753 on Build GRCh38
rs141870909 Chr.3: 134478014-134478014 on Build GRCh38
rs899089661 Chr.3: 134477857-134477857 on Build GRCh38
rs897561761 Chr.3: 134477834-134477834 on Build GRCh38
rs915849561 Chr.3: 134477810-134477810 on Build GRCh38
rs1045833257 Chr.3: 134477820-134477820 on Build GRCh38
rs540657367 Chr.3: 134477672-134477672 on Build GRCh38
rs139092859 Chr.3: 134477923-134477923 on Build GRCh38
rs966713805 Chr.3: 134478043-134478043 on Build GRCh38
rs887904714 Chr.3: 134477894-134477894 on Build GRCh38
rs921505940 Chr.3: 134478018-134478018 on Build GRCh38
rs967484207 Chr.3: 134477960-134477960 on Build GRCh38
rs531908053 Chr.3: 134477815-134477815 on Build GRCh38
rs543262396 Chr.3: 134477787-134477787 on Build GRCh38
rs180863601 Chr.3: 134477619-134477619 on Build GRCh38
rs562082425 Chr.3: 134477794-134477794 on Build GRCh38
rs1021033968 Chr.3: 134477697-134477697 on Build GRCh38
rs369403384 Chr.3: 134477851-134477851 on Build GRCh38
rs975610163 Chr.3: 134478012-134478012 on Build GRCh38
rs3178220 Chr.3: 134477706-134477706 on Build GRCh38
rs761069990 Chr.3: 134478028-134478028 on Build GRCh38
rs994717817 Chr.3: 134477887-134477887 on Build GRCh38
rs1033144679 Chr.3: 134477979-134477979 on Build GRCh38

View Full Product Details