Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111319009-111319546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHIA
Entrez Gene ID
27159
Gene Name
chitinase, acidic
Gene Aliases
AMCASE, CHIT2, TSA1902
UniGene
Hs.128814
Gene Chromosome Location
Chr.1: 111290852-111320566 on Build GRCh38
Transcripts
NM_001040623.2 NM_001258001.1 NM_001258002.1 NM_001258003.1 NM_001258004.1 NM_001258005.1 NM_021797.3 NM_201653.3 XM_006710577.3 XM_017001047.1 XM_017001048.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5421584 c.1162G>A p.G388S Chr.1: 111319453-111319453 on Build GRCh38
1688264 c.1087G>A p.D363N Chr.1: 111319378-111319378 on Build GRCh38
6507620 c.949G>A p.G317R Chr.1: 111319153-111319153 on Build GRCh38
1179735 c.691A>G p.I231V Chr.1: 111319219-111319219 on Build GRCh38
3721241 c.1068C>T p.G356G Chr.1: 111319359-111319359 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770515340 Chr.1: 111319263-111319263 on Build GRCh38
rs139812869 Chr.1: 111319154-111319154 on Build GRCh38
rs371701062 Chr.1: 111319453-111319453 on Build GRCh38
rs374315938 Chr.1: 111319361-111319361 on Build GRCh38
rs76395847 Chr.1: 111319404-111319404 on Build GRCh38
rs749956799 Chr.1: 111319495-111319495 on Build GRCh38
rs201691527 Chr.1: 111319360-111319360 on Build GRCh38
rs559458629 Chr.1: 111319488-111319488 on Build GRCh38
rs141843885 Chr.1: 111319388-111319388 on Build GRCh38
rs373985805 Chr.1: 111319373-111319373 on Build GRCh38
rs756257682 Chr.1: 111319204-111319204 on Build GRCh38
rs1011836963 Chr.1: 111319247-111319247 on Build GRCh38
rs750498281 Chr.1: 111319286-111319286 on Build GRCh38
rs772979534 Chr.1: 111319165-111319165 on Build GRCh38
rs190772785 Chr.1: 111319201-111319201 on Build GRCh38
rs2820092 Chr.1: 111319452-111319452 on Build GRCh38
rs762724976 Chr.1: 111319481-111319481 on Build GRCh38
rs2275253 Chr.1: 111319219-111319219 on Build GRCh38
rs751501173 Chr.1: 111319489-111319489 on Build GRCh38
rs781398038 Chr.1: 111319100-111319100 on Build GRCh38
rs142800191 Chr.1: 111319363-111319363 on Build GRCh38
rs751905455 Chr.1: 111319065-111319065 on Build GRCh38
rs1030825897 Chr.1: 111319308-111319308 on Build GRCh38
rs551555095 Chr.1: 111319507-111319507 on Build GRCh38
rs749286401 Chr.1: 111319141-111319141 on Build GRCh38
rs748471594 Chr.1: 111319448-111319448 on Build GRCh38
rs142736771 Chr.1: 111319127-111319127 on Build GRCh38
rs764439541 Chr.1: 111319270-111319270 on Build GRCh38
rs12046235 Chr.1: 111319276-111319276 on Build GRCh38
rs542694997 Chr.1: 111319458-111319458 on Build GRCh38
rs541197537 Chr.1: 111319407-111319407 on Build GRCh38
rs761418105 Chr.1: 111319159-111319159 on Build GRCh38
rs778215287 Chr.1: 111319081-111319081 on Build GRCh38
rs771636785 Chr.1: 111319160-111319160 on Build GRCh38
rs2786161 Chr.1: 111319092-111319092 on Build GRCh38
rs931946676 Chr.1: 111319074-111319074 on Build GRCh38
rs368203516 Chr.1: 111319434-111319434 on Build GRCh38
rs557915320 Chr.1: 111319278-111319278 on Build GRCh38
rs754884356 Chr.1: 111319313-111319313 on Build GRCh38
rs151326722 Chr.1: 111319160-111319161 on Build GRCh38
rs12094378 Chr.1: 111319456-111319456 on Build GRCh38
rs758302367 Chr.1: 111319328-111319328 on Build GRCh38
rs374240015 Chr.1: 111319319-111319319 on Build GRCh38
rs2275254 Chr.1: 111319352-111319352 on Build GRCh38
rs375947945 Chr.1: 111319198-111319198 on Build GRCh38
rs1024854905 Chr.1: 111319253-111319253 on Build GRCh38
rs374270214 Chr.1: 111319234-111319234 on Build GRCh38
rs767673479 Chr.1: 111319283-111319283 on Build GRCh38
rs189146821 Chr.1: 111319226-111319226 on Build GRCh38
rs756776071 Chr.1: 111319425-111319425 on Build GRCh38
rs773707736 Chr.1: 111319389-111319389 on Build GRCh38
rs766521863 Chr.1: 111319306-111319306 on Build GRCh38
rs752923770 Chr.1: 111319174-111319175 on Build GRCh38
rs970578190 Chr.1: 111319268-111319268 on Build GRCh38
rs868796007 Chr.1: 111319175-111319175 on Build GRCh38
rs763257100 Chr.1: 111319182-111319182 on Build GRCh38
rs147847412 Chr.1: 111319119-111319119 on Build GRCh38
rs776886953 Chr.1: 111319455-111319455 on Build GRCh38
rs759836972 Chr.1: 111319138-111319138 on Build GRCh38
rs762285434 Chr.1: 111319279-111319279 on Build GRCh38
rs36011905 Chr.1: 111319420-111319420 on Build GRCh38
rs754167131 Chr.1: 111319402-111319402 on Build GRCh38
rs751662371 Chr.1: 111319089-111319089 on Build GRCh38
rs140972112 Chr.1: 111319512-111319512 on Build GRCh38
rs528976050 Chr.1: 111319199-111319199 on Build GRCh38
rs962596727 Chr.1: 111319224-111319224 on Build GRCh38
rs144546962 Chr.1: 111319197-111319197 on Build GRCh38
rs114909116 Chr.1: 111319514-111319514 on Build GRCh38
rs144182515 Chr.1: 111319203-111319203 on Build GRCh38
rs748089793 Chr.1: 111319232-111319232 on Build GRCh38
rs779690824 Chr.1: 111319132-111319132 on Build GRCh38
rs765520051 Chr.1: 111319405-111319405 on Build GRCh38
rs765676898 Chr.1: 111319177-111319177 on Build GRCh38
rs374884131 Chr.1: 111319490-111319490 on Build GRCh38
rs151038298 Chr.1: 111319359-111319359 on Build GRCh38
rs746515568 Chr.1: 111319347-111319347 on Build GRCh38
rs912188371 Chr.1: 111319366-111319366 on Build GRCh38
rs972283710 Chr.1: 111319231-111319231 on Build GRCh38
rs756347348 Chr.1: 111319191-111319191 on Build GRCh38
rs745560571 Chr.1: 111319439-111319439 on Build GRCh38
rs776291802 Chr.1: 111319264-111319264 on Build GRCh38
rs777577154 Chr.1: 111319419-111319419 on Build GRCh38
rs776333174 Chr.1: 111319353-111319353 on Build GRCh38
rs867998770 Chr.1: 111319364-111319364 on Build GRCh38
rs938835178 Chr.1: 111319299-111319299 on Build GRCh38
rs147768976 Chr.1: 111319447-111319447 on Build GRCh38
rs760196175 Chr.1: 111319260-111319260 on Build GRCh38
rs182916944 Chr.1: 111319218-111319218 on Build GRCh38
rs781067650 Chr.1: 111319494-111319494 on Build GRCh38
rs921942849 Chr.1: 111319034-111319034 on Build GRCh38
rs777720579 Chr.1: 111319333-111319333 on Build GRCh38
rs200566957 Chr.1: 111319162-111319162 on Build GRCh38
rs556480396 Chr.1: 111319288-111319288 on Build GRCh38
rs758157855 Chr.1: 111319413-111319413 on Build GRCh38
rs777884168 Chr.1: 111319348-111319348 on Build GRCh38
rs1048154802 Chr.1: 111319454-111319454 on Build GRCh38
rs751320088 Chr.1: 111319087-111319087 on Build GRCh38
rs748140882 Chr.1: 111319327-111319327 on Build GRCh38
rs772568916 Chr.1: 111319252-111319252 on Build GRCh38
rs764599211 Chr.1: 111319192-111319192 on Build GRCh38
rs766472526 Chr.1: 111319395-111319395 on Build GRCh38
rs998727555 Chr.1: 111319078-111319078 on Build GRCh38
rs370020615 Chr.1: 111319212-111319212 on Build GRCh38
rs760673420 Chr.1: 111319394-111319394 on Build GRCh38
rs578164623 Chr.1: 111319433-111319433 on Build GRCh38
rs200092579 Chr.1: 111319230-111319230 on Build GRCh38
rs779656421 Chr.1: 111319473-111319473 on Build GRCh38
rs752901913 Chr.1: 111319479-111319479 on Build GRCh38
rs115321756 Chr.1: 111319311-111319311 on Build GRCh38
rs772412644 Chr.1: 111319449-111319449 on Build GRCh38
rs768742033 Chr.1: 111319152-111319152 on Build GRCh38
rs143969273 Chr.1: 111319442-111319442 on Build GRCh38
rs760226016 Chr.1: 111319172-111319172 on Build GRCh38
rs17027410 Chr.1: 111319200-111319200 on Build GRCh38
rs753037237 Chr.1: 111319181-111319181 on Build GRCh38
rs763313855 Chr.1: 111319265-111319265 on Build GRCh38
rs375881947 Chr.1: 111319178-111319179 on Build GRCh38

View Full Product Details