Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111320124-111320625 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHIA
Entrez Gene ID
27159
Gene Name
chitinase, acidic
Gene Aliases
AMCASE, CHIT2, TSA1902
UniGene
Hs.128814
Gene Chromosome Location
Chr.1: 111290852-111320566 on Build GRCh38
Transcripts
NM_001040623.2 NM_001258001.1 NM_001258002.1 NM_001258003.1 NM_001258004.1 NM_001258005.1 NM_021797.3 NM_201653.3 XM_006710577.3 XM_017001047.1 XM_017001048.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4524611 c.958G>A p.G320R Chr.1: 111320317-111320317 on Build GRCh38
358187 c.1382A>T p.Q461L Chr.1: 111320417-111320417 on Build GRCh38
3764094 c.1341C>A p.F447L Chr.1: 111320376-111320376 on Build GRCh38
4672157 c.948G>A p.S316S Chr.1: 111320307-111320307 on Build GRCh38
3471194 c.950G>A p.G317E Chr.1: 111320309-111320309 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545102298 Chr.1: 111320386-111320386 on Build GRCh38
rs752790899 Chr.1: 111320477-111320477 on Build GRCh38
rs771358563 Chr.1: 111320258-111320258 on Build GRCh38
rs763851350 Chr.1: 111320434-111320434 on Build GRCh38
rs544506609 Chr.1: 111320245-111320245 on Build GRCh38
rs915196627 Chr.1: 111320199-111320199 on Build GRCh38
rs759156078 Chr.1: 111320371-111320371 on Build GRCh38
rs780819269 Chr.1: 111320491-111320491 on Build GRCh38
rs750838913 Chr.1: 111320387-111320387 on Build GRCh38
rs1029407341 Chr.1: 111320545-111320545 on Build GRCh38
rs773188690 Chr.1: 111320282-111320282 on Build GRCh38
rs772568990 Chr.1: 111320325-111320360 on Build GRCh38
rs142653250 Chr.1: 111320422-111320422 on Build GRCh38
rs941237426 Chr.1: 111320269-111320269 on Build GRCh38
rs985352253 Chr.1: 111320231-111320231 on Build GRCh38
rs116217812 Chr.1: 111320197-111320197 on Build GRCh38
rs777222509 Chr.1: 111320186-111320186 on Build GRCh38
rs1012841079 Chr.1: 111320544-111320544 on Build GRCh38
rs368010768 Chr.1: 111320350-111320350 on Build GRCh38
rs775634313 Chr.1: 111320192-111320192 on Build GRCh38
rs774698859 Chr.1: 111320349-111320349 on Build GRCh38
rs914871408 Chr.1: 111320379-111320379 on Build GRCh38
rs759695811 Chr.1: 111320188-111320188 on Build GRCh38
rs997960475 Chr.1: 111320399-111320399 on Build GRCh38
rs143551303 Chr.1: 111320351-111320351 on Build GRCh38
rs527737477 Chr.1: 111320415-111320415 on Build GRCh38
rs187769579 Chr.1: 111320256-111320256 on Build GRCh38
rs140461957 Chr.1: 111320421-111320421 on Build GRCh38
rs370014119 Chr.1: 111320204-111320204 on Build GRCh38
rs762901024 Chr.1: 111320274-111320274 on Build GRCh38
rs768276405 Chr.1: 111320518-111320518 on Build GRCh38
rs368211379 Chr.1: 111320376-111320376 on Build GRCh38
rs201456008 Chr.1: 111320372-111320372 on Build GRCh38
rs749363817 Chr.1: 111320195-111320196 on Build GRCh38
rs140459744 Chr.1: 111320227-111320227 on Build GRCh38
rs756577970 Chr.1: 111320165-111320165 on Build GRCh38
rs778041295 Chr.1: 111320479-111320479 on Build GRCh38
rs767455378 Chr.1: 111320207-111320207 on Build GRCh38
rs146040153 Chr.1: 111320461-111320461 on Build GRCh38
rs144623576 Chr.1: 111320267-111320267 on Build GRCh38
rs769643079 Chr.1: 111320504-111320504 on Build GRCh38
rs1023175800 Chr.1: 111320570-111320570 on Build GRCh38
rs769386655 Chr.1: 111320430-111320430 on Build GRCh38
rs536956652 Chr.1: 111320341-111320341 on Build GRCh38
rs145057685 Chr.1: 111320412-111320412 on Build GRCh38
rs755685277 Chr.1: 111320222-111320222 on Build GRCh38
rs141163072 Chr.1: 111320505-111320505 on Build GRCh38
rs760579969 Chr.1: 111320292-111320292 on Build GRCh38
rs946613778 Chr.1: 111320201-111320201 on Build GRCh38
rs547890622 Chr.1: 111320452-111320452 on Build GRCh38
rs781067947 Chr.1: 111320385-111320385 on Build GRCh38
rs769991985 Chr.1: 111320191-111320191 on Build GRCh38
rs761167444 Chr.1: 111320437-111320437 on Build GRCh38
rs145023911 Chr.1: 111320255-111320255 on Build GRCh38
rs369442393 Chr.1: 111320343-111320343 on Build GRCh38
rs771591815 Chr.1: 111320184-111320184 on Build GRCh38
rs1053752843 Chr.1: 111320516-111320516 on Build GRCh38
rs756487510 Chr.1: 111320400-111320400 on Build GRCh38
rs910434832 Chr.1: 111320441-111320441 on Build GRCh38
rs765854980 Chr.1: 111320363-111320363 on Build GRCh38
rs769753703 Chr.1: 111320420-111320420 on Build GRCh38
rs774377527 Chr.1: 111320277-111320277 on Build GRCh38
rs887793812 Chr.1: 111320520-111320520 on Build GRCh38
rs918440510 Chr.1: 111320439-111320439 on Build GRCh38
rs199811046 Chr.1: 111320275-111320275 on Build GRCh38
rs773368726 Chr.1: 111320417-111320417 on Build GRCh38
rs936288007 Chr.1: 111320513-111320513 on Build GRCh38
rs746366495 Chr.1: 111320266-111320266 on Build GRCh38
rs757738885 Chr.1: 111320249-111320251 on Build GRCh38
rs781781725 Chr.1: 111320260-111320260 on Build GRCh38
rs766355170 Chr.1: 111320360-111320360 on Build GRCh38
rs139802202 Chr.1: 111320488-111320488 on Build GRCh38
rs756720776 Chr.1: 111320340-111320340 on Build GRCh38
rs758117743 Chr.1: 111320512-111320512 on Build GRCh38
rs766636380 Chr.1: 111320448-111320448 on Build GRCh38
rs754393219 Chr.1: 111320306-111320306 on Build GRCh38
rs761599968 Chr.1: 111320280-111320280 on Build GRCh38
rs748407448 Chr.1: 111320416-111320416 on Build GRCh38
rs754413469 Chr.1: 111320450-111320450 on Build GRCh38
rs746176151 Chr.1: 111320305-111320316 on Build GRCh38
rs150421495 Chr.1: 111320253-111320253 on Build GRCh38
rs377085074 Chr.1: 111320250-111320250 on Build GRCh38
rs749753951 Chr.1: 111320404-111320404 on Build GRCh38
rs775609194 Chr.1: 111320268-111320268 on Build GRCh38
rs2256721 Chr.1: 111320330-111320330 on Build GRCh38
rs763835872 Chr.1: 111320203-111320203 on Build GRCh38
rs149869566 Chr.1: 111320311-111320311 on Build GRCh38
rs761464220 Chr.1: 111320206-111320206 on Build GRCh38
rs757756209 Chr.1: 111320314-111320314 on Build GRCh38
rs1008709377 Chr.1: 111320219-111320219 on Build GRCh38
rs544028644 Chr.1: 111320433-111320433 on Build GRCh38
rs373720681 Chr.1: 111320483-111320483 on Build GRCh38
rs758408491 Chr.1: 111320478-111320478 on Build GRCh38
rs138227985 Chr.1: 111320281-111320281 on Build GRCh38
rs765911193 Chr.1: 111320300-111320300 on Build GRCh38
rs372590829 Chr.1: 111320220-111320220 on Build GRCh38
rs376164222 Chr.1: 111320301-111320301 on Build GRCh38
rs138073907 Chr.1: 111320353-111320353 on Build GRCh38
rs781674786 Chr.1: 111320320-111320320 on Build GRCh38
rs865961404 Chr.1: 111320294-111320294 on Build GRCh38
rs779521502 Chr.1: 111320237-111320237 on Build GRCh38
rs759369751 Chr.1: 111320425-111320425 on Build GRCh38
rs35242709 Chr.1: 111320307-111320307 on Build GRCh38
rs199879175 Chr.1: 111320241-111320241 on Build GRCh38
rs889525088 Chr.1: 111320572-111320572 on Build GRCh38
rs998722107 Chr.1: 111320541-111320541 on Build GRCh38
rs779585241 Chr.1: 111320296-111320307 on Build GRCh38
rs368987955 Chr.1: 111320173-111320173 on Build GRCh38
rs141747675 Chr.1: 111320276-111320276 on Build GRCh38
rs891973389 Chr.1: 111320528-111320528 on Build GRCh38

View Full Product Details