Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 148841729-148842275 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPB1
Entrez Gene ID
1360
Gene Name
carboxypeptidase B1
Gene Aliases
CPB, PASP, PCPB
UniGene
Hs.477891
Gene Chromosome Location
Chr.3: 148827801-148860187 on Build GRCh38
Transcripts
NM_001871.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6197236 c.475-7G>T p.? Chr.3: 148841816-148841816 on Build GRCh38
1039665 c.494A>G p.N165S Chr.3: 148841842-148841842 on Build GRCh38
3002368 c.500C>T p.P167L Chr.3: 148841848-148841848 on Build GRCh38
3002373 c.559C>A p.Q187K Chr.3: 148841907-148841907 on Build GRCh38
4552845 c.572G>A p.R191K Chr.3: 148841920-148841920 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs934028808 Chr.3: 148842123-148842123 on Build GRCh38
rs914850907 Chr.3: 148842112-148842112 on Build GRCh38
rs759928901 Chr.3: 148841924-148841924 on Build GRCh38
rs563356433 Chr.3: 148841946-148841946 on Build GRCh38
rs759588758 Chr.3: 148842219-148842220 on Build GRCh38
rs896165876 Chr.3: 148842222-148842222 on Build GRCh38
rs1016579426 Chr.3: 148841997-148841997 on Build GRCh38
rs370618017 Chr.3: 148841948-148841948 on Build GRCh38
rs1036909041 Chr.3: 148841782-148841782 on Build GRCh38
rs369320381 Chr.3: 148841912-148841912 on Build GRCh38
rs759994751 Chr.3: 148841817-148841817 on Build GRCh38
rs71617489 Chr.3: 148842222-148842223 on Build GRCh38
rs764639486 Chr.3: 148841950-148841950 on Build GRCh38
rs995444957 Chr.3: 148841852-148841852 on Build GRCh38
rs761204342 Chr.3: 148841826-148841826 on Build GRCh38
rs374054782 Chr.3: 148841964-148841964 on Build GRCh38
rs761472580 Chr.3: 148842211-148842211 on Build GRCh38
rs750318844 Chr.3: 148841835-148841835 on Build GRCh38
rs774916914 Chr.3: 148841816-148841816 on Build GRCh38
rs74283797 Chr.3: 148842077-148842077 on Build GRCh38
rs367547720 Chr.3: 148841844-148841844 on Build GRCh38
rs766261602 Chr.3: 148841965-148841965 on Build GRCh38
rs771043964 Chr.3: 148842219-148842219 on Build GRCh38
rs779424164 Chr.3: 148842003-148842003 on Build GRCh38
rs375559471 Chr.3: 148841919-148841919 on Build GRCh38
rs763775444 Chr.3: 148841819-148841819 on Build GRCh38
rs945517845 Chr.3: 148842166-148842166 on Build GRCh38
rs776208932 Chr.3: 148842201-148842201 on Build GRCh38
rs771574655 Chr.3: 148841922-148841922 on Build GRCh38
rs867912539 Chr.3: 148841920-148841920 on Build GRCh38
rs925519536 Chr.3: 148842203-148842203 on Build GRCh38
rs138120056 Chr.3: 148841864-148841864 on Build GRCh38
rs1002264650 Chr.3: 148841878-148841878 on Build GRCh38
rs773570432 Chr.3: 148841786-148841786 on Build GRCh38
rs1037239401 Chr.3: 148842216-148842216 on Build GRCh38
rs141911824 Chr.3: 148841907-148841907 on Build GRCh38
rs749310319 Chr.3: 148841905-148841905 on Build GRCh38
rs753555102 Chr.3: 148842148-148842148 on Build GRCh38
rs1026071717 Chr.3: 148841940-148841940 on Build GRCh38
rs578207825 Chr.3: 148841759-148841759 on Build GRCh38
rs779979694 Chr.3: 148841851-148841851 on Build GRCh38
rs73866670 Chr.3: 148841939-148841939 on Build GRCh38
rs1054680853 Chr.3: 148842221-148842221 on Build GRCh38
rs772719097 Chr.3: 148841954-148841954 on Build GRCh38
rs528127877 Chr.3: 148842144-148842144 on Build GRCh38
rs764839771 Chr.3: 148841827-148841827 on Build GRCh38
rs763555741 Chr.3: 148841796-148841796 on Build GRCh38
rs62274570 Chr.3: 148842133-148842133 on Build GRCh38
rs774397331 Chr.3: 148841804-148841804 on Build GRCh38
rs538631529 Chr.3: 148841858-148841858 on Build GRCh38
rs556841180 Chr.3: 148841928-148841928 on Build GRCh38
rs769939823 Chr.3: 148841775-148841775 on Build GRCh38
rs771200071 Chr.3: 148841815-148841815 on Build GRCh38
rs564987984 Chr.3: 148842229-148842229 on Build GRCh38
rs932643773 Chr.3: 148842121-148842121 on Build GRCh38
rs921267230 Chr.3: 148842085-148842085 on Build GRCh38
rs544990898 Chr.3: 148842204-148842204 on Build GRCh38
rs398106568 Chr.3: 148842233-148842234 on Build GRCh38
rs753565424 Chr.3: 148841821-148841821 on Build GRCh38

View Full Product Details