Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 150571653-150572187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2A
Entrez Gene ID
83939
Gene Name
eukaryotic translation initiation factor 2A
Gene Aliases
CDA02, EIF-2A, MST089, MSTP004, MSTP089
UniGene
Hs.655782
Gene Chromosome Location
Chr.3: 150546678-150586016 on Build GRCh38
Transcripts
NM_001319043.1 NM_001319044.1 NM_001319045.1 NM_001319046.1 NM_032025.4 XM_011513224.1 XM_011512698.2

Gene Symbol
SERP1
Entrez Gene ID
27230
Gene Name
stress-associated endoplasmic reticulum protein 1
Gene Aliases
RAMP4
UniGene
Hs.518326
Gene Chromosome Location
Chr.3: 150541993-150603177 on Build GRCh38
Transcripts
NM_001319043.1 NM_001319044.1 NM_001319045.1 NM_001319046.1 NM_032025.4 XM_011513224.1 XM_011512698.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5521337 c.837G>A p.L279L Chr.3: 150572058-150572058 on Build GRCh38
3003071 c.949C>T p.R317C Chr.3: 150572095-150572095 on Build GRCh38
5658433 c.1002T>G p.F334L Chr.3: 150572148-150572148 on Build GRCh38
5562085 c.886C>T p.P296S Chr.3: 150572032-150572032 on Build GRCh38
7110980 c.856G>C p.D286H Chr.3: 150572077-150572077 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs574153109 Chr.3: 150571984-150571984 on Build GRCh38
rs775350419 Chr.3: 150572071-150572071 on Build GRCh38
rs755744018 Chr.3: 150571963-150571963 on Build GRCh38
rs759266778 Chr.3: 150571943-150571943 on Build GRCh38
rs571586146 Chr.3: 150571915-150571915 on Build GRCh38
rs537162152 Chr.3: 150571944-150571944 on Build GRCh38
rs115089475 Chr.3: 150571762-150571762 on Build GRCh38
rs753237375 Chr.3: 150572089-150572089 on Build GRCh38
rs1031660728 Chr.3: 150571822-150571822 on Build GRCh38
rs939825079 Chr.3: 150571832-150571832 on Build GRCh38
rs372563492 Chr.3: 150572032-150572032 on Build GRCh38
rs765271029 Chr.3: 150571947-150571948 on Build GRCh38
rs929840918 Chr.3: 150572061-150572061 on Build GRCh38
rs762888932 Chr.3: 150572020-150572020 on Build GRCh38
rs765635866 Chr.3: 150571927-150571927 on Build GRCh38
rs763549541 Chr.3: 150571703-150571703 on Build GRCh38
rs376820577 Chr.3: 150572054-150572054 on Build GRCh38
rs762086747 Chr.3: 150571944-150571945 on Build GRCh38
rs201442630 Chr.3: 150572075-150572075 on Build GRCh38
rs750351034 Chr.3: 150572104-150572104 on Build GRCh38
rs186828790 Chr.3: 150572057-150572057 on Build GRCh38
rs758244130 Chr.3: 150572111-150572111 on Build GRCh38
rs1011982013 Chr.3: 150571767-150571767 on Build GRCh38
rs182147341 Chr.3: 150571701-150571701 on Build GRCh38
rs536892447 Chr.3: 150572025-150572025 on Build GRCh38
rs953391686 Chr.3: 150571978-150571978 on Build GRCh38
rs566472251 Chr.3: 150571814-150571814 on Build GRCh38
rs767848883 Chr.3: 150572129-150572129 on Build GRCh38
rs944060589 Chr.3: 150572003-150572003 on Build GRCh38
rs375957860 Chr.3: 150571954-150571954 on Build GRCh38
rs777251347 Chr.3: 150571909-150571909 on Build GRCh38
rs751969085 Chr.3: 150571820-150571820 on Build GRCh38
rs768789125 Chr.3: 150572039-150572039 on Build GRCh38
rs770163717 Chr.3: 150572043-150572043 on Build GRCh38
rs199994540 Chr.3: 150571970-150571970 on Build GRCh38
rs529851826 Chr.3: 150571764-150571764 on Build GRCh38
rs773404488 Chr.3: 150571938-150571938 on Build GRCh38
rs773742465 Chr.3: 150572047-150572047 on Build GRCh38
rs777054719 Chr.3: 150571943-150571943 on Build GRCh38
rs370372937 Chr.3: 150571753-150571753 on Build GRCh38
rs751997175 Chr.3: 150572131-150572134 on Build GRCh38
rs906683184 Chr.3: 150571809-150571809 on Build GRCh38
rs755131926 Chr.3: 150572138-150572138 on Build GRCh38
rs745610288 Chr.3: 150572016-150572016 on Build GRCh38
rs138050779 Chr.3: 150571855-150571855 on Build GRCh38
rs750490771 Chr.3: 150571958-150571959 on Build GRCh38
rs538532663 Chr.3: 150571816-150571816 on Build GRCh38
rs34211587 Chr.3: 150571842-150571843 on Build GRCh38
rs749257989 Chr.3: 150571800-150571800 on Build GRCh38
rs748715488 Chr.3: 150572042-150572042 on Build GRCh38
rs776505300 Chr.3: 150572041-150572041 on Build GRCh38
rs757247841 Chr.3: 150572095-150572095 on Build GRCh38
rs781367108 Chr.3: 150572152-150572152 on Build GRCh38
rs747611262 Chr.3: 150572096-150572096 on Build GRCh38
rs573613349 Chr.3: 150572124-150572124 on Build GRCh38
rs766901852 Chr.3: 150572047-150572048 on Build GRCh38
rs753819789 Chr.3: 150571981-150571981 on Build GRCh38
rs947778224 Chr.3: 150572153-150572153 on Build GRCh38
rs762218366 Chr.3: 150571910-150571910 on Build GRCh38
rs546388285 Chr.3: 150571768-150571768 on Build GRCh38
rs919832401 Chr.3: 150571902-150571902 on Build GRCh38
rs779696885 Chr.3: 150571700-150571700 on Build GRCh38
rs763676059 Chr.3: 150572084-150572084 on Build GRCh38
rs758158284 Chr.3: 150572019-150572019 on Build GRCh38
rs757083026 Chr.3: 150571982-150571982 on Build GRCh38

View Full Product Details