Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 151198885-151199412 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR171
Entrez Gene ID
29909
Gene Name
G protein-coupled receptor 171
Gene Aliases
H963
UniGene
Hs.549152
Gene Chromosome Location
Chr.3: 151197754-151203674 on Build GRCh38
Transcripts
NM_013308.3 XM_005247402.3 XM_005247403.3 XM_017006274.1 NM_053002.5 XM_006713487.3 XM_011512390.2 XM_011512394.2 XM_011512399.2 XM_017005676.1 XM_017005677.1 XM_017005678.1 XM_017005679.1 XM_017005680.1

Gene Symbol
MED12L
Entrez Gene ID
116931
Gene Name
mediator complex subunit 12 like
Gene Aliases
NOPAR, TNRC11L, TRALP, TRALPUSH
UniGene
Hs.744234
Gene Chromosome Location
Chr.3: 151085665-151436677 on Build GRCh38
Transcripts
NM_013308.3 XM_005247402.3 XM_005247403.3 XM_017006274.1 NM_053002.5 XM_006713487.3 XM_011512390.2 XM_011512394.2 XM_011512399.2 XM_017005676.1 XM_017005677.1 XM_017005678.1 XM_017005679.1 XM_017005680.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7174314 c.123G>T p.Q41H Chr.3: 151199264-151199264 on Build GRCh38
7274125 c.421A>G p.M141V Chr.3: 151198966-151198966 on Build GRCh38
222008 c.399G>A p.M133I Chr.3: 151198988-151198988 on Build GRCh38
1693600 c.302C>T p.S101L Chr.3: 151199085-151199085 on Build GRCh38
1752967 c.371G>A p.R124Q Chr.3: 151199016-151199016 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752246127 Chr.3: 151199054-151199054 on Build GRCh38
rs757701419 Chr.3: 151199053-151199053 on Build GRCh38
rs369126102 Chr.3: 151199016-151199016 on Build GRCh38
rs778504761 Chr.3: 151199298-151199298 on Build GRCh38
rs151172001 Chr.3: 151199056-151199056 on Build GRCh38
rs769805677 Chr.3: 151199242-151199242 on Build GRCh38
rs148323443 Chr.3: 151199333-151199333 on Build GRCh38
rs777389820 Chr.3: 151199173-151199173 on Build GRCh38
rs770822530 Chr.3: 151199011-151199011 on Build GRCh38
rs745692929 Chr.3: 151199096-151199096 on Build GRCh38
rs752344726 Chr.3: 151199154-151199154 on Build GRCh38
rs752410524 Chr.3: 151199164-151199164 on Build GRCh38
rs773431728 Chr.3: 151199265-151199265 on Build GRCh38
rs960856320 Chr.3: 151199292-151199292 on Build GRCh38
rs756728573 Chr.3: 151198922-151198922 on Build GRCh38
rs767867924 Chr.3: 151199378-151199378 on Build GRCh38
rs77550571 Chr.3: 151199215-151199215 on Build GRCh38
rs373786920 Chr.3: 151199257-151199257 on Build GRCh38
rs758775275 Chr.3: 151199158-151199158 on Build GRCh38
rs776655100 Chr.3: 151199257-151199258 on Build GRCh38
rs771393895 Chr.3: 151199367-151199367 on Build GRCh38
rs762433822 Chr.3: 151199135-151199135 on Build GRCh38
rs140237087 Chr.3: 151199188-151199188 on Build GRCh38
rs141844823 Chr.3: 151199219-151199219 on Build GRCh38
rs141482865 Chr.3: 151199372-151199372 on Build GRCh38
rs200562228 Chr.3: 151198978-151198978 on Build GRCh38
rs766785001 Chr.3: 151199256-151199256 on Build GRCh38
rs750878406 Chr.3: 151199156-151199156 on Build GRCh38
rs778377744 Chr.3: 151198928-151198928 on Build GRCh38
rs111283981 Chr.3: 151199126-151199126 on Build GRCh38
rs775872022 Chr.3: 151199116-151199116 on Build GRCh38
rs779992389 Chr.3: 151199198-151199198 on Build GRCh38
rs112520431 Chr.3: 151199047-151199047 on Build GRCh38
rs138457154 Chr.3: 151199274-151199274 on Build GRCh38
rs752799514 Chr.3: 151198914-151198914 on Build GRCh38
rs142089629 Chr.3: 151198995-151198995 on Build GRCh38
rs374880382 Chr.3: 151198935-151198935 on Build GRCh38
rs775816839 Chr.3: 151199026-151199026 on Build GRCh38
rs150296778 Chr.3: 151199210-151199210 on Build GRCh38
rs758396711 Chr.3: 151199306-151199306 on Build GRCh38
rs769212562 Chr.3: 151199003-151199003 on Build GRCh38
rs779080458 Chr.3: 151198971-151198971 on Build GRCh38
rs764025838 Chr.3: 151199035-151199035 on Build GRCh38
rs764301710 Chr.3: 151199131-151199131 on Build GRCh38
rs1048503998 Chr.3: 151198931-151198931 on Build GRCh38
rs766662546 Chr.3: 151199160-151199160 on Build GRCh38
rs774324192 Chr.3: 151199094-151199094 on Build GRCh38
rs769137703 Chr.3: 151199002-151199002 on Build GRCh38
rs756754600 Chr.3: 151199180-151199180 on Build GRCh38
rs776561323 Chr.3: 151199134-151199134 on Build GRCh38
rs867128601 Chr.3: 151199099-151199099 on Build GRCh38
rs762101365 Chr.3: 151199196-151199196 on Build GRCh38
rs772000549 Chr.3: 151199192-151199192 on Build GRCh38
rs768957309 Chr.3: 151199066-151199067 on Build GRCh38
rs369956369 Chr.3: 151199207-151199207 on Build GRCh38
rs910530378 Chr.3: 151199347-151199347 on Build GRCh38
rs560925200 Chr.3: 151199373-151199373 on Build GRCh38
rs757655164 Chr.3: 151198963-151198963 on Build GRCh38
rs769928153 Chr.3: 151199348-151199348 on Build GRCh38
rs774272776 Chr.3: 151199015-151199015 on Build GRCh38
rs747740700 Chr.3: 151199070-151199070 on Build GRCh38
rs749110096 Chr.3: 151199008-151199008 on Build GRCh38
rs756843318 Chr.3: 151199286-151199286 on Build GRCh38
rs754374608 Chr.3: 151199052-151199052 on Build GRCh38
rs141031765 Chr.3: 151198979-151198979 on Build GRCh38
rs34426089 Chr.3: 151199330-151199330 on Build GRCh38
rs753483581 Chr.3: 151199275-151199275 on Build GRCh38
rs552659115 Chr.3: 151199090-151199090 on Build GRCh38
rs761962260 Chr.3: 151199317-151199317 on Build GRCh38
rs755653141 Chr.3: 151199071-151199071 on Build GRCh38
rs143998423 Chr.3: 151199310-151199310 on Build GRCh38
rs1018695450 Chr.3: 151198998-151198998 on Build GRCh38
rs773263911 Chr.3: 151199350-151199350 on Build GRCh38
rs780842892 Chr.3: 151199069-151199069 on Build GRCh38
rs745746327 Chr.3: 151199190-151199190 on Build GRCh38
rs748712688 Chr.3: 151199175-151199175 on Build GRCh38
rs758720300 Chr.3: 151199058-151199058 on Build GRCh38
rs956892282 Chr.3: 151199105-151199105 on Build GRCh38
rs763446113 Chr.3: 151199255-151199255 on Build GRCh38
rs926957796 Chr.3: 151199233-151199233 on Build GRCh38
rs767820026 Chr.3: 151199024-151199024 on Build GRCh38
rs771947167 Chr.3: 151199102-151199102 on Build GRCh38
rs755704657 Chr.3: 151199167-151199167 on Build GRCh38
rs913975629 Chr.3: 151199322-151199323 on Build GRCh38
rs760865029 Chr.3: 151199028-151199028 on Build GRCh38
rs3773614 Chr.3: 151199213-151199213 on Build GRCh38
rs750456669 Chr.3: 151199301-151199301 on Build GRCh38
rs147118688 Chr.3: 151199252-151199252 on Build GRCh38
rs928147855 Chr.3: 151199268-151199268 on Build GRCh38
rs773712549 Chr.3: 151199245-151199245 on Build GRCh38
rs368287689 Chr.3: 151199079-151199079 on Build GRCh38
rs754254367 Chr.3: 151198953-151198953 on Build GRCh38
rs746750520 Chr.3: 151199315-151199315 on Build GRCh38
rs904445995 Chr.3: 151199040-151199040 on Build GRCh38
rs780720030 Chr.3: 151198980-151198980 on Build GRCh38
rs558551250 Chr.3: 151199259-151199259 on Build GRCh38
rs763495491 Chr.3: 151199363-151199363 on Build GRCh38
rs267599649 Chr.3: 151199211-151199211 on Build GRCh38

View Full Product Details