Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 157148118-157148633 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNL1
Entrez Gene ID
57018
Gene Name
cyclin L1
Gene Aliases
ANIA6A, BM-001, PRO1073, ania-6a
UniGene
Hs.4859
Gene Chromosome Location
Chr.3: 157146502-157160760 on Build GRCh38
Transcripts
NM_001308185.1 NM_020307.3 XM_005247648.3 XM_006713710.3 XM_011513011.2 XM_017006891.1 XM_017006892.1 XM_017006893.1 XM_017006894.1 XM_017006895.1 XM_017006896.1 XM_017006897.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3589331 c.1435C>T p.R479W Chr.3: 157148387-157148387 on Build GRCh38
5964814 c.1243A>G p.R415G Chr.3: 157148579-157148579 on Build GRCh38
4794357 c.1467G>A p.R489R Chr.3: 157148355-157148355 on Build GRCh38
3589330 c.1490A>G p.D497G Chr.3: 157148332-157148332 on Build GRCh38
3774673 c.1325A>T p.N442I Chr.3: 157148497-157148497 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752191984 Chr.3: 157148314-157148314 on Build GRCh38
rs781270124 Chr.3: 157148503-157148503 on Build GRCh38
rs1021349455 Chr.3: 157148173-157148173 on Build GRCh38
rs747331518 Chr.3: 157148530-157148530 on Build GRCh38
rs747051633 Chr.3: 157148344-157148344 on Build GRCh38
rs778485731 Chr.3: 157148438-157148438 on Build GRCh38
rs776185132 Chr.3: 157148343-157148343 on Build GRCh38
rs969292368 Chr.3: 157148180-157148180 on Build GRCh38
rs184355678 Chr.3: 157148158-157148158 on Build GRCh38
rs118144837 Chr.3: 157148169-157148169 on Build GRCh38
rs200316598 Chr.3: 157148387-157148387 on Build GRCh38
rs1051650634 Chr.3: 157148467-157148467 on Build GRCh38
rs768813144 Chr.3: 157148380-157148380 on Build GRCh38
rs929152364 Chr.3: 157148205-157148205 on Build GRCh38
rs761277889 Chr.3: 157148587-157148587 on Build GRCh38
rs767422683 Chr.3: 157148410-157148410 on Build GRCh38
rs746328075 Chr.3: 157148246-157148246 on Build GRCh38
rs866943376 Chr.3: 157148310-157148310 on Build GRCh38
rs146971876 Chr.3: 157148304-157148304 on Build GRCh38
rs771814832 Chr.3: 157148569-157148569 on Build GRCh38
rs943062654 Chr.3: 157148174-157148174 on Build GRCh38
rs546060937 Chr.3: 157148404-157148404 on Build GRCh38
rs369328973 Chr.3: 157148305-157148305 on Build GRCh38
rs776591955 Chr.3: 157148384-157148384 on Build GRCh38
rs948726021 Chr.3: 157148327-157148327 on Build GRCh38
rs746220067 Chr.3: 157148464-157148464 on Build GRCh38
rs764909288 Chr.3: 157148598-157148598 on Build GRCh38
rs762041871 Chr.3: 157148386-157148386 on Build GRCh38
rs751470106 Chr.3: 157148190-157148190 on Build GRCh38
rs758957176 Chr.3: 157148212-157148212 on Build GRCh38
rs769435206 Chr.3: 157148225-157148225 on Build GRCh38
rs766152219 Chr.3: 157148295-157148295 on Build GRCh38
rs774195037 Chr.3: 157148403-157148403 on Build GRCh38
rs898992606 Chr.3: 157148202-157148202 on Build GRCh38
rs777356907 Chr.3: 157148171-157148171 on Build GRCh38
rs769303845 Chr.3: 157148495-157148495 on Build GRCh38
rs906411651 Chr.3: 157148269-157148269 on Build GRCh38
rs755672783 Chr.3: 157148319-157148319 on Build GRCh38
rs1010937591 Chr.3: 157148381-157148381 on Build GRCh38
rs776949111 Chr.3: 157148289-157148289 on Build GRCh38
rs1029844869 Chr.3: 157148492-157148492 on Build GRCh38
rs371314438 Chr.3: 157148198-157148198 on Build GRCh38
rs773357422 Chr.3: 157148531-157148531 on Build GRCh38
rs552574994 Chr.3: 157148157-157148157 on Build GRCh38
rs746282584 Chr.3: 157148334-157148334 on Build GRCh38
rs774540849 Chr.3: 157148488-157148488 on Build GRCh38
rs200033201 Chr.3: 157148203-157148203 on Build GRCh38
rs202095674 Chr.3: 157148326-157148326 on Build GRCh38
rs142030218 Chr.3: 157148500-157148500 on Build GRCh38
rs780626373 Chr.3: 157148224-157148224 on Build GRCh38
rs748246366 Chr.3: 157148509-157148509 on Build GRCh38
rs764386846 Chr.3: 157148266-157148266 on Build GRCh38
rs777461968 Chr.3: 157148226-157148226 on Build GRCh38
rs376017453 Chr.3: 157148260-157148260 on Build GRCh38
rs747466174 Chr.3: 157148494-157148494 on Build GRCh38
rs756077496 Chr.3: 157148429-157148429 on Build GRCh38
rs774483966 Chr.3: 157148328-157148328 on Build GRCh38
rs528424179 Chr.3: 157148479-157148479 on Build GRCh38
rs753373067 Chr.3: 157148583-157148583 on Build GRCh38
rs777374443 Chr.3: 157148320-157148320 on Build GRCh38
rs749596489 Chr.3: 157148236-157148236 on Build GRCh38
rs780594103 Chr.3: 157148480-157148480 on Build GRCh38
rs368281773 Chr.3: 157148313-157148313 on Build GRCh38
rs540619875 Chr.3: 157148153-157148153 on Build GRCh38
rs760893912 Chr.3: 157148263-157148263 on Build GRCh38
rs775706047 Chr.3: 157148256-157148256 on Build GRCh38
rs751329404 Chr.3: 157148301-157148301 on Build GRCh38
rs774609871 Chr.3: 157148245-157148245 on Build GRCh38
rs112810419 Chr.3: 157148412-157148412 on Build GRCh38
rs372986934 Chr.3: 157148608-157148608 on Build GRCh38
rs764005118 Chr.3: 157148581-157148581 on Build GRCh38
rs181135757 Chr.3: 157148550-157148550 on Build GRCh38
rs772710305 Chr.3: 157148249-157148249 on Build GRCh38
rs772691946 Chr.3: 157148337-157148337 on Build GRCh38
rs753292523 Chr.3: 157148432-157148432 on Build GRCh38
rs750053960 Chr.3: 157148599-157148599 on Build GRCh38
rs771366552 Chr.3: 157148237-157148237 on Build GRCh38
rs763612660 Chr.3: 157148430-157148430 on Build GRCh38
rs760539080 Chr.3: 157148575-157148575 on Build GRCh38
rs761969024 Chr.3: 157148292-157148292 on Build GRCh38
rs879513582 Chr.3: 157148283-157148283 on Build GRCh38
rs1047572751 Chr.3: 157148253-157148253 on Build GRCh38
rs779270092 Chr.3: 157148332-157148332 on Build GRCh38
rs964410288 Chr.3: 157148604-157148604 on Build GRCh38
rs756930379 Chr.3: 157148434-157148434 on Build GRCh38

View Full Product Details