Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 185078495-185078994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf70
Entrez Gene ID
285382
Gene Name
chromosome 3 open reading frame 70
Gene Aliases
-
UniGene
Hs.380048
Gene Chromosome Location
Chr.3: 185078050-185153014 on Build GRCh38
Transcripts
NM_001025266.1

SNP Detail

SNP ID Chromosome Location
rs931385962 Chr.3: 185078772-185078772 on Build GRCh38
rs1037500989 Chr.3: 185078696-185078696 on Build GRCh38
rs887095462 Chr.3: 185078843-185078843 on Build GRCh38
rs75347208 Chr.3: 185078537-185078537 on Build GRCh38
rs917308006 Chr.3: 185078560-185078560 on Build GRCh38
rs1034259455 Chr.3: 185078805-185078805 on Build GRCh38
rs943277210 Chr.3: 185078664-185078664 on Build GRCh38
rs910508423 Chr.3: 185078636-185078636 on Build GRCh38
rs558724486 Chr.3: 185078745-185078745 on Build GRCh38
rs192962639 Chr.3: 185078938-185078938 on Build GRCh38
rs537003842 Chr.3: 185078676-185078676 on Build GRCh38
rs913537563 Chr.3: 185078572-185078572 on Build GRCh38
rs910841299 Chr.3: 185078648-185078648 on Build GRCh38
rs758497564 Chr.3: 185078918-185078918 on Build GRCh38
rs976755030 Chr.3: 185078965-185078965 on Build GRCh38
rs576040341 Chr.3: 185078969-185078969 on Build GRCh38
rs962174490 Chr.3: 185078567-185078567 on Build GRCh38
rs182834304 Chr.3: 185078813-185078813 on Build GRCh38
rs184892823 Chr.3: 185078538-185078538 on Build GRCh38
rs780465519 Chr.3: 185078967-185078967 on Build GRCh38
rs1041866322 Chr.3: 185078750-185078750 on Build GRCh38
rs534542353 Chr.3: 185078803-185078803 on Build GRCh38
rs567370844 Chr.3: 185078588-185078588 on Build GRCh38
rs7627623 Chr.3: 185078577-185078577 on Build GRCh38
rs910787427 Chr.3: 185078944-185078944 on Build GRCh38
rs114301808 Chr.3: 185078547-185078547 on Build GRCh38
rs778975483 Chr.3: 185078559-185078559 on Build GRCh38
rs1013579353 Chr.3: 185078844-185078844 on Build GRCh38
rs1014650662 Chr.3: 185078835-185078835 on Build GRCh38
rs556792807 Chr.3: 185078961-185078961 on Build GRCh38
rs944954338 Chr.3: 185078964-185078964 on Build GRCh38
rs878937738 Chr.3: 185078728-185078728 on Build GRCh38
rs78365662 Chr.3: 185078848-185078848 on Build GRCh38
rs957769431 Chr.3: 185078540-185078540 on Build GRCh38
rs755117956 Chr.3: 185078661-185078661 on Build GRCh38
rs188207866 Chr.3: 185078865-185078865 on Build GRCh38
rs952207820 Chr.3: 185078892-185078892 on Build GRCh38
rs150867358 Chr.3: 185078778-185078778 on Build GRCh38
rs190194281 Chr.3: 185078739-185078739 on Build GRCh38
rs972138154 Chr.3: 185078890-185078890 on Build GRCh38
rs1032485905 Chr.3: 185078876-185078876 on Build GRCh38
rs1050636071 Chr.3: 185078623-185078623 on Build GRCh38
rs987384918 Chr.3: 185078626-185078626 on Build GRCh38
rs1000069573 Chr.3: 185078782-185078782 on Build GRCh38
rs566523268 Chr.3: 185078856-185078856 on Build GRCh38
rs1002411579 Chr.3: 185078875-185078875 on Build GRCh38
rs986292119 Chr.3: 185078939-185078939 on Build GRCh38
rs1040741125 Chr.3: 185078741-185078741 on Build GRCh38
rs958460130 Chr.3: 185078891-185078891 on Build GRCh38
rs758820478 Chr.3: 185078852-185078852 on Build GRCh38
rs1009959366 Chr.3: 185078746-185078746 on Build GRCh38

View Full Product Details