Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186570588-186571076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB11
Entrez Gene ID
51726
Gene Name
DnaJ heat shock protein family (Hsp40) member B11
Gene Aliases
ABBP-2, ABBP2, DJ9, Dj-9, EDJ, ERdj3, ERj3, ERj3p, PRO1080, UNQ537
UniGene
Hs.317192
Gene Chromosome Location
Chr.3: 186570676-186585800 on Build GRCh38
Transcripts
NM_016306.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5800498 c.56C>T p.A19V Chr.3: 186570953-186570953 on Build GRCh38
4429821 c.35T>C p.L12S Chr.3: 186570932-186570932 on Build GRCh38
302083 c.67G>T p.G23* Chr.3: 186570964-186570964 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs538824536 Chr.3: 186570924-186570924 on Build GRCh38
rs977963649 Chr.3: 186570774-186570774 on Build GRCh38
rs375248047 Chr.3: 186570939-186570939 on Build GRCh38
rs112135705 Chr.3: 186570647-186570647 on Build GRCh38
rs773582025 Chr.3: 186570849-186570849 on Build GRCh38
rs949274824 Chr.3: 186570650-186570650 on Build GRCh38
rs373431036 Chr.3: 186570920-186570920 on Build GRCh38
rs368637259 Chr.3: 186570795-186570795 on Build GRCh38
rs192676872 Chr.3: 186570737-186570737 on Build GRCh38
rs766548495 Chr.3: 186570738-186570738 on Build GRCh38
rs1019498920 Chr.3: 186570831-186570831 on Build GRCh38
rs996377568 Chr.3: 186570674-186570674 on Build GRCh38
rs756897398 Chr.3: 186570911-186570911 on Build GRCh38
rs778181474 Chr.3: 186571030-186571030 on Build GRCh38
rs753314675 Chr.3: 186570963-186570963 on Build GRCh38
rs201275361 Chr.3: 186570989-186570989 on Build GRCh38
rs1025282709 Chr.3: 186570951-186570951 on Build GRCh38
rs756952533 Chr.3: 186570964-186570964 on Build GRCh38
rs1057393151 Chr.3: 186570761-186570761 on Build GRCh38
rs895988653 Chr.3: 186570648-186570648 on Build GRCh38
rs902614499 Chr.3: 186570656-186570656 on Build GRCh38
rs988628815 Chr.3: 186570725-186570725 on Build GRCh38
rs751630079 Chr.3: 186571015-186571016 on Build GRCh38
rs558361447 Chr.3: 186570891-186570891 on Build GRCh38
rs569655075 Chr.3: 186570830-186570830 on Build GRCh38
rs868587136 Chr.3: 186570741-186570741 on Build GRCh38
rs368324560 Chr.3: 186570942-186570942 on Build GRCh38
rs1053833012 Chr.3: 186570784-186570784 on Build GRCh38
rs780496316 Chr.3: 186570866-186570867 on Build GRCh38
rs374445957 Chr.3: 186570611-186570611 on Build GRCh38
rs770056805 Chr.3: 186570928-186570928 on Build GRCh38
rs973853213 Chr.3: 186570692-186570692 on Build GRCh38
rs1030611852 Chr.3: 186570688-186570688 on Build GRCh38
rs899346499 Chr.3: 186571031-186571031 on Build GRCh38
rs1042846186 Chr.3: 186570654-186570654 on Build GRCh38
rs758932906 Chr.3: 186570861-186570863 on Build GRCh38
rs369204533 Chr.3: 186571008-186571008 on Build GRCh38
rs3733160 Chr.3: 186571021-186571021 on Build GRCh38
rs539963666 Chr.3: 186570696-186570696 on Build GRCh38
rs1035914562 Chr.3: 186570634-186570634 on Build GRCh38
rs565966225 Chr.3: 186570743-186570743 on Build GRCh38
rs760013263 Chr.3: 186570883-186570883 on Build GRCh38
rs879434099 Chr.3: 186571022-186571022 on Build GRCh38
rs763255626 Chr.3: 186570861-186570861 on Build GRCh38
rs565048503 Chr.3: 186570689-186570689 on Build GRCh38
rs953398277 Chr.3: 186570796-186570796 on Build GRCh38
rs535800631 Chr.3: 186571035-186571035 on Build GRCh38
rs780786338 Chr.3: 186570720-186570720 on Build GRCh38
rs74688115 Chr.3: 186570886-186570886 on Build GRCh38
rs761152683 Chr.3: 186570959-186570959 on Build GRCh38
rs753543507 Chr.3: 186570905-186570905 on Build GRCh38
rs949217135 Chr.3: 186570751-186570751 on Build GRCh38
rs114104753 Chr.3: 186570629-186570629 on Build GRCh38
rs1015733122 Chr.3: 186570719-186570719 on Build GRCh38
rs751926194 Chr.3: 186570881-186570881 on Build GRCh38
rs917862773 Chr.3: 186570745-186570745 on Build GRCh38
rs780046263 Chr.3: 186570922-186570922 on Build GRCh38
rs959058608 Chr.3: 186570742-186570742 on Build GRCh38
rs59536219 Chr.3: 186570649-186570649 on Build GRCh38
rs543934788 Chr.3: 186571038-186571038 on Build GRCh38
rs561286345 Chr.3: 186571024-186571024 on Build GRCh38
rs936646669 Chr.3: 186570657-186570657 on Build GRCh38
rs186971810 Chr.3: 186570665-186570665 on Build GRCh38
rs767704483 Chr.3: 186570947-186570947 on Build GRCh38
rs3806714 Chr.3: 186570776-186570776 on Build GRCh38
rs773296409 Chr.3: 186570853-186570854 on Build GRCh38
rs953994666 Chr.3: 186570706-186570706 on Build GRCh38
rs3806713 Chr.3: 186570732-186570732 on Build GRCh38
rs372490167 Chr.3: 186571016-186571016 on Build GRCh38
rs779703156 Chr.3: 186570995-186570995 on Build GRCh38
rs1005493863 Chr.3: 186570777-186570777 on Build GRCh38
rs149168314 Chr.3: 186570892-186570892 on Build GRCh38
rs994870661 Chr.3: 186570789-186570789 on Build GRCh38
rs1005071040 Chr.3: 186570715-186570715 on Build GRCh38
rs879881683 Chr.3: 186570878-186570878 on Build GRCh38
rs548183319 Chr.3: 186570782-186570782 on Build GRCh38
rs775986965 Chr.3: 186570948-186570948 on Build GRCh38
rs766724299 Chr.3: 186570864-186570864 on Build GRCh38
rs755963875 Chr.3: 186570936-186570936 on Build GRCh38
rs143266285 Chr.3: 186570992-186570992 on Build GRCh38
rs936964427 Chr.3: 186570747-186570747 on Build GRCh38
rs917527648 Chr.3: 186570993-186570993 on Build GRCh38
rs879661648 Chr.3: 186570655-186570655 on Build GRCh38
rs778612390 Chr.3: 186570913-186570913 on Build GRCh38
rs900498544 Chr.3: 186570821-186570821 on Build GRCh38
rs184825975 Chr.3: 186570617-186570617 on Build GRCh38
rs943871538 Chr.3: 186570721-186570721 on Build GRCh38
rs764962665 Chr.3: 186570980-186570980 on Build GRCh38
rs900551224 Chr.3: 186570668-186570668 on Build GRCh38
rs978525871 Chr.3: 186570896-186570896 on Build GRCh38
rs1036531268 Chr.3: 186570782-186570782 on Build GRCh38
rs973749614 Chr.3: 186570607-186570607 on Build GRCh38
rs1005428561 Chr.3: 186570903-186570903 on Build GRCh38
rs763656566 Chr.3: 186570961-186570961 on Build GRCh38

View Full Product Details