Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 190402244-190402778 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN16
Entrez Gene ID
10686
Gene Name
claudin 16
Gene Aliases
HOMG3, PCLN1
UniGene
Hs.251391
Gene Chromosome Location
Chr.3: 190387872-190412143 on Build GRCh38
Transcripts
NM_006580.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3714426 c.387C>T p.R129R Chr.3: 190402399-190402399 on Build GRCh38
4547201 c.417G>A p.A139A Chr.3: 190402429-190402429 on Build GRCh38
5059568 c.340C>T p.R114* Chr.3: 190402352-190402352 on Build GRCh38
1042521 c.385C>T p.R129C Chr.3: 190402397-190402397 on Build GRCh38
4931862 c.350G>T p.W117L Chr.3: 190402362-190402362 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs950141430 Chr.3: 190402417-190402417 on Build GRCh38
rs183936028 Chr.3: 190402665-190402665 on Build GRCh38
rs766544519 Chr.3: 190402679-190402679 on Build GRCh38
rs573024259 Chr.3: 190402378-190402378 on Build GRCh38
rs973965903 Chr.3: 190402607-190402607 on Build GRCh38
rs572698245 Chr.3: 190402747-190402747 on Build GRCh38
rs958967873 Chr.3: 190402652-190402652 on Build GRCh38
rs747654138 Chr.3: 190402391-190402391 on Build GRCh38
rs758799163 Chr.3: 190402432-190402432 on Build GRCh38
rs931894180 Chr.3: 190402753-190402753 on Build GRCh38
rs536119123 Chr.3: 190402557-190402557 on Build GRCh38
rs746660695 Chr.3: 190402305-190402305 on Build GRCh38
rs572445470 Chr.3: 190402496-190402496 on Build GRCh38
rs117811086 Chr.3: 190402640-190402640 on Build GRCh38
rs770764817 Chr.3: 190402476-190402476 on Build GRCh38
rs201755973 Chr.3: 190402730-190402733 on Build GRCh38
rs780312463 Chr.3: 190402384-190402384 on Build GRCh38
rs773087384 Chr.3: 190402327-190402327 on Build GRCh38
rs900345083 Chr.3: 190402623-190402623 on Build GRCh38
rs369835183 Chr.3: 190402315-190402315 on Build GRCh38
rs1023189854 Chr.3: 190402727-190402727 on Build GRCh38
rs928370591 Chr.3: 190402510-190402510 on Build GRCh38
rs552956716 Chr.3: 190402515-190402515 on Build GRCh38
rs764353770 Chr.3: 190402344-190402344 on Build GRCh38
rs908907436 Chr.3: 190402751-190402751 on Build GRCh38
rs1008684926 Chr.3: 190402578-190402578 on Build GRCh38
rs762831659 Chr.3: 190402398-190402398 on Build GRCh38
rs999764081 Chr.3: 190402467-190402467 on Build GRCh38
rs756914384 Chr.3: 190402449-190402450 on Build GRCh38
rs765249393 Chr.3: 190402316-190402316 on Build GRCh38
rs758739389 Chr.3: 190402382-190402382 on Build GRCh38
rs146727044 Chr.3: 190402361-190402361 on Build GRCh38
rs766383362 Chr.3: 190402399-190402399 on Build GRCh38
rs890091664 Chr.3: 190402572-190402572 on Build GRCh38
rs1031705285 Chr.3: 190402693-190402693 on Build GRCh38
rs111986587 Chr.3: 190402300-190402300 on Build GRCh38
rs104893732 Chr.3: 190402362-190402362 on Build GRCh38
rs768289133 Chr.3: 190402500-190402500 on Build GRCh38
rs144105475 Chr.3: 190402387-190402387 on Build GRCh38
rs552541824 Chr.3: 190402746-190402746 on Build GRCh38
rs753331509 Chr.3: 190402299-190402300 on Build GRCh38
rs762740548 Chr.3: 190402330-190402330 on Build GRCh38
rs962259504 Chr.3: 190402592-190402592 on Build GRCh38
rs761824865 Chr.3: 190402415-190402415 on Build GRCh38
rs747813363 Chr.3: 190402460-190402460 on Build GRCh38
rs753901053 Chr.3: 190402352-190402352 on Build GRCh38
rs560084879 Chr.3: 190402742-190402742 on Build GRCh38
rs759388455 Chr.3: 190402410-190402410 on Build GRCh38
rs566495939 Chr.3: 190402534-190402534 on Build GRCh38
rs1036267501 Chr.3: 190402745-190402745 on Build GRCh38
rs757395583 Chr.3: 190402353-190402353 on Build GRCh38
rs374846039 Chr.3: 190402731-190402734 on Build GRCh38
rs56147287 Chr.3: 190402393-190402393 on Build GRCh38
rs149965853 Chr.3: 190402371-190402371 on Build GRCh38
rs991664500 Chr.3: 190402659-190402659 on Build GRCh38
rs570599662 Chr.3: 190402493-190402493 on Build GRCh38
rs540351930 Chr.3: 190402719-190402719 on Build GRCh38
rs990385956 Chr.3: 190402449-190402449 on Build GRCh38
rs556056997 Chr.3: 190402613-190402613 on Build GRCh38
rs893701631 Chr.3: 190402529-190402529 on Build GRCh38
rs780658485 Chr.3: 190402443-190402443 on Build GRCh38
rs142152395 Chr.3: 190402341-190402341 on Build GRCh38
rs79157522 Chr.3: 190402307-190402307 on Build GRCh38
rs765256758 Chr.3: 190402428-190402428 on Build GRCh38
rs577671505 Chr.3: 190402695-190402695 on Build GRCh38
rs199651054 Chr.3: 190402414-190402414 on Build GRCh38
rs776248582 Chr.3: 190402311-190402311 on Build GRCh38
rs61752083 Chr.3: 190402383-190402383 on Build GRCh38
rs137882210 Chr.3: 190402429-190402429 on Build GRCh38
rs774413495 Chr.3: 190402483-190402483 on Build GRCh38
rs747440607 Chr.3: 190402385-190402385 on Build GRCh38
rs751959432 Chr.3: 190402444-190402444 on Build GRCh38
rs191766695 Chr.3: 190402294-190402294 on Build GRCh38
rs773141087 Chr.3: 190402396-190402396 on Build GRCh38
rs777119599 Chr.3: 190402472-190402472 on Build GRCh38
rs528344809 Chr.3: 190402339-190402339 on Build GRCh38
rs1015083535 Chr.3: 190402583-190402583 on Build GRCh38
rs908877703 Chr.3: 190402577-190402577 on Build GRCh38
rs748881419 Chr.3: 190402475-190402475 on Build GRCh38
rs920823997 Chr.3: 190402373-190402373 on Build GRCh38
rs77798557 Chr.3: 190402463-190402463 on Build GRCh38
rs189008859 Chr.3: 190402685-190402685 on Build GRCh38
rs370592530 Chr.3: 190402372-190402372 on Build GRCh38
rs1028633906 Chr.3: 190402268-190402268 on Build GRCh38
rs755455995 Chr.3: 190402452-190402452 on Build GRCh38
rs780921178 Chr.3: 190402456-190402456 on Build GRCh38
rs773999256 Chr.3: 190402409-190402409 on Build GRCh38
rs758630693 Chr.3: 190402645-190402645 on Build GRCh38
rs558079402 Chr.3: 190402681-190402681 on Build GRCh38
rs1055737562 Chr.3: 190402390-190402390 on Build GRCh38
rs746240348 Chr.3: 190402298-190402298 on Build GRCh38
rs775152656 Chr.3: 190402304-190402304 on Build GRCh38

View Full Product Details