Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 2630844-2631353 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM193A
Entrez Gene ID
8603
Gene Name
family with sequence similarity 193 member A
Gene Aliases
C4orf8, RES4-22
UniGene
Hs.652364
Gene Chromosome Location
Chr.4: 2596288-2732575 on Build GRCh38
Transcripts
NM_001256666.1 NM_001256667.1 NM_001256668.1 NM_003704.3 NR_046335.1 NR_046336.1 XM_006713930.2 XM_006713932.2 XM_011513590.1 XM_011513593.2 XM_017008788.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
340693 c.106G>A p.E36K Chr.4: 2631110-2631110 on Build GRCh38
1054706 c.114C>T p.Y38Y Chr.4: 2631118-2631118 on Build GRCh38
4844605 c.90C>G p.F30L Chr.4: 2631094-2631094 on Build GRCh38
1054707 c.117C>T p.G39G Chr.4: 2631121-2631121 on Build GRCh38
4846906 c.106G>A p.E36K Chr.4: 2631110-2631110 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777305061 Chr.4: 2631034-2631034 on Build GRCh38
rs746220944 Chr.4: 2631285-2631285 on Build GRCh38
rs941488968 Chr.4: 2631184-2631184 on Build GRCh38
rs963711815 Chr.4: 2631262-2631262 on Build GRCh38
rs1032732091 Chr.4: 2631305-2631305 on Build GRCh38
rs764638107 Chr.4: 2630893-2630906 on Build GRCh38
rs748824110 Chr.4: 2630922-2630922 on Build GRCh38
rs776038615 Chr.4: 2630942-2630942 on Build GRCh38
rs754782954 Chr.4: 2630910-2630910 on Build GRCh38
rs563665335 Chr.4: 2631316-2631316 on Build GRCh38
rs756554870 Chr.4: 2631204-2631204 on Build GRCh38
rs751351871 Chr.4: 2630908-2630908 on Build GRCh38
rs976862756 Chr.4: 2631265-2631265 on Build GRCh38
rs781770559 Chr.4: 2631201-2631201 on Build GRCh38
rs375119879 Chr.4: 2631173-2631174 on Build GRCh38
rs1001591822 Chr.4: 2630871-2630871 on Build GRCh38
rs373437678 Chr.4: 2631040-2631040 on Build GRCh38
rs759463274 Chr.4: 2631074-2631074 on Build GRCh38
rs766336343 Chr.4: 2630893-2630893 on Build GRCh38
rs145959580 Chr.4: 2631033-2631033 on Build GRCh38
rs772041994 Chr.4: 2631142-2631142 on Build GRCh38
rs753218634 Chr.4: 2631203-2631203 on Build GRCh38
rs761584276 Chr.4: 2631181-2631181 on Build GRCh38
rs775355104 Chr.4: 2631146-2631146 on Build GRCh38
rs755610595 Chr.4: 2631094-2631094 on Build GRCh38
rs763059861 Chr.4: 2630974-2630974 on Build GRCh38
rs138840649 Chr.4: 2631039-2631039 on Build GRCh38
rs752153023 Chr.4: 2630957-2630957 on Build GRCh38
rs1035700620 Chr.4: 2631244-2631244 on Build GRCh38
rs182761703 Chr.4: 2631312-2631312 on Build GRCh38
rs549427304 Chr.4: 2630991-2630991 on Build GRCh38
rs922369082 Chr.4: 2631267-2631267 on Build GRCh38
rs772491215 Chr.4: 2630941-2630941 on Build GRCh38
rs754367808 Chr.4: 2630908-2630909 on Build GRCh38
rs35176325 Chr.4: 2631147-2631148 on Build GRCh38
rs748519121 Chr.4: 2631042-2631042 on Build GRCh38
rs754335021 Chr.4: 2631079-2631079 on Build GRCh38
rs150727963 Chr.4: 2631085-2631085 on Build GRCh38
rs762968373 Chr.4: 2631060-2631060 on Build GRCh38
rs975233613 Chr.4: 2631322-2631322 on Build GRCh38
rs779276123 Chr.4: 2631255-2631255 on Build GRCh38
rs760491986 Chr.4: 2631149-2631149 on Build GRCh38
rs747761208 Chr.4: 2630913-2630913 on Build GRCh38
rs747355342 Chr.4: 2630946-2630946 on Build GRCh38
rs748574144 Chr.4: 2631110-2631110 on Build GRCh38
rs866122091 Chr.4: 2631157-2631157 on Build GRCh38
rs139878795 Chr.4: 2631077-2631077 on Build GRCh38
rs540872958 Chr.4: 2631257-2631257 on Build GRCh38
rs533280356 Chr.4: 2631304-2631304 on Build GRCh38
rs777319424 Chr.4: 2630919-2630919 on Build GRCh38
rs781755414 Chr.4: 2631099-2631099 on Build GRCh38
rs139977579 Chr.4: 2631117-2631117 on Build GRCh38
rs575968322 Chr.4: 2631127-2631127 on Build GRCh38
rs773462616 Chr.4: 2631059-2631059 on Build GRCh38
rs201858739 Chr.4: 2631183-2631183 on Build GRCh38
rs767237009 Chr.4: 2630996-2630996 on Build GRCh38
rs753444105 Chr.4: 2631180-2631180 on Build GRCh38
rs546990334 Chr.4: 2631249-2631249 on Build GRCh38
rs568706269 Chr.4: 2631280-2631280 on Build GRCh38
rs149864699 Chr.4: 2631136-2631136 on Build GRCh38
rs750397636 Chr.4: 2630889-2630889 on Build GRCh38
rs375509750 Chr.4: 2630988-2630988 on Build GRCh38
rs748796211 Chr.4: 2631014-2631014 on Build GRCh38
rs756724055 Chr.4: 2631082-2631082 on Build GRCh38
rs1043296144 Chr.4: 2631320-2631320 on Build GRCh38
rs763881604 Chr.4: 2631178-2631178 on Build GRCh38
rs749606840 Chr.4: 2631213-2631213 on Build GRCh38
rs755828198 Chr.4: 2631001-2631001 on Build GRCh38
rs141509573 Chr.4: 2631078-2631078 on Build GRCh38
rs752525151 Chr.4: 2631075-2631075 on Build GRCh38
rs373548111 Chr.4: 2630969-2630969 on Build GRCh38
rs771221166 Chr.4: 2631214-2631214 on Build GRCh38
rs1035902628 Chr.4: 2630873-2630873 on Build GRCh38
rs942649117 Chr.4: 2630878-2630878 on Build GRCh38
rs201163715 Chr.4: 2631196-2631196 on Build GRCh38
rs755628154 Chr.4: 2630914-2630914 on Build GRCh38
rs558706176 Chr.4: 2631005-2631005 on Build GRCh38
rs770236370 Chr.4: 2631119-2631119 on Build GRCh38
rs774164919 Chr.4: 2631072-2631072 on Build GRCh38
rs769099752 Chr.4: 2630951-2630951 on Build GRCh38
rs375526486 Chr.4: 2631121-2631121 on Build GRCh38
rs770934285 Chr.4: 2631133-2631133 on Build GRCh38
rs1009812733 Chr.4: 2631256-2631256 on Build GRCh38
rs555382518 Chr.4: 2631185-2631185 on Build GRCh38
rs762067878 Chr.4: 2630956-2630956 on Build GRCh38
rs556150749 Chr.4: 2631122-2631122 on Build GRCh38
rs566227606 Chr.4: 2631024-2631024 on Build GRCh38
rs770183264 Chr.4: 2631048-2631048 on Build GRCh38
rs752363732 Chr.4: 2630999-2630999 on Build GRCh38
rs973020395 Chr.4: 2630905-2630905 on Build GRCh38
rs376773733 Chr.4: 2631067-2631067 on Build GRCh38
rs754763360 Chr.4: 2630992-2630992 on Build GRCh38
rs564225899 Chr.4: 2631261-2631261 on Build GRCh38
rs777368679 Chr.4: 2631013-2631013 on Build GRCh38
rs749887769 Chr.4: 2631088-2631088 on Build GRCh38
rs905359391 Chr.4: 2631023-2631023 on Build GRCh38
rs778146547 Chr.4: 2631205-2631205 on Build GRCh38
rs112748325 Chr.4: 2630891-2630891 on Build GRCh38
rs761142467 Chr.4: 2630891-2630892 on Build GRCh38
rs759576739 Chr.4: 2631141-2631141 on Build GRCh38
rs780849061 Chr.4: 2630911-2630911 on Build GRCh38
rs749463103 Chr.4: 2631130-2631130 on Build GRCh38
rs776997521 Chr.4: 2630955-2630955 on Build GRCh38
rs905937130 Chr.4: 2630870-2630870 on Build GRCh38

View Full Product Details