Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 2884514-2885016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADD1
Entrez Gene ID
118
Gene Name
adducin 1
Gene Aliases
ADDA
UniGene
Hs.183706
Gene Chromosome Location
Chr.4: 2843732-2930076 on Build GRCh38
Transcripts
NM_001119.4 NM_001286645.1 NM_014189.3 NM_014190.3 NM_176801.2 XM_005247933.2 XM_005247934.2 XM_005247935.2 XM_005247936.2 XM_005247937.2 XM_017007703.1 XM_017007704.1 XM_017007705.1 XM_017007706.1 XM_017007707.1 XM_017007708.1 XM_017007709.1 XM_017007710.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5013988 c.436C>T p.R146W Chr.4: 2884592-2884592 on Build GRCh38
6865433 c.381G>A p.V127V Chr.4: 2884537-2884537 on Build GRCh38
6873149 c.478G>T p.G160W Chr.4: 2884634-2884634 on Build GRCh38
3603266 c.404C>T p.S135F Chr.4: 2884560-2884560 on Build GRCh38
6595104 c.411G>A p.A137A Chr.4: 2884567-2884567 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745452586 Chr.4: 2884713-2884713 on Build GRCh38
rs752623937 Chr.4: 2884670-2884670 on Build GRCh38
rs759674972 Chr.4: 2884656-2884656 on Build GRCh38
rs777115034 Chr.4: 2884626-2884626 on Build GRCh38
rs771500177 Chr.4: 2884716-2884716 on Build GRCh38
rs192899881 Chr.4: 2884633-2884633 on Build GRCh38
rs770382552 Chr.4: 2884594-2884594 on Build GRCh38
rs539462843 Chr.4: 2884925-2884925 on Build GRCh38
rs369225621 Chr.4: 2884578-2884578 on Build GRCh38
rs180940335 Chr.4: 2884965-2884965 on Build GRCh38
rs1015084995 Chr.4: 2884976-2884976 on Build GRCh38
rs776509726 Chr.4: 2884817-2884817 on Build GRCh38
rs59790260 Chr.4: 2884923-2884923 on Build GRCh38
rs778528002 Chr.4: 2884704-2884704 on Build GRCh38
rs1032861027 Chr.4: 2884900-2884900 on Build GRCh38
rs954902081 Chr.4: 2884955-2884955 on Build GRCh38
rs781252718 Chr.4: 2884553-2884553 on Build GRCh38
rs189426139 Chr.4: 2884928-2884928 on Build GRCh38
rs78892897 Chr.4: 2884882-2884882 on Build GRCh38
rs745364833 Chr.4: 2884609-2884609 on Build GRCh38
rs973176220 Chr.4: 2884613-2884613 on Build GRCh38
rs778437831 Chr.4: 2884608-2884608 on Build GRCh38
rs1046177786 Chr.4: 2884865-2884865 on Build GRCh38
rs766430572 Chr.4: 2884540-2884540 on Build GRCh38
rs147887114 Chr.4: 2884971-2884971 on Build GRCh38
rs777305817 Chr.4: 2884579-2884579 on Build GRCh38
rs142505906 Chr.4: 2884599-2884599 on Build GRCh38
rs374216380 Chr.4: 2884640-2884640 on Build GRCh38
rs201569938 Chr.4: 2884541-2884541 on Build GRCh38
rs748923951 Chr.4: 2884592-2884592 on Build GRCh38
rs536186215 Chr.4: 2884840-2884840 on Build GRCh38
rs532064677 Chr.4: 2884778-2884778 on Build GRCh38
rs767568364 Chr.4: 2884659-2884659 on Build GRCh38
rs137927328 Chr.4: 2884567-2884567 on Build GRCh38
rs112288208 Chr.4: 2884684-2884684 on Build GRCh38
rs983295533 Chr.4: 2884890-2884890 on Build GRCh38
rs772859513 Chr.4: 2884575-2884575 on Build GRCh38
rs373466803 Chr.4: 2884834-2884834 on Build GRCh38
rs769212546 Chr.4: 2884614-2884614 on Build GRCh38
rs762085283 Chr.4: 2884632-2884632 on Build GRCh38
rs760841629 Chr.4: 2884667-2884667 on Build GRCh38
rs201776350 Chr.4: 2884673-2884673 on Build GRCh38
rs145970845 Chr.4: 2884654-2884654 on Build GRCh38
rs920124759 Chr.4: 2884622-2884622 on Build GRCh38
rs895000447 Chr.4: 2884831-2884831 on Build GRCh38
rs777585727 Chr.4: 2884688-2884688 on Build GRCh38
rs755872563 Chr.4: 2884570-2884570 on Build GRCh38
rs753548401 Chr.4: 2884695-2884695 on Build GRCh38
rs867742075 Chr.4: 2884711-2884711 on Build GRCh38
rs35580670 Chr.4: 2884735-2884735 on Build GRCh38
rs773325179 Chr.4: 2884635-2884635 on Build GRCh38
rs763194143 Chr.4: 2884636-2884636 on Build GRCh38
rs972928661 Chr.4: 2884803-2884803 on Build GRCh38
rs973591119 Chr.4: 2884969-2884969 on Build GRCh38
rs759866387 Chr.4: 2884855-2884855 on Build GRCh38
rs1001396039 Chr.4: 2884789-2884789 on Build GRCh38
rs756952817 Chr.4: 2884954-2884955 on Build GRCh38
rs201672253 Chr.4: 2884946-2884946 on Build GRCh38
rs34395973 Chr.4: 2884696-2884696 on Build GRCh38

View Full Product Details