Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 2898099-2898597 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADD1
Entrez Gene ID
118
Gene Name
adducin 1
Gene Aliases
ADDA
UniGene
Hs.183706
Gene Chromosome Location
Chr.4: 2843732-2930076 on Build GRCh38
Transcripts
NM_001119.4 NM_001286645.1 NM_014189.3 NM_014190.3 NM_176801.2 XM_005247933.2 XM_005247934.2 XM_005247935.2 XM_005247936.2 XM_005247937.2 XM_017007703.1 XM_017007704.1 XM_017007705.1 XM_017007706.1 XM_017007707.1 XM_017007708.1 XM_017007709.1 XM_017007710.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3603271 c.777C>T p.S259S Chr.4: 2898219-2898219 on Build GRCh38
1495796 c.899G>C p.R300P Chr.4: 2898446-2898446 on Build GRCh38
5047868 c.905A>G p.H302R Chr.4: 2898452-2898452 on Build GRCh38
3603272 c.777C>T p.S259S Chr.4: 2898219-2898219 on Build GRCh38
5175811 c.899G>A p.R300Q Chr.4: 2898446-2898446 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780039355 Chr.4: 2898346-2898346 on Build GRCh38
rs752858644 Chr.4: 2898205-2898205 on Build GRCh38
rs780723149 Chr.4: 2898477-2898477 on Build GRCh38
rs750443352 Chr.4: 2898445-2898445 on Build GRCh38
rs773882600 Chr.4: 2898162-2898162 on Build GRCh38
rs1043552038 Chr.4: 2898407-2898407 on Build GRCh38
rs980802365 Chr.4: 2898122-2898122 on Build GRCh38
rs756208883 Chr.4: 2898208-2898208 on Build GRCh38
rs1038706433 Chr.4: 2898263-2898263 on Build GRCh38
rs753930563 Chr.4: 2898331-2898331 on Build GRCh38
rs754031100 Chr.4: 2898420-2898420 on Build GRCh38
rs759997340 Chr.4: 2898387-2898387 on Build GRCh38
rs1006381446 Chr.4: 2898497-2898497 on Build GRCh38
rs745783830 Chr.4: 2898399-2898399 on Build GRCh38
rs759831736 Chr.4: 2898302-2898302 on Build GRCh38
rs551701080 Chr.4: 2898480-2898480 on Build GRCh38
rs764302827 Chr.4: 2898419-2898419 on Build GRCh38
rs867660137 Chr.4: 2898290-2898290 on Build GRCh38
rs776334018 Chr.4: 2898286-2898286 on Build GRCh38
rs372354688 Chr.4: 2898314-2898314 on Build GRCh38
rs754755610 Chr.4: 2898467-2898467 on Build GRCh38
rs1016665300 Chr.4: 2898217-2898217 on Build GRCh38
rs746781628 Chr.4: 2898349-2898349 on Build GRCh38
rs768484562 Chr.4: 2898364-2898364 on Build GRCh38
rs761817740 Chr.4: 2898422-2898422 on Build GRCh38
rs747618884 Chr.4: 2898378-2898378 on Build GRCh38
rs4971 Chr.4: 2898250-2898250 on Build GRCh38
rs887730609 Chr.4: 2898227-2898227 on Build GRCh38
rs926837565 Chr.4: 2898456-2898456 on Build GRCh38
rs145444405 Chr.4: 2898340-2898340 on Build GRCh38
rs372777117 Chr.4: 2898494-2898494 on Build GRCh38
rs771690920 Chr.4: 2898136-2898136 on Build GRCh38
rs765359110 Chr.4: 2898572-2898572 on Build GRCh38
rs764286338 Chr.4: 2898330-2898330 on Build GRCh38
rs202124018 Chr.4: 2898446-2898446 on Build GRCh38
rs997117092 Chr.4: 2898296-2898296 on Build GRCh38
rs761021688 Chr.4: 2898415-2898415 on Build GRCh38
rs780129873 Chr.4: 2898459-2898459 on Build GRCh38
rs200904553 Chr.4: 2898265-2898265 on Build GRCh38
rs540870548 Chr.4: 2898124-2898124 on Build GRCh38
rs766884352 Chr.4: 2898184-2898184 on Build GRCh38
rs763652826 Chr.4: 2898149-2898149 on Build GRCh38
rs377154696 Chr.4: 2898389-2898389 on Build GRCh38
rs759364800 Chr.4: 2898135-2898135 on Build GRCh38
rs564277533 Chr.4: 2898166-2898166 on Build GRCh38
rs199940053 Chr.4: 2898226-2898226 on Build GRCh38
rs775906982 Chr.4: 2898501-2898501 on Build GRCh38
rs566037365 Chr.4: 2898457-2898457 on Build GRCh38
rs144809220 Chr.4: 2898279-2898279 on Build GRCh38
rs769012762 Chr.4: 2898538-2898538 on Build GRCh38
rs778903128 Chr.4: 2898228-2898228 on Build GRCh38
rs548954532 Chr.4: 2898343-2898343 on Build GRCh38
rs746733652 Chr.4: 2898123-2898123 on Build GRCh38
rs765696659 Chr.4: 2898372-2898372 on Build GRCh38
rs777870723 Chr.4: 2898214-2898214 on Build GRCh38
rs765316520 Chr.4: 2898427-2898427 on Build GRCh38
rs760278723 Chr.4: 2898144-2898144 on Build GRCh38
rs373602091 Chr.4: 2898138-2898138 on Build GRCh38
rs889184367 Chr.4: 2898413-2898413 on Build GRCh38
rs767731754 Chr.4: 2898202-2898202 on Build GRCh38
rs563417710 Chr.4: 2898300-2898300 on Build GRCh38
rs868277399 Chr.4: 2898360-2898360 on Build GRCh38
rs747789048 Chr.4: 2898478-2898478 on Build GRCh38
rs767821435 Chr.4: 2898309-2898309 on Build GRCh38
rs771931191 Chr.4: 2898268-2898268 on Build GRCh38
rs147521356 Chr.4: 2898460-2898460 on Build GRCh38
rs371614186 Chr.4: 2898222-2898222 on Build GRCh38
rs368155471 Chr.4: 2898288-2898288 on Build GRCh38
rs745766050 Chr.4: 2898342-2898342 on Build GRCh38
rs772631725 Chr.4: 2898499-2898499 on Build GRCh38
rs993803357 Chr.4: 2898123-2898123 on Build GRCh38
rs757292746 Chr.4: 2898338-2898338 on Build GRCh38
rs1056579146 Chr.4: 2898145-2898145 on Build GRCh38
rs367745191 Chr.4: 2898199-2898199 on Build GRCh38
rs374826670 Chr.4: 2898530-2898530 on Build GRCh38
rs745651446 Chr.4: 2898245-2898245 on Build GRCh38
rs776746947 Chr.4: 2898539-2898539 on Build GRCh38
rs761038171 Chr.4: 2898514-2898514 on Build GRCh38
rs776344429 Chr.4: 2898373-2898373 on Build GRCh38
rs774742702 Chr.4: 2898385-2898385 on Build GRCh38
rs1022451736 Chr.4: 2898347-2898347 on Build GRCh38
rs374634966 Chr.4: 2898271-2898271 on Build GRCh38
rs755368185 Chr.4: 2898200-2898200 on Build GRCh38
rs1007470627 Chr.4: 2898262-2898262 on Build GRCh38
rs374411956 Chr.4: 2898298-2898298 on Build GRCh38
rs762131516 Chr.4: 2898563-2898563 on Build GRCh38
rs776755028 Chr.4: 2898475-2898475 on Build GRCh38
rs980416472 Chr.4: 2898243-2898243 on Build GRCh38
rs750578656 Chr.4: 2898573-2898573 on Build GRCh38
rs756376615 Chr.4: 2898315-2898315 on Build GRCh38
rs775890518 Chr.4: 2898404-2898404 on Build GRCh38
rs897884647 Chr.4: 2898167-2898167 on Build GRCh38

View Full Product Details