Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 7042000-7042504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC96
Entrez Gene ID
257236
Gene Name
coiled-coil domain containing 96
Gene Aliases
-
UniGene
Hs.656757
Gene Chromosome Location
Chr.4: 7040849-7043001 on Build GRCh38
Transcripts
NM_153376.2 NR_033828.1

Gene Symbol
LOC100129931
Entrez Gene ID
100129931
Gene Name
uncharacterized LOC100129931
Gene Aliases
-
UniGene
Hs.707753
Gene Chromosome Location
Chr.4: 7030554-7046231 on Build GRCh38
Transcripts
NM_153376.2 NR_033828.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6330977 c.559G>A p.G187R Chr.4: 7042380-7042380 on Build GRCh38
4670131 c.504G>T p.E168D Chr.4: 7042435-7042435 on Build GRCh38
2829753 c.458A>G p.E153G Chr.4: 7042481-7042481 on Build GRCh38
2829749 c.896C>T p.A299V Chr.4: 7042043-7042043 on Build GRCh38
6311856 c.772C>T p.R258C Chr.4: 7042167-7042167 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369759265 Chr.4: 7042226-7042226 on Build GRCh38
rs765789625 Chr.4: 7042364-7042364 on Build GRCh38
rs890109660 Chr.4: 7042191-7042191 on Build GRCh38
rs754856312 Chr.4: 7042290-7042290 on Build GRCh38
rs766840027 Chr.4: 7042042-7042042 on Build GRCh38
rs375336133 Chr.4: 7042225-7042225 on Build GRCh38
rs757691825 Chr.4: 7042113-7042113 on Build GRCh38
rs758250249 Chr.4: 7042453-7042453 on Build GRCh38
rs760913132 Chr.4: 7042227-7042227 on Build GRCh38
rs757439679 Chr.4: 7042024-7042024 on Build GRCh38
rs945157957 Chr.4: 7042412-7042412 on Build GRCh38
rs775356893 Chr.4: 7042108-7042108 on Build GRCh38
rs368710664 Chr.4: 7042316-7042316 on Build GRCh38
rs904197839 Chr.4: 7042449-7042449 on Build GRCh38
rs763575585 Chr.4: 7042327-7042327 on Build GRCh38
rs181515863 Chr.4: 7042218-7042218 on Build GRCh38
rs768301828 Chr.4: 7042184-7042184 on Build GRCh38
rs771913930 Chr.4: 7042086-7042086 on Build GRCh38
rs747147340 Chr.4: 7042422-7042422 on Build GRCh38
rs755458335 Chr.4: 7042071-7042071 on Build GRCh38
rs141859161 Chr.4: 7042294-7042294 on Build GRCh38
rs766550264 Chr.4: 7042234-7042234 on Build GRCh38
rs765930839 Chr.4: 7042439-7042439 on Build GRCh38
rs764528349 Chr.4: 7042424-7042424 on Build GRCh38
rs543659626 Chr.4: 7042213-7042213 on Build GRCh38
rs371063673 Chr.4: 7042267-7042267 on Build GRCh38
rs775051750 Chr.4: 7042432-7042432 on Build GRCh38
rs761930296 Chr.4: 7042342-7042342 on Build GRCh38
rs1051723762 Chr.4: 7042187-7042187 on Build GRCh38
rs753590452 Chr.4: 7042365-7042365 on Build GRCh38
rs749069171 Chr.4: 7042183-7042183 on Build GRCh38
rs776971237 Chr.4: 7042311-7042311 on Build GRCh38
rs374287436 Chr.4: 7042299-7042299 on Build GRCh38
rs377461773 Chr.4: 7042137-7042137 on Build GRCh38
rs748263193 Chr.4: 7042403-7042404 on Build GRCh38
rs758850837 Chr.4: 7042031-7042032 on Build GRCh38
rs778794847 Chr.4: 7042369-7042369 on Build GRCh38
rs147077157 Chr.4: 7042438-7042438 on Build GRCh38
rs534252962 Chr.4: 7042366-7042366 on Build GRCh38
rs549220952 Chr.4: 7042390-7042390 on Build GRCh38
rs981156598 Chr.4: 7042326-7042326 on Build GRCh38
rs1000346404 Chr.4: 7042451-7042451 on Build GRCh38
rs775952467 Chr.4: 7042415-7042415 on Build GRCh38
rs750608989 Chr.4: 7042043-7042043 on Build GRCh38
rs781467139 Chr.4: 7042371-7042372 on Build GRCh38
rs754692962 Chr.4: 7042444-7042444 on Build GRCh38
rs59204906 Chr.4: 7042051-7042051 on Build GRCh38
rs746489810 Chr.4: 7042089-7042089 on Build GRCh38
rs777172785 Chr.4: 7042074-7042074 on Build GRCh38
rs755731779 Chr.4: 7042345-7042345 on Build GRCh38
rs754155331 Chr.4: 7042129-7042129 on Build GRCh38
rs139612326 Chr.4: 7042254-7042254 on Build GRCh38
rs770317293 Chr.4: 7042092-7042092 on Build GRCh38
rs879145223 Chr.4: 7042351-7042351 on Build GRCh38
rs764841283 Chr.4: 7042446-7042446 on Build GRCh38
rs936390227 Chr.4: 7042038-7042038 on Build GRCh38
rs577899866 Chr.4: 7042461-7042461 on Build GRCh38
rs758160715 Chr.4: 7042386-7042386 on Build GRCh38
rs139316839 Chr.4: 7042198-7042198 on Build GRCh38
rs201971169 Chr.4: 7042096-7042096 on Build GRCh38
rs868326302 Chr.4: 7042091-7042091 on Build GRCh38
rs999951915 Chr.4: 7042244-7042244 on Build GRCh38
rs751623687 Chr.4: 7042105-7042105 on Build GRCh38
rs750714848 Chr.4: 7042127-7042127 on Build GRCh38
rs769858588 Chr.4: 7042410-7042410 on Build GRCh38
rs781354755 Chr.4: 7042463-7042463 on Build GRCh38
rs533110978 Chr.4: 7042242-7042242 on Build GRCh38
rs755472502 Chr.4: 7042130-7042130 on Build GRCh38
rs926644626 Chr.4: 7042332-7042332 on Build GRCh38
rs756849015 Chr.4: 7042150-7042150 on Build GRCh38
rs550858107 Chr.4: 7042214-7042214 on Build GRCh38
rs199844890 Chr.4: 7042044-7042044 on Build GRCh38
rs370554794 Chr.4: 7042347-7042347 on Build GRCh38
rs956904773 Chr.4: 7042297-7042297 on Build GRCh38
rs747859864 Chr.4: 7042385-7042385 on Build GRCh38
rs942142748 Chr.4: 7042132-7042132 on Build GRCh38
rs142049391 Chr.4: 7042103-7042103 on Build GRCh38
rs769884585 Chr.4: 7042480-7042480 on Build GRCh38
rs114895671 Chr.4: 7042068-7042068 on Build GRCh38
rs376178176 Chr.4: 7042177-7042177 on Build GRCh38
rs780297516 Chr.4: 7042383-7042383 on Build GRCh38
rs760823406 Chr.4: 7042128-7042128 on Build GRCh38
rs748866058 Chr.4: 7042280-7042280 on Build GRCh38
rs1052840563 Chr.4: 7042456-7042456 on Build GRCh38
rs753978103 Chr.4: 7042241-7042241 on Build GRCh38
rs770977069 Chr.4: 7042308-7042308 on Build GRCh38
rs746409481 Chr.4: 7042124-7042124 on Build GRCh38
rs917909855 Chr.4: 7042105-7042105 on Build GRCh38
rs752325111 Chr.4: 7042447-7042447 on Build GRCh38
rs781041540 Chr.4: 7042098-7042098 on Build GRCh38
rs752078199 Chr.4: 7042342-7042342 on Build GRCh38
rs771065049 Chr.4: 7042394-7042394 on Build GRCh38
rs753313893 Chr.4: 7042260-7042260 on Build GRCh38
rs764654703 Chr.4: 7042341-7042341 on Build GRCh38
rs201135458 Chr.4: 7042462-7042462 on Build GRCh38
rs755859167 Chr.4: 7042277-7042277 on Build GRCh38
rs567489271 Chr.4: 7042399-7042399 on Build GRCh38
rs375345995 Chr.4: 7042173-7042173 on Build GRCh38
rs769512219 Chr.4: 7042174-7042174 on Build GRCh38
rs200732240 Chr.4: 7042116-7042116 on Build GRCh38
rs1015688672 Chr.4: 7042161-7042161 on Build GRCh38
rs747133690 Chr.4: 7042300-7042300 on Build GRCh38
rs370637587 Chr.4: 7042159-7042159 on Build GRCh38
rs75719146 Chr.4: 7042279-7042279 on Build GRCh38
rs561818755 Chr.4: 7042138-7042138 on Build GRCh38
rs146317749 Chr.4: 7042189-7042189 on Build GRCh38
rs372351063 Chr.4: 7042441-7042441 on Build GRCh38
rs746113925 Chr.4: 7042404-7042404 on Build GRCh38
rs759673956 Chr.4: 7042312-7042312 on Build GRCh38
rs745894418 Chr.4: 7042466-7042466 on Build GRCh38
rs997077695 Chr.4: 7042107-7042107 on Build GRCh38
rs749902765 Chr.4: 7042270-7042270 on Build GRCh38
rs919088701 Chr.4: 7042370-7042370 on Build GRCh38
rs374259671 Chr.4: 7042246-7042246 on Build GRCh38
rs780845168 Chr.4: 7042172-7042172 on Build GRCh38
rs767736461 Chr.4: 7042034-7042034 on Build GRCh38
rs762809305 Chr.4: 7042109-7042109 on Build GRCh38

View Full Product Details