Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 15718766-15719304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BST1
Entrez Gene ID
683
Gene Name
bone marrow stromal cell antigen 1
Gene Aliases
CD157
UniGene
Hs.720344
Gene Chromosome Location
Chr.4: 15686347-15774177 on Build GRCh38
Transcripts
NM_004334.2 XM_005248185.1 XM_005248186.1 XM_011513878.2 XM_011513879.1 XM_011513881.2 XM_017008565.1 XM_017008566.1 XM_017008567.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7122446 c.774C>T p.S258S Chr.4: 15718976-15718976 on Build GRCh38
4584874 c.727A>G p.M243V Chr.4: 15718929-15718929 on Build GRCh38
3129839 c.761G>A p.G254E Chr.4: 15718963-15718963 on Build GRCh38
3129838 c.735C>T p.V245V Chr.4: 15718937-15718937 on Build GRCh38
5865961 c.725G>A p.S242N Chr.4: 15718927-15718927 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770921286 Chr.4: 15718957-15718957 on Build GRCh38
rs757658291 Chr.4: 15719036-15719036 on Build GRCh38
rs925800718 Chr.4: 15719071-15719071 on Build GRCh38
rs765162961 Chr.4: 15719010-15719010 on Build GRCh38
rs371427757 Chr.4: 15719280-15719280 on Build GRCh38
rs553991686 Chr.4: 15719264-15719264 on Build GRCh38
rs571638897 Chr.4: 15719187-15719187 on Build GRCh38
rs767973076 Chr.4: 15718955-15718955 on Build GRCh38
rs146288579 Chr.4: 15718916-15718916 on Build GRCh38
rs549220748 Chr.4: 15719087-15719087 on Build GRCh38
rs773037252 Chr.4: 15718992-15718992 on Build GRCh38
rs1033179031 Chr.4: 15719206-15719206 on Build GRCh38
rs562456632 Chr.4: 15718855-15718855 on Build GRCh38
rs200411213 Chr.4: 15718949-15718949 on Build GRCh38
rs753956494 Chr.4: 15718867-15718867 on Build GRCh38
rs1006583717 Chr.4: 15719245-15719245 on Build GRCh38
rs534117100 Chr.4: 15719196-15719196 on Build GRCh38
rs371481195 Chr.4: 15718799-15718799 on Build GRCh38
rs888565392 Chr.4: 15719098-15719098 on Build GRCh38
rs758794842 Chr.4: 15718898-15718898 on Build GRCh38
rs888457133 Chr.4: 15718809-15718809 on Build GRCh38
rs1020917404 Chr.4: 15719146-15719146 on Build GRCh38
rs376767442 Chr.4: 15718964-15718964 on Build GRCh38
rs918444020 Chr.4: 15719070-15719070 on Build GRCh38
rs758887306 Chr.4: 15719028-15719028 on Build GRCh38
rs370948510 Chr.4: 15718993-15718993 on Build GRCh38
rs537871580 Chr.4: 15719119-15719119 on Build GRCh38
rs534744267 Chr.4: 15718844-15718844 on Build GRCh38
rs935843706 Chr.4: 15719091-15719091 on Build GRCh38
rs937402872 Chr.4: 15719100-15719100 on Build GRCh38
rs896107545 Chr.4: 15718861-15718861 on Build GRCh38
rs996241005 Chr.4: 15718870-15718870 on Build GRCh38
rs1000808661 Chr.4: 15719205-15719205 on Build GRCh38
rs1015466574 Chr.4: 15719195-15719195 on Build GRCh38
rs748097807 Chr.4: 15718963-15718963 on Build GRCh38
rs753086388 Chr.4: 15718893-15718893 on Build GRCh38
rs1034341306 Chr.4: 15718851-15718851 on Build GRCh38
rs34163939 Chr.4: 15718917-15718917 on Build GRCh38
rs754971020 Chr.4: 15718871-15718871 on Build GRCh38
rs765793572 Chr.4: 15718891-15718891 on Build GRCh38
rs75812843 Chr.4: 15719061-15719061 on Build GRCh38
rs749106641 Chr.4: 15718866-15718866 on Build GRCh38
rs1040418567 Chr.4: 15719112-15719112 on Build GRCh38
rs756469603 Chr.4: 15719052-15719052 on Build GRCh38
rs560382910 Chr.4: 15719067-15719067 on Build GRCh38
rs149266397 Chr.4: 15719060-15719060 on Build GRCh38
rs374266490 Chr.4: 15719020-15719020 on Build GRCh38
rs748864788 Chr.4: 15718929-15718929 on Build GRCh38
rs891826770 Chr.4: 15718832-15718832 on Build GRCh38
rs1049128310 Chr.4: 15719137-15719137 on Build GRCh38
rs184195255 Chr.4: 15719077-15719077 on Build GRCh38
rs759673773 Chr.4: 15718999-15718999 on Build GRCh38
rs910870661 Chr.4: 15719023-15719023 on Build GRCh38
rs766617549 Chr.4: 15718994-15718994 on Build GRCh38
rs1015174676 Chr.4: 15718961-15718961 on Build GRCh38
rs1048481174 Chr.4: 15719095-15719095 on Build GRCh38
rs565207752 Chr.4: 15719045-15719045 on Build GRCh38
rs977872560 Chr.4: 15718924-15718924 on Build GRCh38
rs903360564 Chr.4: 15719190-15719190 on Build GRCh38
rs551633356 Chr.4: 15719171-15719171 on Build GRCh38
rs774364119 Chr.4: 15718959-15718959 on Build GRCh38
rs1005044648 Chr.4: 15719177-15719177 on Build GRCh38
rs776783544 Chr.4: 15718995-15718995 on Build GRCh38
rs935944054 Chr.4: 15718797-15718797 on Build GRCh38
rs1014525917 Chr.4: 15719124-15719124 on Build GRCh38
rs530454248 Chr.4: 15719107-15719107 on Build GRCh38
rs867381056 Chr.4: 15719069-15719069 on Build GRCh38
rs764677327 Chr.4: 15719032-15719032 on Build GRCh38
rs1032474688 Chr.4: 15718934-15718934 on Build GRCh38
rs779824817 Chr.4: 15718918-15718918 on Build GRCh38
rs913157983 Chr.4: 15719105-15719105 on Build GRCh38
rs1028252197 Chr.4: 15718885-15718885 on Build GRCh38
rs751454804 Chr.4: 15719005-15719005 on Build GRCh38
rs1013761010 Chr.4: 15719217-15719217 on Build GRCh38
rs866242780 Chr.4: 15719225-15719225 on Build GRCh38
rs757334277 Chr.4: 15718877-15718877 on Build GRCh38

View Full Product Details