Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 24806035-24806545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC149
Entrez Gene ID
91050
Gene Name
coiled-coil domain containing 149
Gene Aliases
-
UniGene
Hs.106432
Gene Chromosome Location
Chr.4: 24806117-24980204 on Build GRCh38
Transcripts
NM_001130726.2 NM_173463.4 XM_005248210.3 XM_011513906.2 XM_011513907.2 XM_011513908.1 XM_017008827.1 XM_017008828.1

SNP Detail

SNP ID Chromosome Location
rs1036017708 Chr.4: 24806100-24806100 on Build GRCh38
rs149654839 Chr.4: 24806516-24806516 on Build GRCh38
rs567214454 Chr.4: 24806199-24806199 on Build GRCh38
rs1022248785 Chr.4: 24806302-24806302 on Build GRCh38
rs769405688 Chr.4: 24806198-24806198 on Build GRCh38
rs182770101 Chr.4: 24806416-24806416 on Build GRCh38
rs756445326 Chr.4: 24806112-24806113 on Build GRCh38
rs113632496 Chr.4: 24806206-24806206 on Build GRCh38
rs748020190 Chr.4: 24806140-24806140 on Build GRCh38
rs185290807 Chr.4: 24806193-24806193 on Build GRCh38
rs2324580 Chr.4: 24806369-24806369 on Build GRCh38
rs1002892090 Chr.4: 24806286-24806286 on Build GRCh38
rs530350136 Chr.4: 24806136-24806136 on Build GRCh38
rs971847723 Chr.4: 24806447-24806447 on Build GRCh38
rs536089639 Chr.4: 24806411-24806411 on Build GRCh38
rs753136599 Chr.4: 24806464-24806464 on Build GRCh38
rs140783926 Chr.4: 24806434-24806434 on Build GRCh38
rs762459781 Chr.4: 24806308-24806308 on Build GRCh38
rs764316677 Chr.4: 24806471-24806471 on Build GRCh38
rs768081762 Chr.4: 24806458-24806458 on Build GRCh38
rs1009639170 Chr.4: 24806289-24806289 on Build GRCh38
rs552550089 Chr.4: 24806231-24806231 on Build GRCh38
rs187310531 Chr.4: 24806465-24806465 on Build GRCh38
rs573634710 Chr.4: 24806405-24806405 on Build GRCh38
rs898322459 Chr.4: 24806200-24806201 on Build GRCh38
rs771420965 Chr.4: 24806412-24806412 on Build GRCh38
rs922201562 Chr.4: 24806478-24806478 on Build GRCh38
rs1022871793 Chr.4: 24806248-24806248 on Build GRCh38
rs557053057 Chr.4: 24806370-24806370 on Build GRCh38
rs34066951 Chr.4: 24806415-24806415 on Build GRCh38
rs960884412 Chr.4: 24806415-24806415 on Build GRCh38
rs770537338 Chr.4: 24806200-24806200 on Build GRCh38
rs190547499 Chr.4: 24806222-24806222 on Build GRCh38
rs10805251 Chr.4: 24806084-24806084 on Build GRCh38
rs1040230823 Chr.4: 24806276-24806276 on Build GRCh38
rs558507995 Chr.4: 24806290-24806290 on Build GRCh38
rs142417106 Chr.4: 24806124-24806124 on Build GRCh38
rs910441202 Chr.4: 24806503-24806503 on Build GRCh38
rs13129946 Chr.4: 24806282-24806282 on Build GRCh38
rs1034362971 Chr.4: 24806397-24806397 on Build GRCh38
rs930544692 Chr.4: 24806192-24806192 on Build GRCh38
rs1050690809 Chr.4: 24806115-24806115 on Build GRCh38
rs908739823 Chr.4: 24806431-24806431 on Build GRCh38

View Full Product Details