Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 24837141-24837648 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC149
Entrez Gene ID
91050
Gene Name
coiled-coil domain containing 149
Gene Aliases
-
UniGene
Hs.106432
Gene Chromosome Location
Chr.4: 24806117-24980204 on Build GRCh38
Transcripts
NM_001130726.2 NM_173463.4 XM_005248210.3 XM_011513906.2 XM_011513907.2 XM_011513908.1 XM_017008827.1 XM_017008828.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7285151 c.477C>T p.D159D Chr.4: 24837248-24837248 on Build GRCh38
6100150 c.432G>T p.L144L Chr.4: 24837293-24837293 on Build GRCh38
4960613 c.582G>A p.R194R Chr.4: 24837308-24837308 on Build GRCh38
3603040 c.364G>C p.E122Q Chr.4: 24837361-24837361 on Build GRCh38
6512074 c.508G>T p.D170Y Chr.4: 24837382-24837382 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs927461268 Chr.4: 24837578-24837578 on Build GRCh38
rs771268292 Chr.4: 24837430-24837430 on Build GRCh38
rs754591249 Chr.4: 24837214-24837214 on Build GRCh38
rs538367240 Chr.4: 24837516-24837516 on Build GRCh38
rs202018132 Chr.4: 24837382-24837382 on Build GRCh38
rs897752633 Chr.4: 24837514-24837514 on Build GRCh38
rs931734760 Chr.4: 24837417-24837417 on Build GRCh38
rs1036312558 Chr.4: 24837481-24837481 on Build GRCh38
rs757231145 Chr.4: 24837378-24837378 on Build GRCh38
rs762264627 Chr.4: 24837301-24837301 on Build GRCh38
rs753515249 Chr.4: 24837203-24837203 on Build GRCh38
rs764772506 Chr.4: 24837215-24837215 on Build GRCh38
rs144305412 Chr.4: 24837271-24837271 on Build GRCh38
rs954037775 Chr.4: 24837190-24837190 on Build GRCh38
rs764498345 Chr.4: 24837178-24837178 on Build GRCh38
rs756621811 Chr.4: 24837415-24837415 on Build GRCh38
rs780726716 Chr.4: 24837220-24837220 on Build GRCh38
rs775028675 Chr.4: 24837438-24837438 on Build GRCh38
rs533907448 Chr.4: 24837308-24837308 on Build GRCh38
rs770877513 Chr.4: 24837585-24837585 on Build GRCh38
rs749473340 Chr.4: 24837398-24837398 on Build GRCh38
rs762091842 Chr.4: 24837201-24837201 on Build GRCh38
rs763611050 Chr.4: 24837350-24837350 on Build GRCh38
rs775699302 Chr.4: 24837265-24837265 on Build GRCh38
rs750233654 Chr.4: 24837383-24837383 on Build GRCh38
rs199797697 Chr.4: 24837254-24837254 on Build GRCh38
rs979371211 Chr.4: 24837576-24837576 on Build GRCh38
rs369672838 Chr.4: 24837420-24837420 on Build GRCh38
rs774850176 Chr.4: 24837183-24837183 on Build GRCh38
rs770122005 Chr.4: 24837264-24837264 on Build GRCh38
rs762626313 Chr.4: 24837449-24837449 on Build GRCh38
rs913515761 Chr.4: 24837567-24837567 on Build GRCh38
rs58692972 Chr.4: 24837362-24837362 on Build GRCh38
rs371812850 Chr.4: 24837337-24837337 on Build GRCh38
rs149187284 Chr.4: 24837272-24837272 on Build GRCh38
rs1019544338 Chr.4: 24837515-24837515 on Build GRCh38
rs1008732198 Chr.4: 24837469-24837469 on Build GRCh38
rs1002722759 Chr.4: 24837577-24837577 on Build GRCh38
rs747474506 Chr.4: 24837238-24837238 on Build GRCh38
rs748724226 Chr.4: 24837262-24837263 on Build GRCh38
rs146803158 Chr.4: 24837336-24837336 on Build GRCh38
rs761029952 Chr.4: 24837424-24837424 on Build GRCh38
rs180741946 Chr.4: 24837287-24837287 on Build GRCh38
rs143719102 Chr.4: 24837303-24837303 on Build GRCh38
rs369087231 Chr.4: 24837248-24837248 on Build GRCh38
rs764863024 Chr.4: 24837333-24837333 on Build GRCh38
rs779015464 Chr.4: 24837412-24837412 on Build GRCh38
rs770119292 Chr.4: 24837306-24837306 on Build GRCh38
rs989039927 Chr.4: 24837537-24837537 on Build GRCh38
rs376023103 Chr.4: 24837386-24837386 on Build GRCh38
rs188323020 Chr.4: 24837202-24837202 on Build GRCh38
rs748700027 Chr.4: 24837416-24837416 on Build GRCh38
rs763567768 Chr.4: 24837175-24837175 on Build GRCh38
rs539608551 Chr.4: 24837466-24837466 on Build GRCh38
rs1014830297 Chr.4: 24837209-24837209 on Build GRCh38
rs1013467539 Chr.4: 24837377-24837377 on Build GRCh38
rs954153237 Chr.4: 24837477-24837477 on Build GRCh38
rs1006948383 Chr.4: 24837505-24837505 on Build GRCh38
rs968908476 Chr.4: 24837569-24837569 on Build GRCh38
rs200792138 Chr.4: 24837253-24837253 on Build GRCh38
rs780147944 Chr.4: 24837395-24837395 on Build GRCh38
rs768645366 Chr.4: 24837400-24837400 on Build GRCh38
rs746908590 Chr.4: 24837242-24837242 on Build GRCh38
rs937640047 Chr.4: 24837314-24837314 on Build GRCh38

View Full Product Details