Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 36281536-36282031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTHD1
Entrez Gene ID
401124
Gene Name
death domain containing 1
Gene Aliases
-
UniGene
Hs.363407
Gene Chromosome Location
Chr.4: 36281593-36347511 on Build GRCh38
Transcripts
NM_001136536.4 XM_006714014.3 XM_011513693.2 XM_011513694.2 XM_011513695.2 XM_011513696.2 XM_017008191.1 XM_017008192.1 XM_017008193.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1429498 c.17+2C>T p.? Chr.4: 36282031-36282031 on Build GRCh38
3696632 c.9C>T p.L3L Chr.4: 36282021-36282021 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1019944479 Chr.4: 36281676-36281676 on Build GRCh38
rs914726961 Chr.4: 36281808-36281808 on Build GRCh38
rs937555842 Chr.4: 36281867-36281867 on Build GRCh38
rs576995653 Chr.4: 36281779-36281779 on Build GRCh38
rs995193584 Chr.4: 36281699-36281699 on Build GRCh38
rs1057233389 Chr.4: 36281891-36281891 on Build GRCh38
rs959237057 Chr.4: 36282007-36282007 on Build GRCh38
rs937581267 Chr.4: 36281783-36281783 on Build GRCh38
rs927581734 Chr.4: 36281858-36281858 on Build GRCh38
rs908244934 Chr.4: 36281744-36281744 on Build GRCh38
rs1018748595 Chr.4: 36281753-36281753 on Build GRCh38
rs982517902 Chr.4: 36281741-36281741 on Build GRCh38
rs572049223 Chr.4: 36281834-36281834 on Build GRCh38
rs903228833 Chr.4: 36281893-36281893 on Build GRCh38
rs370704809 Chr.4: 36281915-36281915 on Build GRCh38
rs1041775532 Chr.4: 36281847-36281847 on Build GRCh38
rs944042071 Chr.4: 36281897-36281897 on Build GRCh38
rs970379494 Chr.4: 36281757-36281757 on Build GRCh38
rs369416543 Chr.4: 36281722-36281722 on Build GRCh38
rs755760827 Chr.4: 36281760-36281760 on Build GRCh38
rs147471410 Chr.4: 36281768-36281768 on Build GRCh38
rs7695130 Chr.4: 36281691-36281691 on Build GRCh38
rs1033326910 Chr.4: 36281988-36281988 on Build GRCh38
rs888275504 Chr.4: 36281922-36281922 on Build GRCh38
rs917414067 Chr.4: 36281923-36281923 on Build GRCh38
rs928858002 Chr.4: 36281574-36281574 on Build GRCh38
rs1041723650 Chr.4: 36281652-36281652 on Build GRCh38
rs980821778 Chr.4: 36281845-36281845 on Build GRCh38
rs564849323 Chr.4: 36281825-36281825 on Build GRCh38
rs7671251 Chr.4: 36281653-36281653 on Build GRCh38
rs953387078 Chr.4: 36281732-36281732 on Build GRCh38
rs1008131045 Chr.4: 36281656-36281656 on Build GRCh38
rs937509795 Chr.4: 36281587-36281587 on Build GRCh38
rs914938767 Chr.4: 36281806-36281806 on Build GRCh38
rs1036394424 Chr.4: 36281958-36281958 on Build GRCh38
rs937543592 Chr.4: 36281617-36281617 on Build GRCh38
rs940952976 Chr.4: 36281568-36281568 on Build GRCh38
rs527686960 Chr.4: 36281623-36281623 on Build GRCh38
rs186371797 Chr.4: 36281562-36281562 on Build GRCh38
rs981734746 Chr.4: 36281763-36281763 on Build GRCh38
rs1015355583 Chr.4: 36281941-36281941 on Build GRCh38
rs10013944 Chr.4: 36281953-36281953 on Build GRCh38
rs748863988 Chr.4: 36281985-36281985 on Build GRCh38
rs952595735 Chr.4: 36281740-36281740 on Build GRCh38
rs940813928 Chr.4: 36281930-36281930 on Build GRCh38
rs1041014888 Chr.4: 36281902-36281902 on Build GRCh38
rs369115562 Chr.4: 36281590-36281590 on Build GRCh38
rs1011591684 Chr.4: 36281635-36281635 on Build GRCh38
rs931777233 Chr.4: 36281599-36281600 on Build GRCh38
rs1026798485 Chr.4: 36281727-36281727 on Build GRCh38
rs970433328 Chr.4: 36281943-36281943 on Build GRCh38
rs899759137 Chr.4: 36281907-36281907 on Build GRCh38
rs1027264032 Chr.4: 36281916-36281916 on Build GRCh38
rs569944301 Chr.4: 36281694-36281694 on Build GRCh38
rs906048854 Chr.4: 36281609-36281609 on Build GRCh38
rs914907508 Chr.4: 36281592-36281592 on Build GRCh38
rs140029129 Chr.4: 36281967-36281967 on Build GRCh38
rs1004006675 Chr.4: 36281934-36281934 on Build GRCh38
rs534785133 Chr.4: 36281909-36281909 on Build GRCh38
rs1046006688 Chr.4: 36281573-36281573 on Build GRCh38
rs947502910 Chr.4: 36281809-36281809 on Build GRCh38
rs553148810 Chr.4: 36281969-36281969 on Build GRCh38
rs987880169 Chr.4: 36281774-36281774 on Build GRCh38
rs149406847 Chr.4: 36281599-36281599 on Build GRCh38
rs1056384338 Chr.4: 36281799-36281799 on Build GRCh38

View Full Product Details