Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 36294603-36295122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTHD1
Entrez Gene ID
401124
Gene Name
death domain containing 1
Gene Aliases
-
UniGene
Hs.363407
Gene Chromosome Location
Chr.4: 36281593-36347511 on Build GRCh38
Transcripts
NM_001136536.4 NM_001170700.2 XM_006714014.3 XM_011513693.2 XM_011513694.2 XM_011513695.2 XM_011513696.2 XM_017008191.1 XM_017008192.1 XM_017008193.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
160607 c.610C>G p.L204V Chr.4: 36294876-36294876 on Build GRCh38
7002766 c.1255T>G p.S419A Chr.4: 36295026-36295026 on Build GRCh38
7412034 c.770A>G p.N257S Chr.4: 36295036-36295036 on Build GRCh38
1429502 c.668T>C p.L223S Chr.4: 36294934-36294934 on Build GRCh38
7002767 c.760T>G p.S254A Chr.4: 36295026-36295026 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1006425300 Chr.4: 36294844-36294844 on Build GRCh38
rs991482123 Chr.4: 36294651-36294651 on Build GRCh38
rs958970039 Chr.4: 36294647-36294647 on Build GRCh38
rs769724268 Chr.4: 36294830-36294830 on Build GRCh38
rs767042064 Chr.4: 36294970-36294970 on Build GRCh38
rs756413588 Chr.4: 36294979-36294979 on Build GRCh38
rs914498826 Chr.4: 36294659-36294659 on Build GRCh38
rs374555683 Chr.4: 36295028-36295028 on Build GRCh38
rs1043369396 Chr.4: 36294725-36294725 on Build GRCh38
rs1043055500 Chr.4: 36295017-36295017 on Build GRCh38
rs74616570 Chr.4: 36295071-36295071 on Build GRCh38
rs78418755 Chr.4: 36295070-36295070 on Build GRCh38
rs554345243 Chr.4: 36294653-36294653 on Build GRCh38
rs866698643 Chr.4: 36294873-36294873 on Build GRCh38
rs763048359 Chr.4: 36294872-36294872 on Build GRCh38
rs755519744 Chr.4: 36295005-36295005 on Build GRCh38
rs905747243 Chr.4: 36294753-36294753 on Build GRCh38
rs1054316512 Chr.4: 36294718-36294731 on Build GRCh38
rs776388146 Chr.4: 36295030-36295030 on Build GRCh38
rs766789305 Chr.4: 36294986-36294986 on Build GRCh38
rs775552771 Chr.4: 36294838-36294838 on Build GRCh38
rs75111589 Chr.4: 36295069-36295069 on Build GRCh38
rs932331513 Chr.4: 36294784-36294784 on Build GRCh38
rs1027656421 Chr.4: 36294898-36294898 on Build GRCh38
rs74786126 Chr.4: 36295093-36295093 on Build GRCh38
rs776034617 Chr.4: 36294897-36294897 on Build GRCh38
rs912711755 Chr.4: 36294948-36294948 on Build GRCh38
rs746352706 Chr.4: 36294928-36294928 on Build GRCh38
rs754339190 Chr.4: 36294994-36294994 on Build GRCh38
rs747996318 Chr.4: 36295059-36295059 on Build GRCh38
rs779417332 Chr.4: 36295012-36295012 on Build GRCh38
rs773778842 Chr.4: 36294918-36294918 on Build GRCh38
rs866210741 Chr.4: 36294807-36294807 on Build GRCh38
rs772136247 Chr.4: 36294789-36294789 on Build GRCh38
rs199975273 Chr.4: 36294865-36294865 on Build GRCh38
rs189410090 Chr.4: 36295038-36295038 on Build GRCh38
rs1036531579 Chr.4: 36294845-36294845 on Build GRCh38
rs747066255 Chr.4: 36294744-36294744 on Build GRCh38
rs185536233 Chr.4: 36294642-36294642 on Build GRCh38
rs543438501 Chr.4: 36294768-36294768 on Build GRCh38
rs977339400 Chr.4: 36295023-36295023 on Build GRCh38
rs920995004 Chr.4: 36294779-36294779 on Build GRCh38
rs758507951 Chr.4: 36295021-36295021 on Build GRCh38
rs769705093 Chr.4: 36295039-36295039 on Build GRCh38
rs777500404 Chr.4: 36294717-36294717 on Build GRCh38
rs746654237 Chr.4: 36294720-36294720 on Build GRCh38
rs750097471 Chr.4: 36294973-36294973 on Build GRCh38
rs748372231 Chr.4: 36294781-36294781 on Build GRCh38
rs1003370127 Chr.4: 36294632-36294632 on Build GRCh38
rs748136362 Chr.4: 36295020-36295020 on Build GRCh38
rs35618738 Chr.4: 36294952-36294952 on Build GRCh38
rs986070785 Chr.4: 36294929-36294929 on Build GRCh38
rs780163054 Chr.4: 36294704-36294704 on Build GRCh38
rs563179017 Chr.4: 36294808-36294808 on Build GRCh38
rs1003012905 Chr.4: 36294869-36294869 on Build GRCh38
rs1013364174 Chr.4: 36294981-36294981 on Build GRCh38
rs943813843 Chr.4: 36294764-36294764 on Build GRCh38
rs775461053 Chr.4: 36295053-36295053 on Build GRCh38
rs561023019 Chr.4: 36294664-36294664 on Build GRCh38
rs533626351 Chr.4: 36294965-36294965 on Build GRCh38
rs958801424 Chr.4: 36294916-36294916 on Build GRCh38
rs888063339 Chr.4: 36294821-36294821 on Build GRCh38
rs745581607 Chr.4: 36294809-36294809 on Build GRCh38
rs146277370 Chr.4: 36294754-36294754 on Build GRCh38

View Full Product Details