Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 36308103-36308610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTHD1
Entrez Gene ID
401124
Gene Name
death domain containing 1
Gene Aliases
-
UniGene
Hs.363407
Gene Chromosome Location
Chr.4: 36281593-36347511 on Build GRCh38
Transcripts
NM_001136536.4 NM_001170700.2 XM_006714014.3 XM_011513693.2 XM_011513694.2 XM_011513695.2 XM_011513696.2 XM_017008191.1 XM_017008192.1 XM_017008193.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
447814 c.1177G>A p.E393K Chr.4: 36308445-36308445 on Build GRCh38
4470303 c.1157C>T p.S386F Chr.4: 36308425-36308425 on Build GRCh38
5703032 c.1214T>A p.I405K Chr.4: 36308482-36308482 on Build GRCh38
1429508 c.1117G>A p.D373N Chr.4: 36308385-36308385 on Build GRCh38
1727988 c.1523C>T p.A508V Chr.4: 36308296-36308296 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563835721 Chr.4: 36308275-36308275 on Build GRCh38
rs911353379 Chr.4: 36308296-36308296 on Build GRCh38
rs900726166 Chr.4: 36308413-36308413 on Build GRCh38
rs937962562 Chr.4: 36308219-36308219 on Build GRCh38
rs781583416 Chr.4: 36308215-36308215 on Build GRCh38
rs750691784 Chr.4: 36308442-36308442 on Build GRCh38
rs771480211 Chr.4: 36308265-36308265 on Build GRCh38
rs753137234 Chr.4: 36308375-36308375 on Build GRCh38
rs542690773 Chr.4: 36308480-36308480 on Build GRCh38
rs757598775 Chr.4: 36308180-36308180 on Build GRCh38
rs1042448705 Chr.4: 36308298-36308298 on Build GRCh38
rs749054756 Chr.4: 36308217-36308217 on Build GRCh38
rs201839505 Chr.4: 36308401-36308401 on Build GRCh38
rs908333748 Chr.4: 36308129-36308129 on Build GRCh38
rs775716091 Chr.4: 36308404-36308404 on Build GRCh38
rs1053903285 Chr.4: 36308355-36308355 on Build GRCh38
rs200787538 Chr.4: 36308341-36308341 on Build GRCh38
rs1041114036 Chr.4: 36308308-36308308 on Build GRCh38
rs1028369845 Chr.4: 36308418-36308418 on Build GRCh38
rs867402448 Chr.4: 36308523-36308523 on Build GRCh38
rs187161267 Chr.4: 36308324-36308324 on Build GRCh38
rs955067649 Chr.4: 36308425-36308425 on Build GRCh38
rs942783392 Chr.4: 36308300-36308300 on Build GRCh38
rs961141007 Chr.4: 36308438-36308438 on Build GRCh38
rs917931531 Chr.4: 36308273-36308273 on Build GRCh38
rs746447611 Chr.4: 36308505-36308505 on Build GRCh38
rs770529433 Chr.4: 36308326-36308326 on Build GRCh38
rs778835630 Chr.4: 36308458-36308458 on Build GRCh38
rs1042396378 Chr.4: 36308537-36308537 on Build GRCh38
rs961656631 Chr.4: 36308145-36308145 on Build GRCh38
rs374066496 Chr.4: 36308416-36308416 on Build GRCh38
rs536272239 Chr.4: 36308227-36308227 on Build GRCh38
rs61739432 Chr.4: 36308443-36308443 on Build GRCh38
rs12507599 Chr.4: 36308427-36308427 on Build GRCh38
rs1048544394 Chr.4: 36308251-36308251 on Build GRCh38
rs971700996 Chr.4: 36308451-36308451 on Build GRCh38
rs922681413 Chr.4: 36308570-36308570 on Build GRCh38
rs554575267 Chr.4: 36308228-36308228 on Build GRCh38
rs748178118 Chr.4: 36308250-36308250 on Build GRCh38
rs759836208 Chr.4: 36308346-36308346 on Build GRCh38
rs917865362 Chr.4: 36308510-36308510 on Build GRCh38
rs868814877 Chr.4: 36308364-36308364 on Build GRCh38
rs976860039 Chr.4: 36308568-36308568 on Build GRCh38
rs748005265 Chr.4: 36308461-36308461 on Build GRCh38
rs751698031 Chr.4: 36308156-36308156 on Build GRCh38
rs140254738 Chr.4: 36308232-36308232 on Build GRCh38
rs73117691 Chr.4: 36308354-36308354 on Build GRCh38
rs369210518 Chr.4: 36308428-36308428 on Build GRCh38
rs1041727993 Chr.4: 36308581-36308581 on Build GRCh38
rs115555517 Chr.4: 36308233-36308233 on Build GRCh38
rs150903588 Chr.4: 36308511-36308511 on Build GRCh38
rs763551614 Chr.4: 36308387-36308387 on Build GRCh38
rs377338124 Chr.4: 36308573-36308573 on Build GRCh38
rs375987946 Chr.4: 36308454-36308454 on Build GRCh38
rs942834400 Chr.4: 36308550-36308550 on Build GRCh38
rs973931401 Chr.4: 36308173-36308173 on Build GRCh38
rs963612933 Chr.4: 36308569-36308569 on Build GRCh38
rs777595338 Chr.4: 36308497-36308497 on Build GRCh38
rs952996813 Chr.4: 36308460-36308460 on Build GRCh38
rs376898600 Chr.4: 36308220-36308220 on Build GRCh38
rs1034205141 Chr.4: 36308421-36308421 on Build GRCh38
rs772819020 Chr.4: 36308291-36308291 on Build GRCh38

View Full Product Details