Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 48499888-48500407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRYL
Entrez Gene ID
285527
Gene Name
FRY like transcription coactivator
Gene Aliases
AF4p12, KIAA0826, MOR2
UniGene
Hs.595553
Gene Chromosome Location
Chr.4: 48497363-48780312 on Build GRCh38
Transcripts
NM_015030.1 XM_005248090.4 XM_005248093.4 XM_011513680.2 XM_011513681.2 XM_011513682.2 XM_011513683.2 XM_011513684.2 XM_017008046.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2849073 c.8783G>C p.R2928T Chr.4: 48500030-48500030 on Build GRCh38
1241192 c.8600A>G p.N2867S Chr.4: 48500213-48500213 on Build GRCh38
7191966 c.8657C>A p.S2886Y Chr.4: 48500156-48500156 on Build GRCh38
5014422 c.8764G>A p.A2922T Chr.4: 48500049-48500049 on Build GRCh38
5401280 c.8635C>T p.L2879F Chr.4: 48500178-48500178 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562452192 Chr.4: 48500211-48500211 on Build GRCh38
rs181257019 Chr.4: 48499979-48499979 on Build GRCh38
rs748062017 Chr.4: 48500251-48500251 on Build GRCh38
rs571789225 Chr.4: 48499934-48499934 on Build GRCh38
rs778278290 Chr.4: 48500205-48500205 on Build GRCh38
rs953979163 Chr.4: 48500287-48500287 on Build GRCh38
rs757579235 Chr.4: 48500216-48500216 on Build GRCh38
rs1003059027 Chr.4: 48500178-48500178 on Build GRCh38
rs781430421 Chr.4: 48500227-48500227 on Build GRCh38
rs759080724 Chr.4: 48500311-48500311 on Build GRCh38
rs188895645 Chr.4: 48500149-48500149 on Build GRCh38
rs745907238 Chr.4: 48500135-48500135 on Build GRCh38
rs181814536 Chr.4: 48500319-48500319 on Build GRCh38
rs1022082979 Chr.4: 48500258-48500258 on Build GRCh38
rs772060329 Chr.4: 48500252-48500252 on Build GRCh38
rs773133549 Chr.4: 48500270-48500270 on Build GRCh38
rs375724636 Chr.4: 48500155-48500155 on Build GRCh38
rs749489262 Chr.4: 48500296-48500296 on Build GRCh38
rs774664519 Chr.4: 48500332-48500332 on Build GRCh38
rs187249022 Chr.4: 48500325-48500325 on Build GRCh38
rs748961044 Chr.4: 48500268-48500269 on Build GRCh38
rs759034087 Chr.4: 48500191-48500191 on Build GRCh38
rs747572215 Chr.4: 48499985-48499988 on Build GRCh38
rs747222952 Chr.4: 48500079-48500079 on Build GRCh38
rs753277835 Chr.4: 48500053-48500053 on Build GRCh38
rs546254706 Chr.4: 48500018-48500018 on Build GRCh38
rs758794120 Chr.4: 48500059-48500059 on Build GRCh38
rs777236055 Chr.4: 48500266-48500267 on Build GRCh38
rs765789722 Chr.4: 48500004-48500004 on Build GRCh38
rs559700547 Chr.4: 48500341-48500341 on Build GRCh38
rs779417959 Chr.4: 48500023-48500023 on Build GRCh38
rs746111141 Chr.4: 48500234-48500234 on Build GRCh38
rs540707705 Chr.4: 48500154-48500154 on Build GRCh38
rs576992241 Chr.4: 48500167-48500167 on Build GRCh38
rs912418218 Chr.4: 48499987-48499987 on Build GRCh38
rs569528521 Chr.4: 48500016-48500016 on Build GRCh38
rs62619178 Chr.4: 48500184-48500184 on Build GRCh38
rs200708718 Chr.4: 48500160-48500160 on Build GRCh38
rs781275636 Chr.4: 48500133-48500133 on Build GRCh38
rs769789021 Chr.4: 48500146-48500146 on Build GRCh38
rs769494934 Chr.4: 48500006-48500006 on Build GRCh38
rs918355082 Chr.4: 48500111-48500111 on Build GRCh38
rs952467376 Chr.4: 48500374-48500374 on Build GRCh38
rs540689433 Chr.4: 48500087-48500087 on Build GRCh38
rs780154880 Chr.4: 48500243-48500243 on Build GRCh38
rs965161086 Chr.4: 48500349-48500349 on Build GRCh38
rs767650657 Chr.4: 48500371-48500371 on Build GRCh38
rs954099936 Chr.4: 48499976-48499976 on Build GRCh38
rs867610050 Chr.4: 48500172-48500172 on Build GRCh38
rs185751021 Chr.4: 48500071-48500071 on Build GRCh38
rs142593800 Chr.4: 48500151-48500151 on Build GRCh38
rs188385001 Chr.4: 48499970-48499970 on Build GRCh38
rs759035579 Chr.4: 48500163-48500163 on Build GRCh38
rs756189820 Chr.4: 48500238-48500238 on Build GRCh38
rs373666417 Chr.4: 48500161-48500161 on Build GRCh38
rs760365675 Chr.4: 48500001-48500001 on Build GRCh38
rs972554361 Chr.4: 48500355-48500355 on Build GRCh38
rs759486083 Chr.4: 48500359-48500359 on Build GRCh38
rs771251980 Chr.4: 48500112-48500112 on Build GRCh38

View Full Product Details