Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70628666-70629178 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENAM
Entrez Gene ID
10117
Gene Name
enamelin
Gene Aliases
ADAI, AI1C, AIH2
UniGene
Hs.667018
Gene Chromosome Location
Chr.4: 70627471-70646819 on Build GRCh38
Transcripts
NM_031889.2 XM_006714056.3

SNP Detail

SNP ID Chromosome Location
rs916207987 Chr.4: 70628865-70628865 on Build GRCh38
rs902288165 Chr.4: 70628904-70628904 on Build GRCh38
rs909117386 Chr.4: 70628819-70628819 on Build GRCh38
rs949109951 Chr.4: 70628875-70628875 on Build GRCh38
rs998628914 Chr.4: 70629093-70629093 on Build GRCh38
rs899776067 Chr.4: 70628846-70628846 on Build GRCh38
rs151125662 Chr.4: 70629088-70629088 on Build GRCh38
rs983279033 Chr.4: 70628811-70628811 on Build GRCh38
rs943683100 Chr.4: 70628757-70628757 on Build GRCh38
rs142969834 Chr.4: 70629048-70629048 on Build GRCh38
rs145814085 Chr.4: 70628764-70628764 on Build GRCh38
rs913572577 Chr.4: 70628690-70628690 on Build GRCh38
rs893992558 Chr.4: 70629060-70629060 on Build GRCh38
rs897843739 Chr.4: 70629085-70629085 on Build GRCh38
rs756356063 Chr.4: 70628975-70628975 on Build GRCh38
rs1018946292 Chr.4: 70629071-70629071 on Build GRCh38
rs990702461 Chr.4: 70628863-70628863 on Build GRCh38
rs935468390 Chr.4: 70628934-70628934 on Build GRCh38
rs1007114594 Chr.4: 70629063-70629063 on Build GRCh38
rs1041372247 Chr.4: 70628829-70628829 on Build GRCh38
rs886059580 Chr.4: 70628807-70628807 on Build GRCh38
rs950573608 Chr.4: 70628807-70628807 on Build GRCh38
rs941921398 Chr.4: 70628847-70628847 on Build GRCh38
rs984180578 Chr.4: 70629146-70629146 on Build GRCh38
rs1993579 Chr.4: 70628902-70628902 on Build GRCh38
rs951490803 Chr.4: 70629142-70629142 on Build GRCh38
rs886059582 Chr.4: 70628914-70628914 on Build GRCh38
rs566616822 Chr.4: 70629106-70629106 on Build GRCh38
rs886059581 Chr.4: 70628913-70628913 on Build GRCh38
rs969707105 Chr.4: 70629110-70629110 on Build GRCh38

View Full Product Details