Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70629250-70629748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENAM
Entrez Gene ID
10117
Gene Name
enamelin
Gene Aliases
ADAI, AI1C, AIH2
UniGene
Hs.667018
Gene Chromosome Location
Chr.4: 70627471-70646819 on Build GRCh38
Transcripts
NM_031889.2 XM_006714056.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5205930 c.10_11insT p.R5fs*10 Chr.4: 70629510-70629511 on Build GRCh38
7150039 c.20G>A p.R7K Chr.4: 70629520-70629520 on Build GRCh38
4680944 c.22C>A p.L8I Chr.4: 70629522-70629522 on Build GRCh38
1137198 c.39T>G p.P13P Chr.4: 70629539-70629539 on Build GRCh38
4680945 c.53T>C p.L18S Chr.4: 70629553-70629553 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs771999287 Chr.4: 70629459-70629459 on Build GRCh38
rs781501514 Chr.4: 70629566-70629566 on Build GRCh38
rs761432111 Chr.4: 70629529-70629529 on Build GRCh38
rs776093568 Chr.4: 70629582-70629582 on Build GRCh38
rs758546918 Chr.4: 70629554-70629554 on Build GRCh38
rs543158403 Chr.4: 70629665-70629665 on Build GRCh38
rs142329753 Chr.4: 70629313-70629313 on Build GRCh38
rs564475011 Chr.4: 70629694-70629694 on Build GRCh38
rs771475268 Chr.4: 70629478-70629478 on Build GRCh38
rs923568044 Chr.4: 70629697-70629697 on Build GRCh38
rs749590630 Chr.4: 70629540-70629540 on Build GRCh38
rs182835987 Chr.4: 70629437-70629437 on Build GRCh38
rs779281301 Chr.4: 70629657-70629657 on Build GRCh38
rs963172139 Chr.4: 70629621-70629621 on Build GRCh38
rs767353566 Chr.4: 70629531-70629531 on Build GRCh38
rs909718258 Chr.4: 70629469-70629469 on Build GRCh38
rs753680535 Chr.4: 70629553-70629553 on Build GRCh38
rs770340121 Chr.4: 70629513-70629513 on Build GRCh38
rs942886482 Chr.4: 70629443-70629443 on Build GRCh38
rs1032531961 Chr.4: 70629636-70629636 on Build GRCh38
rs574877044 Chr.4: 70629421-70629421 on Build GRCh38
rs746260829 Chr.4: 70629567-70629567 on Build GRCh38
rs777978626 Chr.4: 70629557-70629557 on Build GRCh38
rs763552842 Chr.4: 70629515-70629515 on Build GRCh38
rs1023533471 Chr.4: 70629653-70629653 on Build GRCh38
rs982208663 Chr.4: 70629278-70629278 on Build GRCh38
rs1009719280 Chr.4: 70629664-70629664 on Build GRCh38
rs956362477 Chr.4: 70629698-70629698 on Build GRCh38
rs778175689 Chr.4: 70629476-70629476 on Build GRCh38
rs750213131 Chr.4: 70629532-70629532 on Build GRCh38
rs958212105 Chr.4: 70629639-70629639 on Build GRCh38
rs1014086848 Chr.4: 70629558-70629558 on Build GRCh38
rs971027128 Chr.4: 70629693-70629693 on Build GRCh38
rs777122356 Chr.4: 70629480-70629480 on Build GRCh38
rs748261893 Chr.4: 70629456-70629456 on Build GRCh38
rs770007828 Chr.4: 70629568-70629568 on Build GRCh38
rs920924230 Chr.4: 70629331-70629331 on Build GRCh38
rs778923094 Chr.4: 70629450-70629450 on Build GRCh38
rs546065827 Chr.4: 70629589-70629589 on Build GRCh38
rs768200116 Chr.4: 70629591-70629591 on Build GRCh38
rs1026044731 Chr.4: 70629506-70629506 on Build GRCh38
rs775804067 Chr.4: 70629514-70629514 on Build GRCh38
rs747206681 Chr.4: 70629477-70629477 on Build GRCh38
rs146402422 Chr.4: 70629405-70629405 on Build GRCh38
rs780052281 Chr.4: 70629551-70629551 on Build GRCh38
rs932616675 Chr.4: 70629337-70629337 on Build GRCh38
rs200390024 Chr.4: 70629548-70629548 on Build GRCh38
rs905159533 Chr.4: 70629580-70629580 on Build GRCh38
rs554151732 Chr.4: 70629444-70629444 on Build GRCh38
rs773833086 Chr.4: 70629602-70629602 on Build GRCh38
rs760029562 Chr.4: 70629497-70629497 on Build GRCh38
rs771278744 Chr.4: 70629561-70629561 on Build GRCh38
rs991088184 Chr.4: 70629627-70629627 on Build GRCh38
rs998181343 Chr.4: 70629486-70629486 on Build GRCh38
rs761520315 Chr.4: 70629606-70629606 on Build GRCh38
rs751252664 Chr.4: 70629526-70629526 on Build GRCh38
rs563529697 Chr.4: 70629716-70629716 on Build GRCh38
rs935039584 Chr.4: 70629382-70629382 on Build GRCh38

View Full Product Details