Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 71754461-71755007 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GC
Entrez Gene ID
2638
Gene Name
GC, vitamin D binding protein
Gene Aliases
DBP, DBP/GC, GRD3, Gc-MAF, GcMAF, HEL-S-51, VDBG, VDBP
UniGene
Hs.418497
Gene Chromosome Location
Chr.4: 71741693-71805520 on Build GRCh38
Transcripts
NM_000583.3 NM_001204306.1 NM_001204307.1 XM_006714177.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5866700 c.1136A>C p.E379A Chr.4: 71755006-71755006 on Build GRCh38
3392948 c.1219A>G p.K407E Chr.4: 71754980-71754980 on Build GRCh38
5401494 c.1183G>A p.E395K Chr.4: 71754490-71754490 on Build GRCh38
3721901 c.1199C>G p.S400* Chr.4: 71755000-71755000 on Build GRCh38
5045322 c.1135G>A p.E379K Chr.4: 71755007-71755007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1046917621 Chr.4: 71754653-71754653 on Build GRCh38
rs879087577 Chr.4: 71754976-71754976 on Build GRCh38
rs377573160 Chr.4: 71754599-71754600 on Build GRCh38
rs982258168 Chr.4: 71754838-71754838 on Build GRCh38
rs142448358 Chr.4: 71754495-71754495 on Build GRCh38
rs762519132 Chr.4: 71754961-71754961 on Build GRCh38
rs541898001 Chr.4: 71754692-71754692 on Build GRCh38
rs755201392 Chr.4: 71754546-71754546 on Build GRCh38
rs530168528 Chr.4: 71754654-71754654 on Build GRCh38
rs532433982 Chr.4: 71754946-71754946 on Build GRCh38
rs1038193053 Chr.4: 71754972-71754972 on Build GRCh38
rs768097606 Chr.4: 71754965-71754965 on Build GRCh38
rs761684766 Chr.4: 71754980-71754980 on Build GRCh38
rs571405700 Chr.4: 71754931-71754931 on Build GRCh38
rs769360876 Chr.4: 71754674-71754674 on Build GRCh38
rs926162022 Chr.4: 71754755-71754755 on Build GRCh38
rs368943930 Chr.4: 71754541-71754541 on Build GRCh38
rs754055486 Chr.4: 71754542-71754542 on Build GRCh38
rs1042250707 Chr.4: 71754736-71754736 on Build GRCh38
rs775348510 Chr.4: 71754960-71754960 on Build GRCh38
rs764119415 Chr.4: 71754525-71754525 on Build GRCh38
rs899178492 Chr.4: 71754756-71754756 on Build GRCh38
rs942610985 Chr.4: 71754506-71754506 on Build GRCh38
rs762704585 Chr.4: 71754795-71754795 on Build GRCh38
rs754247794 Chr.4: 71754703-71754703 on Build GRCh38
rs917292793 Chr.4: 71754683-71754683 on Build GRCh38
rs79444700 Chr.4: 71754840-71754840 on Build GRCh38
rs535412686 Chr.4: 71754597-71754598 on Build GRCh38
rs975142493 Chr.4: 71754876-71754876 on Build GRCh38
rs933931711 Chr.4: 71754913-71754913 on Build GRCh38
rs562889416 Chr.4: 71754632-71754632 on Build GRCh38
rs886954178 Chr.4: 71754926-71754926 on Build GRCh38
rs781413286 Chr.4: 71754945-71754945 on Build GRCh38
rs200525599 Chr.4: 71754494-71754494 on Build GRCh38
rs531426883 Chr.4: 71754816-71754816 on Build GRCh38
rs952603226 Chr.4: 71754879-71754879 on Build GRCh38
rs900973826 Chr.4: 71754563-71754563 on Build GRCh38
rs1016236869 Chr.4: 71754742-71754742 on Build GRCh38
rs368318594 Chr.4: 71754949-71754949 on Build GRCh38
rs922506065 Chr.4: 71754886-71754886 on Build GRCh38
rs560769002 Chr.4: 71754704-71754704 on Build GRCh38
rs1031203694 Chr.4: 71754839-71754839 on Build GRCh38
rs758541386 Chr.4: 71754929-71754929 on Build GRCh38
rs186634163 Chr.4: 71754537-71754537 on Build GRCh38
rs1046865786 Chr.4: 71754925-71754925 on Build GRCh38
rs762608701 Chr.4: 71754502-71754502 on Build GRCh38
rs952711737 Chr.4: 71754583-71754583 on Build GRCh38
rs989654550 Chr.4: 71754849-71754849 on Build GRCh38
rs1034268668 Chr.4: 71754907-71754907 on Build GRCh38
rs766130195 Chr.4: 71754800-71754800 on Build GRCh38
rs771278551 Chr.4: 71754943-71754943 on Build GRCh38
rs752764207 Chr.4: 71754553-71754553 on Build GRCh38
rs774292637 Chr.4: 71754970-71754970 on Build GRCh38
rs747519094 Chr.4: 71754932-71754932 on Build GRCh38
rs914874309 Chr.4: 71754856-71754856 on Build GRCh38
rs761316644 Chr.4: 71754577-71754577 on Build GRCh38
rs971002784 Chr.4: 71754834-71754834 on Build GRCh38
rs372194035 Chr.4: 71754528-71754528 on Build GRCh38
rs767234420 Chr.4: 71754540-71754540 on Build GRCh38
rs984077044 Chr.4: 71754606-71754606 on Build GRCh38
rs1012460679 Chr.4: 71754803-71754803 on Build GRCh38
rs546498722 Chr.4: 71754860-71754860 on Build GRCh38
rs59000278 Chr.4: 71754969-71754970 on Build GRCh38
rs112727179 Chr.4: 71754709-71754709 on Build GRCh38

View Full Product Details