Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 71755000-71755512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GC
Entrez Gene ID
2638
Gene Name
GC, vitamin D binding protein
Gene Aliases
DBP, DBP/GC, GRD3, Gc-MAF, GcMAF, HEL-S-51, VDBG, VDBP
UniGene
Hs.418497
Gene Chromosome Location
Chr.4: 71741693-71805520 on Build GRCh38
Transcripts
NM_000583.3 NM_001204306.1 NM_001204307.1 XM_006714177.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5866700 c.1136A>C p.E379A Chr.4: 71755006-71755006 on Build GRCh38
3605596 c.1129G>A p.D377N Chr.4: 71755013-71755013 on Build GRCh38
4969433 c.1122T>A p.H374Q Chr.4: 71755077-71755077 on Build GRCh38
1540896 c.1096G>T p.E366* Chr.4: 71755046-71755046 on Build GRCh38
5736990 c.1070C>T p.P357L Chr.4: 71755072-71755072 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866426372 Chr.4: 71755415-71755415 on Build GRCh38
rs986265978 Chr.4: 71755353-71755353 on Build GRCh38
rs996624916 Chr.4: 71755414-71755414 on Build GRCh38
rs528273370 Chr.4: 71755307-71755307 on Build GRCh38
rs371860432 Chr.4: 71755146-71755148 on Build GRCh38
rs756109741 Chr.4: 71755313-71755313 on Build GRCh38
rs892758663 Chr.4: 71755346-71755346 on Build GRCh38
rs756171443 Chr.4: 71755233-71755233 on Build GRCh38
rs58603194 Chr.4: 71755177-71755178 on Build GRCh38
rs773204732 Chr.4: 71755113-71755113 on Build GRCh38
rs571440091 Chr.4: 71755220-71755220 on Build GRCh38
rs929700042 Chr.4: 71755148-71755148 on Build GRCh38
rs544872395 Chr.4: 71755193-71755193 on Build GRCh38
rs1022482754 Chr.4: 71755371-71755371 on Build GRCh38
rs377233580 Chr.4: 71755390-71755390 on Build GRCh38
rs765442276 Chr.4: 71755028-71755028 on Build GRCh38
rs1038240656 Chr.4: 71755288-71755288 on Build GRCh38
rs1029262436 Chr.4: 71755445-71755445 on Build GRCh38
rs559100492 Chr.4: 71755174-71755174 on Build GRCh38
rs117492633 Chr.4: 71755091-71755091 on Build GRCh38
rs750739005 Chr.4: 71755024-71755024 on Build GRCh38
rs899243610 Chr.4: 71755302-71755302 on Build GRCh38
rs200641845 Chr.4: 71755178-71755178 on Build GRCh38
rs754718134 Chr.4: 71755149-71755156 on Build GRCh38
rs781120578 Chr.4: 71755155-71755156 on Build GRCh38
rs76803094 Chr.4: 71755071-71755071 on Build GRCh38
rs568820346 Chr.4: 71755046-71755046 on Build GRCh38
rs773741845 Chr.4: 71755093-71755093 on Build GRCh38
rs748968666 Chr.4: 71755083-71755083 on Build GRCh38
rs368970048 Chr.4: 71755323-71755323 on Build GRCh38
rs541934907 Chr.4: 71755265-71755265 on Build GRCh38
rs568783825 Chr.4: 71755428-71755428 on Build GRCh38
rs986381592 Chr.4: 71755052-71755052 on Build GRCh38
rs894366273 Chr.4: 71755255-71755255 on Build GRCh38
rs888168807 Chr.4: 71755423-71755423 on Build GRCh38
rs574462312 Chr.4: 71755245-71755245 on Build GRCh38
rs184061554 Chr.4: 71755222-71755222 on Build GRCh38
rs111227171 Chr.4: 71755335-71755335 on Build GRCh38
rs587776830 Chr.4: 71755171-71755174 on Build GRCh38
rs761960440 Chr.4: 71755084-71755086 on Build GRCh38
rs550831840 Chr.4: 71755406-71755406 on Build GRCh38
rs922299442 Chr.4: 71755237-71755237 on Build GRCh38
rs752958552 Chr.4: 71755037-71755037 on Build GRCh38
rs775643300 Chr.4: 71755131-71755131 on Build GRCh38
rs1011491411 Chr.4: 71755350-71755350 on Build GRCh38
rs71673486 Chr.4: 71755148-71755149 on Build GRCh38
rs139868816 Chr.4: 71755287-71755287 on Build GRCh38
rs1047172332 Chr.4: 71755421-71755421 on Build GRCh38
rs764175036 Chr.4: 71755143-71755143 on Build GRCh38
rs910738981 Chr.4: 71755367-71755367 on Build GRCh38
rs369015898 Chr.4: 71755048-71755048 on Build GRCh38
rs1045443727 Chr.4: 71755308-71755308 on Build GRCh38
rs757717977 Chr.4: 71755057-71755057 on Build GRCh38
rs763116380 Chr.4: 71755138-71755138 on Build GRCh38
rs888274455 Chr.4: 71755223-71755223 on Build GRCh38
rs1055528671 Chr.4: 71755462-71755462 on Build GRCh38
rs770318450 Chr.4: 71755077-71755077 on Build GRCh38
rs377318672 Chr.4: 71755141-71755141 on Build GRCh38
rs908342956 Chr.4: 71755285-71755285 on Build GRCh38
rs145351863 Chr.4: 71755324-71755324 on Build GRCh38
rs112725080 Chr.4: 71755108-71755108 on Build GRCh38
rs760432782 Chr.4: 71755114-71755114 on Build GRCh38
rs532339036 Chr.4: 71755352-71755352 on Build GRCh38
rs529682474 Chr.4: 71755429-71755429 on Build GRCh38
rs139133782 Chr.4: 71755072-71755072 on Build GRCh38
rs758537485 Chr.4: 71755063-71755063 on Build GRCh38
rs149359930 Chr.4: 71755107-71755107 on Build GRCh38
rs748063583 Chr.4: 71755102-71755102 on Build GRCh38
rs976359039 Chr.4: 71755380-71755380 on Build GRCh38
rs549780517 Chr.4: 71755187-71755208 on Build GRCh38

View Full Product Details