Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 94875714-94876228 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMPR1B
Entrez Gene ID
658
Gene Name
bone morphogenetic protein receptor type 1B
Gene Aliases
ALK-6, ALK6, AMDD, BDA1D, BDA2, CDw293
UniGene
Hs.598475
Gene Chromosome Location
Chr.4: 94757977-95158453 on Build GRCh38
Transcripts
NM_001203.2 XM_011532201.2 XM_017008558.1 XM_017008559.1

SNP Detail

SNP ID Chromosome Location
rs114050799 Chr.4: 94876158-94876158 on Build GRCh38
rs921764716 Chr.4: 94875940-94875940 on Build GRCh38
rs571345881 Chr.4: 94875910-94875910 on Build GRCh38
rs970695549 Chr.4: 94875837-94875837 on Build GRCh38
rs193168382 Chr.4: 94875760-94875760 on Build GRCh38
rs1036142688 Chr.4: 94876037-94876037 on Build GRCh38
rs958383567 Chr.4: 94876195-94876195 on Build GRCh38
rs1024940322 Chr.4: 94875815-94875815 on Build GRCh38
rs540386868 Chr.4: 94876167-94876167 on Build GRCh38
rs149226776 Chr.4: 94875792-94875792 on Build GRCh38
rs143024463 Chr.4: 94876175-94876175 on Build GRCh38
rs1025179264 Chr.4: 94876153-94876153 on Build GRCh38
rs1054569342 Chr.4: 94876064-94876064 on Build GRCh38
rs1012487763 Chr.4: 94876098-94876098 on Build GRCh38
rs891308714 Chr.4: 94876146-94876146 on Build GRCh38
rs1051672735 Chr.4: 94875949-94875949 on Build GRCh38
rs936549769 Chr.4: 94875904-94875904 on Build GRCh38
rs868679388 Chr.4: 94875737-94875737 on Build GRCh38
rs763470709 Chr.4: 94876188-94876188 on Build GRCh38
rs148290279 Chr.4: 94875884-94875884 on Build GRCh38
rs75697273 Chr.4: 94875896-94875896 on Build GRCh38
rs865802105 Chr.4: 94876065-94876065 on Build GRCh38
rs748910913 Chr.4: 94875913-94875913 on Build GRCh38
rs143230133 Chr.4: 94875831-94875831 on Build GRCh38
rs544185041 Chr.4: 94876092-94876092 on Build GRCh38
rs879440246 Chr.4: 94875969-94875969 on Build GRCh38
rs955452532 Chr.4: 94875889-94875889 on Build GRCh38
rs917076857 Chr.4: 94876030-94876030 on Build GRCh38
rs989542720 Chr.4: 94876196-94876196 on Build GRCh38
rs1027569073 Chr.4: 94876095-94876095 on Build GRCh38
rs542345437 Chr.4: 94876183-94876183 on Build GRCh38
rs536101046 Chr.4: 94875965-94875965 on Build GRCh38
rs895285217 Chr.4: 94876097-94876097 on Build GRCh38
rs1044141982 Chr.4: 94876039-94876039 on Build GRCh38
rs551682952 Chr.4: 94876090-94876090 on Build GRCh38
rs879368421 Chr.4: 94875954-94875954 on Build GRCh38
rs892066962 Chr.4: 94875826-94875826 on Build GRCh38
rs183571919 Chr.4: 94875934-94875934 on Build GRCh38
rs959633302 Chr.4: 94875794-94875794 on Build GRCh38
rs555782427 Chr.4: 94876034-94876034 on Build GRCh38
rs779589676 Chr.4: 94875912-94875912 on Build GRCh38
rs926932028 Chr.4: 94876057-94876057 on Build GRCh38
rs574853739 Chr.4: 94876163-94876163 on Build GRCh38
rs1030277740 Chr.4: 94875888-94875888 on Build GRCh38
rs76172175 Chr.4: 94875745-94875745 on Build GRCh38
rs772498912 Chr.4: 94876001-94876002 on Build GRCh38
rs376525202 Chr.4: 94875948-94875948 on Build GRCh38
rs996064921 Chr.4: 94875774-94875774 on Build GRCh38
rs929252067 Chr.4: 94875998-94875998 on Build GRCh38
rs920502408 Chr.4: 94875981-94875981 on Build GRCh38
rs553500805 Chr.4: 94875946-94875946 on Build GRCh38
rs17500892 Chr.4: 94875801-94875801 on Build GRCh38
rs898921943 Chr.4: 94876063-94876063 on Build GRCh38

View Full Product Details