Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 112264930-112265443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1AR
Entrez Gene ID
55435
Gene Name
adaptor related protein complex 1 associated regulatory protein
Gene Aliases
2C18, C4orf16, GBAR, PRO0971, gamma-BAR
UniGene
Hs.435991
Gene Chromosome Location
Chr.4: 112231727-112273110 on Build GRCh38
Transcripts
NM_001128426.2 NM_018569.5 NR_136622.1 XM_017008378.1 XM_017008379.1 XM_017008380.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1618276 c.410A>G p.Y137C Chr.4: 112265037-112265037 on Build GRCh38
1618275 c.384A>C p.Q128H Chr.4: 112265011-112265011 on Build GRCh38
3392660 c.440+1G>T p.? Chr.4: 112265068-112265068 on Build GRCh38
1660558 c.403C>T p.Q135* Chr.4: 112265030-112265030 on Build GRCh38
1050090 c.385G>T p.E129* Chr.4: 112265012-112265012 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs897940538 Chr.4: 112265386-112265386 on Build GRCh38
rs754517424 Chr.4: 112264957-112264958 on Build GRCh38
rs779005941 Chr.4: 112265083-112265083 on Build GRCh38
rs79411041 Chr.4: 112265153-112265153 on Build GRCh38
rs750176441 Chr.4: 112265086-112265088 on Build GRCh38
rs986989976 Chr.4: 112264997-112264997 on Build GRCh38
rs146737294 Chr.4: 112265156-112265156 on Build GRCh38
rs753997662 Chr.4: 112264981-112264981 on Build GRCh38
rs948140274 Chr.4: 112265232-112265232 on Build GRCh38
rs748255398 Chr.4: 112264986-112264986 on Build GRCh38
rs553034017 Chr.4: 112265371-112265371 on Build GRCh38
rs964471374 Chr.4: 112265016-112265016 on Build GRCh38
rs116585845 Chr.4: 112265042-112265042 on Build GRCh38
rs986008685 Chr.4: 112265133-112265133 on Build GRCh38
rs983767121 Chr.4: 112265139-112265139 on Build GRCh38
rs140495117 Chr.4: 112265384-112265384 on Build GRCh38
rs920259315 Chr.4: 112265082-112265082 on Build GRCh38
rs1052589720 Chr.4: 112265208-112265208 on Build GRCh38
rs781265020 Chr.4: 112264983-112264983 on Build GRCh38
rs751430964 Chr.4: 112265095-112265095 on Build GRCh38
rs779570138 Chr.4: 112265091-112265091 on Build GRCh38
rs181706282 Chr.4: 112265325-112265325 on Build GRCh38
rs779950018 Chr.4: 112264961-112264961 on Build GRCh38
rs903844056 Chr.4: 112265270-112265270 on Build GRCh38
rs770638141 Chr.4: 112265251-112265251 on Build GRCh38
rs907817181 Chr.4: 112265141-112265141 on Build GRCh38
rs758110754 Chr.4: 112265088-112265088 on Build GRCh38
rs368670552 Chr.4: 112265009-112265009 on Build GRCh38
rs746890469 Chr.4: 112265191-112265191 on Build GRCh38
rs377373110 Chr.4: 112264959-112264959 on Build GRCh38
rs999561590 Chr.4: 112265271-112265271 on Build GRCh38
rs373910360 Chr.4: 112264960-112264960 on Build GRCh38
rs924381884 Chr.4: 112265199-112265199 on Build GRCh38
rs764686481 Chr.4: 112265075-112265075 on Build GRCh38
rs34015872 Chr.4: 112265363-112265364 on Build GRCh38
rs757208545 Chr.4: 112264981-112264985 on Build GRCh38
rs777418398 Chr.4: 112265157-112265157 on Build GRCh38
rs1056456198 Chr.4: 112265385-112265385 on Build GRCh38
rs774815303 Chr.4: 112265035-112265035 on Build GRCh38
rs748063101 Chr.4: 112265120-112265120 on Build GRCh38
rs768069384 Chr.4: 112265052-112265052 on Build GRCh38
rs771034493 Chr.4: 112265027-112265027 on Build GRCh38
rs569010687 Chr.4: 112264976-112264976 on Build GRCh38
rs549384439 Chr.4: 112264980-112264981 on Build GRCh38
rs781281467 Chr.4: 112265119-112265119 on Build GRCh38
rs746752326 Chr.4: 112264963-112264963 on Build GRCh38
rs749879785 Chr.4: 112265085-112265085 on Build GRCh38
rs1031652593 Chr.4: 112265304-112265304 on Build GRCh38
rs964419934 Chr.4: 112265348-112265348 on Build GRCh38
rs993579535 Chr.4: 112265396-112265396 on Build GRCh38
rs915369352 Chr.4: 112265245-112265245 on Build GRCh38
rs369918864 Chr.4: 112265350-112265350 on Build GRCh38
rs760852547 Chr.4: 112264980-112264980 on Build GRCh38
rs1045261354 Chr.4: 112265319-112265319 on Build GRCh38
rs1018462779 Chr.4: 112265331-112265331 on Build GRCh38
rs975876897 Chr.4: 112265190-112265190 on Build GRCh38
rs754765804 Chr.4: 112265103-112265103 on Build GRCh38
rs753142668 Chr.4: 112265073-112265073 on Build GRCh38
rs776571852 Chr.4: 112265311-112265311 on Build GRCh38
rs759568655 Chr.4: 112265394-112265394 on Build GRCh38
rs111545718 Chr.4: 112265351-112265351 on Build GRCh38
rs763182366 Chr.4: 112265025-112265025 on Build GRCh38
rs752962207 Chr.4: 112264974-112264975 on Build GRCh38
rs891402452 Chr.4: 112265310-112265310 on Build GRCh38
rs769930818 Chr.4: 112264988-112264988 on Build GRCh38

View Full Product Details