Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 112266523-112267028 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1AR
Entrez Gene ID
55435
Gene Name
adaptor related protein complex 1 associated regulatory protein
Gene Aliases
2C18, C4orf16, GBAR, PRO0971, gamma-BAR
UniGene
Hs.435991
Gene Chromosome Location
Chr.4: 112231727-112273110 on Build GRCh38
Transcripts
NM_001128426.2 NM_018569.5 NR_136622.1 XM_017008378.1 XM_017008379.1 XM_017008380.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1050094 c.609C>A p.S203R Chr.4: 112266682-112266682 on Build GRCh38
1050093 c.532A>G p.T178A Chr.4: 112266605-112266605 on Build GRCh38
1050092 c.517_518delCT p.S174fs*15 Chr.4: 112266590-112266591 on Build GRCh38
1183096 c.520T>G p.S174A Chr.4: 112266593-112266593 on Build GRCh38
3006567 c.577A>G p.K193E Chr.4: 112266650-112266650 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750559912 Chr.4: 112266700-112266700 on Build GRCh38
rs568280938 Chr.4: 112266779-112266779 on Build GRCh38
rs1019679191 Chr.4: 112266889-112266889 on Build GRCh38
rs1052997787 Chr.4: 112266916-112266916 on Build GRCh38
rs759108143 Chr.4: 112266633-112266633 on Build GRCh38
rs774009103 Chr.4: 112266971-112266971 on Build GRCh38
rs746653177 Chr.4: 112266775-112266784 on Build GRCh38
rs960042440 Chr.4: 112266631-112266631 on Build GRCh38
rs770315796 Chr.4: 112266620-112266620 on Build GRCh38
rs779768891 Chr.4: 112266720-112266720 on Build GRCh38
rs956546490 Chr.4: 112266837-112266837 on Build GRCh38
rs1032711972 Chr.4: 112266783-112266783 on Build GRCh38
rs781739255 Chr.4: 112266617-112266617 on Build GRCh38
rs535725400 Chr.4: 112266800-112266800 on Build GRCh38
rs905556447 Chr.4: 112266964-112266964 on Build GRCh38
rs950944769 Chr.4: 112266712-112266712 on Build GRCh38
rs141360156 Chr.4: 112266625-112266625 on Build GRCh38
rs760521490 Chr.4: 112266650-112266650 on Build GRCh38
rs1013182511 Chr.4: 112266683-112266683 on Build GRCh38
rs931347039 Chr.4: 112266976-112266976 on Build GRCh38
rs745924800 Chr.4: 112266579-112266579 on Build GRCh38
rs758687369 Chr.4: 112266580-112266580 on Build GRCh38
rs933392224 Chr.4: 112266812-112266812 on Build GRCh38
rs780260366 Chr.4: 112266754-112266754 on Build GRCh38
rs755846177 Chr.4: 112266565-112266565 on Build GRCh38
rs575301546 Chr.4: 112266967-112266967 on Build GRCh38
rs1001232998 Chr.4: 112266970-112266970 on Build GRCh38
rs755395014 Chr.4: 112266758-112266758 on Build GRCh38
rs758416681 Chr.4: 112266734-112266734 on Build GRCh38
rs1023259676 Chr.4: 112266740-112266740 on Build GRCh38
rs559145232 Chr.4: 112266564-112266564 on Build GRCh38
rs749800833 Chr.4: 112266948-112266948 on Build GRCh38
rs185742307 Chr.4: 112266767-112266767 on Build GRCh38
rs764102584 Chr.4: 112266570-112266570 on Build GRCh38
rs975130004 Chr.4: 112266769-112266769 on Build GRCh38
rs139300460 Chr.4: 112266662-112266662 on Build GRCh38
rs887281791 Chr.4: 112266612-112266612 on Build GRCh38
rs975379340 Chr.4: 112266940-112266940 on Build GRCh38
rs75171772 Chr.4: 112266575-112266575 on Build GRCh38
rs761705111 Chr.4: 112266677-112266677 on Build GRCh38
rs751743849 Chr.4: 112266755-112266755 on Build GRCh38
rs762615107 Chr.4: 112266797-112266797 on Build GRCh38
rs748559180 Chr.4: 112266619-112266619 on Build GRCh38
rs765156038 Chr.4: 112266684-112266684 on Build GRCh38
rs190447139 Chr.4: 112266939-112266939 on Build GRCh38
rs201749009 Chr.4: 112266574-112266574 on Build GRCh38
rs987999666 Chr.4: 112266879-112266879 on Build GRCh38
rs775221472 Chr.4: 112266648-112266648 on Build GRCh38
rs768837483 Chr.4: 112266616-112266616 on Build GRCh38
rs780241899 Chr.4: 112266605-112266605 on Build GRCh38
rs534792394 Chr.4: 112266676-112266676 on Build GRCh38
rs747389238 Chr.4: 112266613-112266613 on Build GRCh38
rs1049157215 Chr.4: 112266601-112266601 on Build GRCh38

View Full Product Details