Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 112268364-112268876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1AR
Entrez Gene ID
55435
Gene Name
adaptor related protein complex 1 associated regulatory protein
Gene Aliases
2C18, C4orf16, GBAR, PRO0971, gamma-BAR
UniGene
Hs.435991
Gene Chromosome Location
Chr.4: 112231727-112273110 on Build GRCh38
Transcripts
NM_001128426.2 NM_018569.5 NR_136622.1 XM_017008378.1 XM_017008379.1 XM_017008380.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6576187 c.899A>C p.Q300P Chr.4: 112268399-112268399 on Build GRCh38
5400304 c.909G>A p.*303* Chr.4: 112268409-112268409 on Build GRCh38
1050096 c.905G>A p.R302Q Chr.4: 112268405-112268405 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs962348160 Chr.4: 112268588-112268588 on Build GRCh38
rs781381804 Chr.4: 112268831-112268831 on Build GRCh38
rs780739798 Chr.4: 112268410-112268410 on Build GRCh38
rs933749264 Chr.4: 112268655-112268655 on Build GRCh38
rs1037063959 Chr.4: 112268690-112268690 on Build GRCh38
rs1016885048 Chr.4: 112268841-112268841 on Build GRCh38
rs540730361 Chr.4: 112268678-112268678 on Build GRCh38
rs773776938 Chr.4: 112268481-112268481 on Build GRCh38
rs1014301139 Chr.4: 112268802-112268804 on Build GRCh38
rs975088559 Chr.4: 112268592-112268592 on Build GRCh38
rs749081245 Chr.4: 112268441-112268441 on Build GRCh38
rs975520238 Chr.4: 112268852-112268852 on Build GRCh38
rs200802231 Chr.4: 112268392-112268392 on Build GRCh38
rs374076320 Chr.4: 112268743-112268743 on Build GRCh38
rs34900583 Chr.4: 112268390-112268390 on Build GRCh38
rs374904998 Chr.4: 112268397-112268397 on Build GRCh38
rs552933023 Chr.4: 112268404-112268404 on Build GRCh38
rs951279466 Chr.4: 112268577-112268577 on Build GRCh38
rs771680540 Chr.4: 112268576-112268576 on Build GRCh38
rs533058438 Chr.4: 112268832-112268832 on Build GRCh38
rs369335927 Chr.4: 112268418-112268418 on Build GRCh38
rs760031744 Chr.4: 112268443-112268460 on Build GRCh38
rs770909409 Chr.4: 112268461-112268461 on Build GRCh38
rs772457728 Chr.4: 112268453-112268455 on Build GRCh38
rs944997325 Chr.4: 112268480-112268480 on Build GRCh38
rs769416777 Chr.4: 112268425-112268425 on Build GRCh38
rs1027988330 Chr.4: 112268853-112268853 on Build GRCh38
rs904764346 Chr.4: 112268713-112268713 on Build GRCh38
rs1014537533 Chr.4: 112268505-112268505 on Build GRCh38
rs962608426 Chr.4: 112268846-112268850 on Build GRCh38
rs1040753122 Chr.4: 112268496-112268496 on Build GRCh38
rs989618638 Chr.4: 112268411-112268411 on Build GRCh38
rs746515200 Chr.4: 112268405-112268405 on Build GRCh38
rs1024566640 Chr.4: 112268804-112268804 on Build GRCh38
rs773033389 Chr.4: 112268431-112268431 on Build GRCh38
rs894589557 Chr.4: 112268790-112268790 on Build GRCh38
rs1024139209 Chr.4: 112268519-112268519 on Build GRCh38
rs1041925921 Chr.4: 112268710-112268710 on Build GRCh38
rs755155758 Chr.4: 112268722-112268722 on Build GRCh38
rs1014184226 Chr.4: 112268765-112268765 on Build GRCh38
rs747752736 Chr.4: 112268419-112268419 on Build GRCh38
rs909562701 Chr.4: 112268586-112268586 on Build GRCh38
rs888274367 Chr.4: 112268666-112268666 on Build GRCh38
rs754444264 Chr.4: 112268407-112268407 on Build GRCh38
rs921043452 Chr.4: 112268635-112268635 on Build GRCh38
rs773563014 Chr.4: 112268579-112268579 on Build GRCh38
rs114615943 Chr.4: 112268668-112268668 on Build GRCh38
rs912023536 Chr.4: 112268676-112268676 on Build GRCh38
rs1000632668 Chr.4: 112268741-112268741 on Build GRCh38
rs569139415 Chr.4: 112268780-112268780 on Build GRCh38
rs897257478 Chr.4: 112268705-112268705 on Build GRCh38
rs774845871 Chr.4: 112268422-112268427 on Build GRCh38
rs958803208 Chr.4: 112268498-112268498 on Build GRCh38
rs957601735 Chr.4: 112268398-112268398 on Build GRCh38
rs747818117 Chr.4: 112268527-112268527 on Build GRCh38

View Full Product Details