Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 112411742-112412247 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALPK1
Entrez Gene ID
80216
Gene Name
alpha kinase 1
Gene Aliases
8430410J10Rik, LAK
UniGene
Hs.652825
Gene Chromosome Location
Chr.4: 112286420-112442618 on Build GRCh38
Transcripts
NM_001102406.1 NM_001253884.1 NM_025144.3 XM_005263245.4 XM_005263246.4 XM_006714326.3 XM_017008633.1 XM_017008634.1 XM_017008635.1 XM_017008636.1 XM_017008637.1 XM_017008638.1 XM_017008639.1 XM_017008640.1 XM_017008641.1 XM_017008642.1 XM_017008643.1 XM_017008644.1 XM_017008645.1 XM_017008646.1 XM_017008647.1 XM_017008648.1 XM_017008650.1 XM_017008651.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5014447 c.367G>A p.V123I Chr.4: 112411917-112411917 on Build GRCh38
4121388 c.391G>A p.A131T Chr.4: 112411941-112411941 on Build GRCh38
6845357 c.405C>A p.H135Q Chr.4: 112411955-112411955 on Build GRCh38
3993398 c.340G>A p.V114M Chr.4: 112411890-112411890 on Build GRCh38
447243 c.433C>T p.P145S Chr.4: 112411983-112411983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755828218 Chr.4: 112411893-112411893 on Build GRCh38
rs569057284 Chr.4: 112412055-112412055 on Build GRCh38
rs770023639 Chr.4: 112412048-112412048 on Build GRCh38
rs531216607 Chr.4: 112411806-112411806 on Build GRCh38
rs780322054 Chr.4: 112411913-112411913 on Build GRCh38
rs775879914 Chr.4: 112411857-112411857 on Build GRCh38
rs757383407 Chr.4: 112412114-112412114 on Build GRCh38
rs780851052 Chr.4: 112411827-112411827 on Build GRCh38
rs879241479 Chr.4: 112412151-112412151 on Build GRCh38
rs370872798 Chr.4: 112411883-112411883 on Build GRCh38
rs778659546 Chr.4: 112411854-112411854 on Build GRCh38
rs532223625 Chr.4: 112412177-112412177 on Build GRCh38
rs770738508 Chr.4: 112411785-112411785 on Build GRCh38
rs926921984 Chr.4: 112411803-112411803 on Build GRCh38
rs754970847 Chr.4: 112412031-112412031 on Build GRCh38
rs751091985 Chr.4: 112411812-112411812 on Build GRCh38
rs367762485 Chr.4: 112411961-112411961 on Build GRCh38
rs756937832 Chr.4: 112411903-112411903 on Build GRCh38
rs564905848 Chr.4: 112412131-112412131 on Build GRCh38
rs781497943 Chr.4: 112412035-112412035 on Build GRCh38
rs142248599 Chr.4: 112411973-112411973 on Build GRCh38
rs768936763 Chr.4: 112411915-112411915 on Build GRCh38
rs747594321 Chr.4: 112411864-112411865 on Build GRCh38
rs747343393 Chr.4: 112412037-112412037 on Build GRCh38
rs796377531 Chr.4: 112412074-112412074 on Build GRCh38
rs934545904 Chr.4: 112411992-112411992 on Build GRCh38
rs376090210 Chr.4: 112411786-112411786 on Build GRCh38
rs754416973 Chr.4: 112411817-112411817 on Build GRCh38
rs576303718 Chr.4: 112412111-112412111 on Build GRCh38
rs772438978 Chr.4: 112411792-112411792 on Build GRCh38
rs772484889 Chr.4: 112412014-112412014 on Build GRCh38
rs778174100 Chr.4: 112412054-112412054 on Build GRCh38
rs752055236 Chr.4: 112412086-112412086 on Build GRCh38
rs78597342 Chr.4: 112411881-112411881 on Build GRCh38
rs532284879 Chr.4: 112412004-112412004 on Build GRCh38
rs942090186 Chr.4: 112411924-112411924 on Build GRCh38
rs770001847 Chr.4: 112411960-112411960 on Build GRCh38
rs780840562 Chr.4: 112411840-112411841 on Build GRCh38
rs201002217 Chr.4: 112411968-112411968 on Build GRCh38
rs775042131 Chr.4: 112411988-112411988 on Build GRCh38
rs146381479 Chr.4: 112412015-112412015 on Build GRCh38
rs757280722 Chr.4: 112411850-112411850 on Build GRCh38
rs754131633 Chr.4: 112411802-112411802 on Build GRCh38
rs758388317 Chr.4: 112411910-112411910 on Build GRCh38
rs747284395 Chr.4: 112411862-112411862 on Build GRCh38
rs541098605 Chr.4: 112412186-112412186 on Build GRCh38
rs750213241 Chr.4: 112412008-112412008 on Build GRCh38
rs529361422 Chr.4: 112411793-112411793 on Build GRCh38
rs373918792 Chr.4: 112411894-112411894 on Build GRCh38
rs751694209 Chr.4: 112412020-112412020 on Build GRCh38
rs765776944 Chr.4: 112411870-112411870 on Build GRCh38
rs112218994 Chr.4: 112411989-112411989 on Build GRCh38
rs759065750 Chr.4: 112411887-112411887 on Build GRCh38
rs755738158 Chr.4: 112411832-112411832 on Build GRCh38
rs768970546 Chr.4: 112411864-112411864 on Build GRCh38
rs748574839 Chr.4: 112411958-112411958 on Build GRCh38
rs753685488 Chr.4: 112411899-112411899 on Build GRCh38
rs1008077460 Chr.4: 112412082-112412082 on Build GRCh38
rs752167593 Chr.4: 112411891-112411891 on Build GRCh38
rs370555520 Chr.4: 112411794-112411794 on Build GRCh38
rs755619682 Chr.4: 112411884-112411886 on Build GRCh38
rs778943984 Chr.4: 112411905-112411905 on Build GRCh38
rs745752579 Chr.4: 112411909-112411909 on Build GRCh38
rs767060999 Chr.4: 112411890-112411890 on Build GRCh38
rs1042983017 Chr.4: 112411931-112411931 on Build GRCh38
rs765648789 Chr.4: 112411809-112411809 on Build GRCh38
rs749035725 Chr.4: 112411841-112411841 on Build GRCh38
rs1003679795 Chr.4: 112412221-112412221 on Build GRCh38
rs974843563 Chr.4: 112412161-112412161 on Build GRCh38
rs908055837 Chr.4: 112411908-112411908 on Build GRCh38
rs763814228 Chr.4: 112411995-112411995 on Build GRCh38
rs374550552 Chr.4: 112411999-112411999 on Build GRCh38
rs34847576 Chr.4: 112412156-112412157 on Build GRCh38
rs550191337 Chr.4: 112412011-112412011 on Build GRCh38
rs772239630 Chr.4: 112411855-112411855 on Build GRCh38
rs777325540 Chr.4: 112411933-112411933 on Build GRCh38
rs924445505 Chr.4: 112411932-112411932 on Build GRCh38
rs943398032 Chr.4: 112412156-112412156 on Build GRCh38
rs761154196 Chr.4: 112411799-112411799 on Build GRCh38
rs773551645 Chr.4: 112411874-112411874 on Build GRCh38
rs776648904 Chr.4: 112411948-112411948 on Build GRCh38
rs762448453 Chr.4: 112411807-112411807 on Build GRCh38
rs543394179 Chr.4: 112412122-112412122 on Build GRCh38
rs542474597 Chr.4: 112412005-112412005 on Build GRCh38
rs368944438 Chr.4: 112412072-112412072 on Build GRCh38
rs759525001 Chr.4: 112411788-112411788 on Build GRCh38
rs746351025 Chr.4: 112411997-112412010 on Build GRCh38
rs777661055 Chr.4: 112411833-112411833 on Build GRCh38
rs775870775 Chr.4: 112412024-112412025 on Build GRCh38
rs375367633 Chr.4: 112411828-112411828 on Build GRCh38
rs928436331 Chr.4: 112411804-112411804 on Build GRCh38
rs376695615 Chr.4: 112411865-112411865 on Build GRCh38
rs748365124 Chr.4: 112412046-112412046 on Build GRCh38

View Full Product Details