Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 112425578-112426077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALPK1
Entrez Gene ID
80216
Gene Name
alpha kinase 1
Gene Aliases
8430410J10Rik, LAK
UniGene
Hs.652825
Gene Chromosome Location
Chr.4: 112286420-112442618 on Build GRCh38
Transcripts
NM_001102406.1 NM_001253884.1 NM_025144.3 XM_005263245.4 XM_005263246.4 XM_006714326.3 XM_017008633.1 XM_017008634.1 XM_017008635.1 XM_017008636.1 XM_017008637.1 XM_017008638.1 XM_017008639.1 XM_017008640.1 XM_017008641.1 XM_017008642.1 XM_017008643.1 XM_017008644.1 XM_017008645.1 XM_017008646.1 XM_017008647.1 XM_017008648.1 XM_017008649.1 XM_017008650.1 XM_017008651.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3599174 c.610C>T p.L204L Chr.4: 112425739-112425739 on Build GRCh38
5419514 c.597C>T p.I199I Chr.4: 112425726-112425726 on Build GRCh38
3599173 c.540C>T p.T180T Chr.4: 112425669-112425669 on Build GRCh38
6598753 c.536-6delT p.? Chr.4: 112425659-112425659 on Build GRCh38
4121390 c.577C>T p.Q193* Chr.4: 112425706-112425706 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373046330 Chr.4: 112425686-112425686 on Build GRCh38
rs532730383 Chr.4: 112425946-112425946 on Build GRCh38
rs754737048 Chr.4: 112425762-112425762 on Build GRCh38
rs770718790 Chr.4: 112425668-112425668 on Build GRCh38
rs765486098 Chr.4: 112425625-112425625 on Build GRCh38
rs77202449 Chr.4: 112425879-112425879 on Build GRCh38
rs921876618 Chr.4: 112425932-112425932 on Build GRCh38
rs968804839 Chr.4: 112425969-112425969 on Build GRCh38
rs978748329 Chr.4: 112425978-112425978 on Build GRCh38
rs780592514 Chr.4: 112426010-112426010 on Build GRCh38
rs10017209 Chr.4: 112425840-112425840 on Build GRCh38
rs138151334 Chr.4: 112425690-112425690 on Build GRCh38
rs202246189 Chr.4: 112425749-112425749 on Build GRCh38
rs1012447959 Chr.4: 112425806-112425806 on Build GRCh38
rs766806450 Chr.4: 112425760-112425760 on Build GRCh38
rs1052359761 Chr.4: 112425694-112425694 on Build GRCh38
rs777488331 Chr.4: 112425665-112425665 on Build GRCh38
rs796918040 Chr.4: 112425925-112425925 on Build GRCh38
rs771854606 Chr.4: 112425708-112425708 on Build GRCh38
rs755509952 Chr.4: 112425651-112425651 on Build GRCh38
rs775130609 Chr.4: 112425710-112425710 on Build GRCh38
rs751963789 Chr.4: 112425761-112425761 on Build GRCh38
rs536655539 Chr.4: 112426003-112426003 on Build GRCh38
rs148738985 Chr.4: 112425711-112425711 on Build GRCh38
rs755596237 Chr.4: 112425773-112425773 on Build GRCh38
rs766249760 Chr.4: 112425692-112425692 on Build GRCh38
rs890661676 Chr.4: 112425754-112425754 on Build GRCh38
rs748790655 Chr.4: 112425667-112425667 on Build GRCh38
rs544012199 Chr.4: 112425703-112425703 on Build GRCh38
rs201171863 Chr.4: 112425722-112425722 on Build GRCh38
rs548087928 Chr.4: 112425968-112425968 on Build GRCh38
rs1001127066 Chr.4: 112425660-112425660 on Build GRCh38
rs762910530 Chr.4: 112425638-112425639 on Build GRCh38
rs773206030 Chr.4: 112425636-112425637 on Build GRCh38
rs536152336 Chr.4: 112425932-112425933 on Build GRCh38
rs959867212 Chr.4: 112425931-112425931 on Build GRCh38
rs973291836 Chr.4: 112425905-112425905 on Build GRCh38
rs577907136 Chr.4: 112425757-112425757 on Build GRCh38
rs867827635 Chr.4: 112426043-112426043 on Build GRCh38
rs776750570 Chr.4: 112425718-112425718 on Build GRCh38
rs565908960 Chr.4: 112425981-112425981 on Build GRCh38
rs554668253 Chr.4: 112425643-112425643 on Build GRCh38
rs757541443 Chr.4: 112425616-112425616 on Build GRCh38
rs559927546 Chr.4: 112425832-112425832 on Build GRCh38
rs765384476 Chr.4: 112425634-112425635 on Build GRCh38
rs780445460 Chr.4: 112425638-112425638 on Build GRCh38
rs751312365 Chr.4: 112425760-112425761 on Build GRCh38
rs753400003 Chr.4: 112425797-112425797 on Build GRCh38
rs911623892 Chr.4: 112425970-112425970 on Build GRCh38
rs764015889 Chr.4: 112425713-112425713 on Build GRCh38
rs191173233 Chr.4: 112425929-112425929 on Build GRCh38
rs1004172435 Chr.4: 112425880-112425880 on Build GRCh38
rs902726844 Chr.4: 112425653-112425653 on Build GRCh38
rs200304892 Chr.4: 112425719-112425719 on Build GRCh38
rs150206897 Chr.4: 112425930-112425930 on Build GRCh38
rs368808111 Chr.4: 112425633-112425633 on Build GRCh38
rs750607501 Chr.4: 112425731-112425731 on Build GRCh38
rs765383617 Chr.4: 112425730-112425730 on Build GRCh38
rs971237540 Chr.4: 112425849-112425849 on Build GRCh38
rs750767117 Chr.4: 112425631-112425631 on Build GRCh38
rs963380451 Chr.4: 112425900-112425900 on Build GRCh38
rs140834461 Chr.4: 112425919-112425919 on Build GRCh38
rs114326978 Chr.4: 112425859-112425859 on Build GRCh38
rs758326801 Chr.4: 112425604-112425604 on Build GRCh38
rs571965076 Chr.4: 112425934-112425934 on Build GRCh38
rs551709172 Chr.4: 112425979-112425979 on Build GRCh38
rs895296020 Chr.4: 112425765-112425765 on Build GRCh38
rs560437040 Chr.4: 112425782-112425782 on Build GRCh38

View Full Product Details