Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 112427492-112427995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALPK1
Entrez Gene ID
80216
Gene Name
alpha kinase 1
Gene Aliases
8430410J10Rik, LAK
UniGene
Hs.652825
Gene Chromosome Location
Chr.4: 112286420-112442618 on Build GRCh38
Transcripts
NM_001102406.1 NM_001253884.1 NM_025144.3 XM_005263245.4 XM_005263246.4 XM_006714326.3 XM_017008633.1 XM_017008634.1 XM_017008635.1 XM_017008636.1 XM_017008637.1 XM_017008638.1 XM_017008639.1 XM_017008640.1 XM_017008641.1 XM_017008642.1 XM_017008643.1 XM_017008644.1 XM_017008645.1 XM_017008646.1 XM_017008647.1 XM_017008648.1 XM_017008649.1 XM_017008650.1 XM_017008651.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4557203 c.718G>A p.G240S Chr.4: 112427588-112427588 on Build GRCh38
731960 c.747G>C p.M249I Chr.4: 112427617-112427617 on Build GRCh38
5974961 c.762T>C p.Y254Y Chr.4: 112427632-112427632 on Build GRCh38
5526534 c.705C>T p.L235L Chr.4: 112427575-112427575 on Build GRCh38
6488330 c.725T>G p.L242R Chr.4: 112427595-112427595 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017760769 Chr.4: 112427952-112427952 on Build GRCh38
rs752240064 Chr.4: 112427711-112427711 on Build GRCh38
rs763837838 Chr.4: 112427717-112427717 on Build GRCh38
rs747432717 Chr.4: 112427687-112427687 on Build GRCh38
rs879199633 Chr.4: 112427737-112427737 on Build GRCh38
rs906917166 Chr.4: 112427884-112427884 on Build GRCh38
rs1040224478 Chr.4: 112427806-112427806 on Build GRCh38
rs986267825 Chr.4: 112427715-112427715 on Build GRCh38
rs764667727 Chr.4: 112427573-112427573 on Build GRCh38
rs777786049 Chr.4: 112427626-112427626 on Build GRCh38
rs113977807 Chr.4: 112427697-112427697 on Build GRCh38
rs772422578 Chr.4: 112427556-112427556 on Build GRCh38
rs1040821997 Chr.4: 112427602-112427602 on Build GRCh38
rs371765193 Chr.4: 112427709-112427709 on Build GRCh38
rs73841017 Chr.4: 112427970-112427970 on Build GRCh38
rs761174009 Chr.4: 112427835-112427835 on Build GRCh38
rs757233009 Chr.4: 112427634-112427634 on Build GRCh38
rs150823391 Chr.4: 112427619-112427619 on Build GRCh38
rs537055391 Chr.4: 112427948-112427948 on Build GRCh38
rs762548610 Chr.4: 112427583-112427583 on Build GRCh38
rs773865358 Chr.4: 112427703-112427703 on Build GRCh38
rs772436859 Chr.4: 112427660-112427660 on Build GRCh38
rs775509415 Chr.4: 112427684-112427684 on Build GRCh38
rs896755370 Chr.4: 112427946-112427946 on Build GRCh38
rs368316265 Chr.4: 112427838-112427838 on Build GRCh38
rs779204222 Chr.4: 112427649-112427649 on Build GRCh38
rs372563699 Chr.4: 112427738-112427739 on Build GRCh38
rs370712989 Chr.4: 112427543-112427543 on Build GRCh38
rs138057180 Chr.4: 112427917-112427917 on Build GRCh38
rs752537126 Chr.4: 112427616-112427616 on Build GRCh38
rs527857871 Chr.4: 112427690-112427690 on Build GRCh38
rs992355440 Chr.4: 112427728-112427728 on Build GRCh38
rs976604330 Chr.4: 112427790-112427790 on Build GRCh38
rs979745267 Chr.4: 112427757-112427757 on Build GRCh38
rs749235785 Chr.4: 112427627-112427627 on Build GRCh38
rs952203327 Chr.4: 112427714-112427714 on Build GRCh38
rs944508350 Chr.4: 112427793-112427793 on Build GRCh38
rs191970455 Chr.4: 112427773-112427773 on Build GRCh38
rs932095150 Chr.4: 112427836-112427836 on Build GRCh38
rs201720771 Chr.4: 112427603-112427603 on Build GRCh38
rs1054397730 Chr.4: 112427555-112427555 on Build GRCh38
rs183069006 Chr.4: 112427896-112427896 on Build GRCh38
rs375431050 Chr.4: 112427933-112427933 on Build GRCh38
rs765067038 Chr.4: 112427802-112427802 on Build GRCh38
rs767094062 Chr.4: 112427610-112427610 on Build GRCh38
rs779504755 Chr.4: 112427638-112427638 on Build GRCh38
rs1004885655 Chr.4: 112427928-112427928 on Build GRCh38
rs748476577 Chr.4: 112427849-112427849 on Build GRCh38
rs896814284 Chr.4: 112427915-112427915 on Build GRCh38
rs913156184 Chr.4: 112427783-112427783 on Build GRCh38
rs4834277 Chr.4: 112427864-112427864 on Build GRCh38
rs541297996 Chr.4: 112427753-112427753 on Build GRCh38
rs776815288 Chr.4: 112427957-112427957 on Build GRCh38
rs767554894 Chr.4: 112427567-112427567 on Build GRCh38
rs779158312 Chr.4: 112427778-112427778 on Build GRCh38
rs762292079 Chr.4: 112427694-112427694 on Build GRCh38
rs74727833 Chr.4: 112427869-112427869 on Build GRCh38
rs753520922 Chr.4: 112427735-112427735 on Build GRCh38
rs781150173 Chr.4: 112427968-112427968 on Build GRCh38
rs768855740 Chr.4: 112427688-112427688 on Build GRCh38
rs893067023 Chr.4: 112427859-112427859 on Build GRCh38
rs1027826323 Chr.4: 112427931-112427931 on Build GRCh38
rs1015782093 Chr.4: 112427912-112427912 on Build GRCh38
rs935926577 Chr.4: 112427571-112427571 on Build GRCh38
rs538444321 Chr.4: 112427816-112427816 on Build GRCh38
rs376492179 Chr.4: 112427850-112427850 on Build GRCh38
rs1052954321 Chr.4: 112427580-112427580 on Build GRCh38
rs372841187 Chr.4: 112427584-112427584 on Build GRCh38
rs760351476 Chr.4: 112427712-112427712 on Build GRCh38
rs554928370 Chr.4: 112427927-112427927 on Build GRCh38
rs373860176 Chr.4: 112427597-112427597 on Build GRCh38
rs201401104 Chr.4: 112427581-112427581 on Build GRCh38
rs377436691 Chr.4: 112427528-112427528 on Build GRCh38
rs199719196 Chr.4: 112427538-112427538 on Build GRCh38
rs78341843 Chr.4: 112427934-112427934 on Build GRCh38
rs1002612288 Chr.4: 112427896-112427897 on Build GRCh38
rs879074644 Chr.4: 112427813-112427813 on Build GRCh38
rs1005588517 Chr.4: 112427655-112427655 on Build GRCh38
rs745934168 Chr.4: 112427652-112427652 on Build GRCh38
rs776059783 Chr.4: 112427557-112427557 on Build GRCh38
rs377361072 Chr.4: 112427539-112427539 on Build GRCh38
rs558361316 Chr.4: 112427966-112427966 on Build GRCh38
rs925647970 Chr.4: 112427764-112427764 on Build GRCh38
rs149307398 Chr.4: 112427737-112427738 on Build GRCh38
rs1022687023 Chr.4: 112427593-112427593 on Build GRCh38
rs934688919 Chr.4: 112427845-112427845 on Build GRCh38
rs888012804 Chr.4: 112427646-112427646 on Build GRCh38

View Full Product Details