Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 112553468-112553988 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZGRF1
Entrez Gene ID
55345
Gene Name
zinc finger GRF-type containing 1
Gene Aliases
C4orf21
UniGene
Hs.380346
Gene Chromosome Location
Chr.4: 112539333-112636995 on Build GRCh38
Transcripts
NM_018392.4 XM_005263115.3 XM_011532091.2 XM_011532092.2 XM_011532093.2 XM_011532094.2 XM_011532097.2 XM_011532098.2 XM_011532099.2 XM_017008369.1 XM_017008370.1 XM_017008371.1 XM_017008372.1 XM_017008373.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3599192 c.5305G>A p.E1769K Chr.4: 112553876-112553876 on Build GRCh38
5746330 c.5297A>G p.K1766R Chr.4: 112553884-112553884 on Build GRCh38
5436480 c.5260G>C p.E1754Q Chr.4: 112553921-112553921 on Build GRCh38
4845840 c.5340G>A p.L1780L Chr.4: 112553841-112553841 on Build GRCh38
5496330 c.5302A>T p.I1768F Chr.4: 112553879-112553879 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773373573 Chr.4: 112553841-112553841 on Build GRCh38
rs759746664 Chr.4: 112553952-112553952 on Build GRCh38
rs755355156 Chr.4: 112553858-112553858 on Build GRCh38
rs892476324 Chr.4: 112553758-112553758 on Build GRCh38
rs1032583442 Chr.4: 112553601-112553601 on Build GRCh38
rs766548463 Chr.4: 112553854-112553854 on Build GRCh38
rs1001891499 Chr.4: 112553546-112553546 on Build GRCh38
rs769812310 Chr.4: 112553932-112553932 on Build GRCh38
rs761771355 Chr.4: 112553815-112553815 on Build GRCh38
rs754158837 Chr.4: 112553694-112553694 on Build GRCh38
rs921389526 Chr.4: 112553707-112553707 on Build GRCh38
rs763173044 Chr.4: 112553937-112553937 on Build GRCh38
rs931635332 Chr.4: 112553675-112553675 on Build GRCh38
rs775179487 Chr.4: 112553793-112553793 on Build GRCh38
rs185116025 Chr.4: 112553522-112553522 on Build GRCh38
rs1026401654 Chr.4: 112553582-112553582 on Build GRCh38
rs969646479 Chr.4: 112553562-112553562 on Build GRCh38
rs538397484 Chr.4: 112553693-112553693 on Build GRCh38
rs868381340 Chr.4: 112553846-112553846 on Build GRCh38
rs1023705810 Chr.4: 112553780-112553780 on Build GRCh38
rs905223543 Chr.4: 112553792-112553792 on Build GRCh38
rs1032634758 Chr.4: 112553840-112553840 on Build GRCh38
rs760795519 Chr.4: 112553512-112553512 on Build GRCh38
rs556807884 Chr.4: 112553708-112553708 on Build GRCh38
rs779635105 Chr.4: 112553905-112553905 on Build GRCh38
rs374230171 Chr.4: 112553947-112553947 on Build GRCh38
rs746832079 Chr.4: 112553802-112553802 on Build GRCh38
rs115828964 Chr.4: 112553606-112553606 on Build GRCh38
rs776503379 Chr.4: 112553812-112553812 on Build GRCh38
rs141688719 Chr.4: 112553892-112553892 on Build GRCh38
rs960353005 Chr.4: 112553551-112553551 on Build GRCh38
rs961035906 Chr.4: 112553515-112553515 on Build GRCh38
rs867732622 Chr.4: 112553906-112553906 on Build GRCh38
rs767919193 Chr.4: 112553859-112553859 on Build GRCh38
rs921580338 Chr.4: 112553661-112553661 on Build GRCh38
rs774503447 Chr.4: 112553948-112553948 on Build GRCh38
rs113227039 Chr.4: 112553725-112553725 on Build GRCh38
rs190389649 Chr.4: 112553630-112553630 on Build GRCh38
rs567857905 Chr.4: 112553660-112553660 on Build GRCh38
rs571878361 Chr.4: 112553738-112553738 on Build GRCh38
rs940534919 Chr.4: 112553720-112553720 on Build GRCh38
rs1051451172 Chr.4: 112553495-112553495 on Build GRCh38
rs1033417861 Chr.4: 112553548-112553548 on Build GRCh38
rs551321669 Chr.4: 112553526-112553526 on Build GRCh38
rs181177506 Chr.4: 112553935-112553935 on Build GRCh38
rs148022472 Chr.4: 112553836-112553836 on Build GRCh38
rs979705913 Chr.4: 112553570-112553570 on Build GRCh38
rs773250597 Chr.4: 112553936-112553936 on Build GRCh38
rs566372613 Chr.4: 112553604-112553604 on Build GRCh38
rs868751185 Chr.4: 112553894-112553894 on Build GRCh38
rs1013652102 Chr.4: 112553778-112553778 on Build GRCh38
rs745424422 Chr.4: 112553893-112553893 on Build GRCh38
rs562754900 Chr.4: 112553506-112553506 on Build GRCh38
rs756564836 Chr.4: 112553872-112553872 on Build GRCh38
rs941531844 Chr.4: 112553685-112553685 on Build GRCh38
rs371251700 Chr.4: 112553794-112553794 on Build GRCh38
rs941772408 Chr.4: 112553558-112553558 on Build GRCh38
rs1009906217 Chr.4: 112553520-112553520 on Build GRCh38
rs748116145 Chr.4: 112553926-112553926 on Build GRCh38
rs766727415 Chr.4: 112553573-112553573 on Build GRCh38
rs763296660 Chr.4: 112553853-112553853 on Build GRCh38
rs780979249 Chr.4: 112553924-112553924 on Build GRCh38
rs753216220 Chr.4: 112553866-112553866 on Build GRCh38
rs965841797 Chr.4: 112553908-112553908 on Build GRCh38
rs896581937 Chr.4: 112553742-112553742 on Build GRCh38
rs538988073 Chr.4: 112553811-112553811 on Build GRCh38
rs767796398 Chr.4: 112553956-112553956 on Build GRCh38
rs747134039 Chr.4: 112553925-112553926 on Build GRCh38
rs956929092 Chr.4: 112553605-112553605 on Build GRCh38
rs751781449 Chr.4: 112553856-112553856 on Build GRCh38
rs991878498 Chr.4: 112553576-112553576 on Build GRCh38
rs944421729 Chr.4: 112553631-112553631 on Build GRCh38
rs150527612 Chr.4: 112553904-112553904 on Build GRCh38
rs934068595 Chr.4: 112553710-112553710 on Build GRCh38

View Full Product Details