Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 124669011-124669551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD50
Entrez Gene ID
57182
Gene Name
ankyrin repeat domain 50
Gene Aliases
-
UniGene
Hs.480694
Gene Chromosome Location
Chr.4: 124664048-124712732 on Build GRCh38
Transcripts
NM_001167882.1 NM_020337.2 XM_017008471.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7317068 c.4027C>T p.Q1343* Chr.4: 124669250-124669250 on Build GRCh38
3917094 c.4197C>T p.S1399S Chr.4: 124669080-124669080 on Build GRCh38
3599811 c.4243G>A p.E1415K Chr.4: 124669034-124669034 on Build GRCh38
6600819 c.3813delG p.K1272fs*26 Chr.4: 124669464-124669464 on Build GRCh38
3599813 c.3809G>A p.G1270E Chr.4: 124669468-124669468 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1874748 Chr.4: 124669323-124669323 on Build GRCh38
rs186680694 Chr.4: 124669507-124669507 on Build GRCh38
rs938391003 Chr.4: 124669159-124669159 on Build GRCh38
rs777168754 Chr.4: 124669091-124669091 on Build GRCh38
rs141676247 Chr.4: 124669348-124669348 on Build GRCh38
rs528056780 Chr.4: 124669305-124669305 on Build GRCh38
rs201541253 Chr.4: 124669111-124669111 on Build GRCh38
rs772106197 Chr.4: 124669205-124669207 on Build GRCh38
rs200657431 Chr.4: 124669286-124669286 on Build GRCh38
rs569034621 Chr.4: 124669363-124669363 on Build GRCh38
rs764755874 Chr.4: 124669373-124669373 on Build GRCh38
rs970688640 Chr.4: 124669134-124669134 on Build GRCh38
rs776964769 Chr.4: 124669151-124669151 on Build GRCh38
rs573566243 Chr.4: 124669143-124669143 on Build GRCh38
rs745787834 Chr.4: 124669129-124669129 on Build GRCh38
rs149234063 Chr.4: 124669193-124669193 on Build GRCh38
rs766708715 Chr.4: 124669082-124669082 on Build GRCh38
rs763774791 Chr.4: 124669201-124669201 on Build GRCh38
rs757818256 Chr.4: 124669394-124669394 on Build GRCh38
rs113942607 Chr.4: 124669519-124669519 on Build GRCh38
rs749896966 Chr.4: 124669383-124669383 on Build GRCh38
rs751523274 Chr.4: 124669496-124669496 on Build GRCh38
rs775628252 Chr.4: 124669045-124669045 on Build GRCh38
rs774979224 Chr.4: 124669428-124669428 on Build GRCh38
rs768974044 Chr.4: 124669142-124669142 on Build GRCh38
rs762055333 Chr.4: 124669156-124669156 on Build GRCh38
rs775091823 Chr.4: 124669328-124669328 on Build GRCh38
rs748412709 Chr.4: 124669412-124669412 on Build GRCh38
rs868546287 Chr.4: 124669429-124669429 on Build GRCh38
rs548469161 Chr.4: 124669447-124669447 on Build GRCh38
rs765888235 Chr.4: 124669074-124669074 on Build GRCh38
rs138487103 Chr.4: 124669080-124669080 on Build GRCh38
rs181647409 Chr.4: 124669334-124669334 on Build GRCh38
rs199669857 Chr.4: 124669059-124669059 on Build GRCh38
rs762306694 Chr.4: 124669336-124669336 on Build GRCh38
rs199500950 Chr.4: 124669227-124669227 on Build GRCh38
rs368511135 Chr.4: 124669173-124669173 on Build GRCh38
rs754837414 Chr.4: 124669409-124669409 on Build GRCh38
rs772425796 Chr.4: 124669438-124669438 on Build GRCh38
rs779732106 Chr.4: 124669296-124669296 on Build GRCh38
rs749457631 Chr.4: 124669524-124669524 on Build GRCh38
rs756371482 Chr.4: 124669245-124669245 on Build GRCh38
rs751442096 Chr.4: 124669402-124669402 on Build GRCh38
rs530037702 Chr.4: 124669433-124669433 on Build GRCh38
rs941271122 Chr.4: 124669427-124669427 on Build GRCh38
rs760525425 Chr.4: 124669421-124669422 on Build GRCh38
rs534048314 Chr.4: 124669077-124669077 on Build GRCh38
rs150964092 Chr.4: 124669460-124669460 on Build GRCh38
rs560573825 Chr.4: 124669283-124669283 on Build GRCh38
rs112459806 Chr.4: 124669518-124669518 on Build GRCh38
rs542073632 Chr.4: 124669246-124669246 on Build GRCh38
rs762585211 Chr.4: 124669479-124669479 on Build GRCh38
rs765752803 Chr.4: 124669486-124669486 on Build GRCh38
rs199504785 Chr.4: 124669212-124669212 on Build GRCh38
rs764083029 Chr.4: 124669067-124669067 on Build GRCh38
rs945749148 Chr.4: 124669234-124669234 on Build GRCh38
rs778120664 Chr.4: 124669420-124669420 on Build GRCh38
rs111651064 Chr.4: 124669337-124669337 on Build GRCh38
rs747222819 Chr.4: 124669138-124669138 on Build GRCh38
rs776809046 Chr.4: 124669072-124669072 on Build GRCh38
rs766687553 Chr.4: 124669176-124669176 on Build GRCh38
rs746864039 Chr.4: 124669312-124669312 on Build GRCh38
rs929619391 Chr.4: 124669357-124669357 on Build GRCh38
rs780108468 Chr.4: 124669395-124669395 on Build GRCh38
rs147174885 Chr.4: 124669423-124669423 on Build GRCh38
rs994632367 Chr.4: 124669422-124669422 on Build GRCh38
rs763894330 Chr.4: 124669070-124669070 on Build GRCh38
rs745794223 Chr.4: 124669278-124669278 on Build GRCh38
rs760012356 Chr.4: 124669340-124669340 on Build GRCh38
rs750041405 Chr.4: 124669469-124669469 on Build GRCh38
rs765620183 Chr.4: 124669411-124669411 on Build GRCh38
rs755940649 Chr.4: 124669514-124669514 on Build GRCh38
rs373345736 Chr.4: 124669521-124669521 on Build GRCh38
rs764916001 Chr.4: 124669464-124669464 on Build GRCh38
rs752299201 Chr.4: 124669089-124669089 on Build GRCh38
rs761965382 Chr.4: 124669073-124669073 on Build GRCh38
rs781158004 Chr.4: 124669506-124669506 on Build GRCh38
rs200614722 Chr.4: 124669419-124669419 on Build GRCh38
rs878936119 Chr.4: 124669036-124669036 on Build GRCh38
rs773870129 Chr.4: 124669164-124669164 on Build GRCh38
rs748326840 Chr.4: 124669324-124669324 on Build GRCh38
rs771895625 Chr.4: 124669039-124669039 on Build GRCh38
rs200711493 Chr.4: 124669122-124669122 on Build GRCh38
rs765398629 Chr.4: 124669157-124669157 on Build GRCh38
rs753498656 Chr.4: 124669352-124669352 on Build GRCh38
rs751904052 Chr.4: 124669190-124669190 on Build GRCh38
rs972600223 Chr.4: 124669087-124669087 on Build GRCh38
rs771772634 Chr.4: 124669130-124669130 on Build GRCh38
rs961444935 Chr.4: 124669085-124669085 on Build GRCh38
rs745801835 Chr.4: 124669053-124669056 on Build GRCh38
rs377291470 Chr.4: 124669110-124669110 on Build GRCh38
rs989696794 Chr.4: 124669235-124669235 on Build GRCh38
rs755753253 Chr.4: 124669090-124669090 on Build GRCh38
rs748843184 Chr.4: 124669093-124669093 on Build GRCh38
rs912993126 Chr.4: 124669214-124669214 on Build GRCh38
rs202030328 Chr.4: 124669338-124669338 on Build GRCh38
rs574799458 Chr.4: 124669228-124669228 on Build GRCh38

View Full Product Details