Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 124669841-124670336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD50
Entrez Gene ID
57182
Gene Name
ankyrin repeat domain 50
Gene Aliases
-
UniGene
Hs.480694
Gene Chromosome Location
Chr.4: 124664048-124712732 on Build GRCh38
Transcripts
NM_001167882.1 NM_020337.2 XM_017008471.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
480816 c.3434A>C p.D1145A Chr.4: 124669843-124669843 on Build GRCh38
4337604 c.3163delG p.E1055fs*11 Chr.4: 124670114-124670114 on Build GRCh38
231758 c.3359C>T p.P1120L Chr.4: 124669918-124669918 on Build GRCh38
5594181 c.3184C>T p.Q1062* Chr.4: 124670093-124670093 on Build GRCh38
4828442 c.3093G>C p.L1031L Chr.4: 124670184-124670184 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757664338 Chr.4: 124670145-124670145 on Build GRCh38
rs768205304 Chr.4: 124670030-124670030 on Build GRCh38
rs139499397 Chr.4: 124670299-124670299 on Build GRCh38
rs764738403 Chr.4: 124670227-124670227 on Build GRCh38
rs967469880 Chr.4: 124670013-124670013 on Build GRCh38
rs201273871 Chr.4: 124670041-124670041 on Build GRCh38
rs774453017 Chr.4: 124670176-124670176 on Build GRCh38
rs779826893 Chr.4: 124669995-124669995 on Build GRCh38
rs779918695 Chr.4: 124670151-124670151 on Build GRCh38
rs775586706 Chr.4: 124670208-124670208 on Build GRCh38
rs781617465 Chr.4: 124669938-124669938 on Build GRCh38
rs770702877 Chr.4: 124670174-124670174 on Build GRCh38
rs753145762 Chr.4: 124670119-124670119 on Build GRCh38
rs1046524337 Chr.4: 124670032-124670032 on Build GRCh38
rs978446637 Chr.4: 124670022-124670022 on Build GRCh38
rs1014095802 Chr.4: 124669943-124669943 on Build GRCh38
rs775544087 Chr.4: 124670308-124670308 on Build GRCh38
rs944175738 Chr.4: 124670182-124670182 on Build GRCh38
rs767573228 Chr.4: 124670109-124670109 on Build GRCh38
rs761745894 Chr.4: 124670034-124670034 on Build GRCh38
rs141095097 Chr.4: 124670265-124670265 on Build GRCh38
rs925727361 Chr.4: 124670029-124670029 on Build GRCh38
rs371573842 Chr.4: 124670273-124670273 on Build GRCh38
rs148154149 Chr.4: 124670260-124670260 on Build GRCh38
rs759594537 Chr.4: 124670105-124670105 on Build GRCh38
rs757069076 Chr.4: 124670202-124670202 on Build GRCh38
rs143672935 Chr.4: 124669916-124669916 on Build GRCh38
rs771797868 Chr.4: 124669993-124669993 on Build GRCh38
rs765633282 Chr.4: 124670239-124670239 on Build GRCh38
rs145805142 Chr.4: 124670240-124670240 on Build GRCh38
rs145189367 Chr.4: 124669893-124669893 on Build GRCh38
rs3822116 Chr.4: 124670268-124670268 on Build GRCh38
rs754611161 Chr.4: 124670244-124670244 on Build GRCh38
rs772382179 Chr.4: 124670305-124670305 on Build GRCh38
rs768520765 Chr.4: 124669885-124669885 on Build GRCh38
rs146823198 Chr.4: 124670171-124670171 on Build GRCh38
rs796080274 Chr.4: 124670039-124670039 on Build GRCh38
rs750651470 Chr.4: 124669912-124669912 on Build GRCh38
rs754391701 Chr.4: 124670229-124670229 on Build GRCh38
rs111803915 Chr.4: 124669997-124669997 on Build GRCh38
rs778214956 Chr.4: 124669977-124669977 on Build GRCh38
rs767522480 Chr.4: 124670199-124670199 on Build GRCh38
rs756447459 Chr.4: 124669973-124669973 on Build GRCh38
rs1034108309 Chr.4: 124669908-124669908 on Build GRCh38
rs760903897 Chr.4: 124670310-124670310 on Build GRCh38
rs765026508 Chr.4: 124669899-124669899 on Build GRCh38
rs773106734 Chr.4: 124670170-124670170 on Build GRCh38
rs911364713 Chr.4: 124670146-124670146 on Build GRCh38
rs149147559 Chr.4: 124669914-124669914 on Build GRCh38
rs200654829 Chr.4: 124670033-124670033 on Build GRCh38
rs1020144166 Chr.4: 124669978-124669978 on Build GRCh38
rs753981794 Chr.4: 124669947-124669947 on Build GRCh38
rs1000014384 Chr.4: 124670084-124670084 on Build GRCh38
rs200351787 Chr.4: 124670130-124670130 on Build GRCh38
rs745819135 Chr.4: 124670262-124670262 on Build GRCh38
rs748058628 Chr.4: 124670162-124670162 on Build GRCh38
rs753236192 Chr.4: 124669971-124669971 on Build GRCh38
rs746657581 Chr.4: 124670154-124670154 on Build GRCh38
rs746096555 Chr.4: 124670303-124670303 on Build GRCh38
rs755055726 Chr.4: 124669937-124669937 on Build GRCh38
rs746537866 Chr.4: 124670024-124670024 on Build GRCh38
rs991406786 Chr.4: 124670309-124670309 on Build GRCh38
rs529948055 Chr.4: 124670088-124670088 on Build GRCh38
rs754303830 Chr.4: 124670144-124670144 on Build GRCh38
rs769772793 Chr.4: 124670168-124670168 on Build GRCh38
rs888533466 Chr.4: 124669866-124669866 on Build GRCh38
rs766629653 Chr.4: 124670085-124670085 on Build GRCh38
rs374920985 Chr.4: 124670043-124670043 on Build GRCh38
rs751688533 Chr.4: 124669920-124669920 on Build GRCh38
rs547418955 Chr.4: 124670011-124670011 on Build GRCh38
rs140645873 Chr.4: 124670190-124670190 on Build GRCh38
rs756600446 Chr.4: 124670123-124670123 on Build GRCh38
rs762715346 Chr.4: 124670077-124670077 on Build GRCh38
rs780715353 Chr.4: 124670157-124670157 on Build GRCh38
rs761578389 Chr.4: 124669898-124669898 on Build GRCh38
rs745546033 Chr.4: 124669979-124669979 on Build GRCh38
rs985628939 Chr.4: 124670052-124670052 on Build GRCh38
rs906651612 Chr.4: 124670047-124670047 on Build GRCh38
rs772947133 Chr.4: 124670059-124670059 on Build GRCh38
rs757752297 Chr.4: 124670233-124670233 on Build GRCh38

View Full Product Details