Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 152909950-152910484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFIP1
Entrez Gene ID
27236
Gene Name
ADP ribosylation factor interacting protein 1
Gene Aliases
HSU52521
UniGene
Hs.416089
Gene Chromosome Location
Chr.4: 152779937-152911912 on Build GRCh38
Transcripts
NM_001025593.2 NM_001025595.2 NM_001287431.1 NM_001287432.1 NM_001287433.1 NM_014447.3 XM_011531868.2 XM_011531869.2 XM_011531870.2 XM_011531871.2 XM_011531872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3600960 c.982A>G p.N328D Chr.4: 152910079-152910079 on Build GRCh38
460670 c.1004A>G p.N335S Chr.4: 152910101-152910101 on Build GRCh38
3660935 c.975A>G p.V325V Chr.4: 152910072-152910072 on Build GRCh38
4842909 c.985C>T p.Q329* Chr.4: 152910082-152910082 on Build GRCh38
1427943 c.1024G>A p.A342T Chr.4: 152910121-152910121 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766785672 Chr.4: 152910132-152910132 on Build GRCh38
rs938254118 Chr.4: 152910018-152910018 on Build GRCh38
rs749929039 Chr.4: 152910117-152910117 on Build GRCh38
rs772979357 Chr.4: 152910239-152910239 on Build GRCh38
rs200681290 Chr.4: 152910111-152910111 on Build GRCh38
rs1004289374 Chr.4: 152910003-152910003 on Build GRCh38
rs771907353 Chr.4: 152910090-152910090 on Build GRCh38
rs137970826 Chr.4: 152910108-152910108 on Build GRCh38
rs199675470 Chr.4: 152910182-152910182 on Build GRCh38
rs995407893 Chr.4: 152910080-152910080 on Build GRCh38
rs966404225 Chr.4: 152910429-152910429 on Build GRCh38
rs745780327 Chr.4: 152910083-152910083 on Build GRCh38
rs539482347 Chr.4: 152910229-152910229 on Build GRCh38
rs1034290954 Chr.4: 152910416-152910416 on Build GRCh38
rs150725835 Chr.4: 152910151-152910151 on Build GRCh38
rs113208221 Chr.4: 152910331-152910331 on Build GRCh38
rs989680570 Chr.4: 152910287-152910287 on Build GRCh38
rs1055769162 Chr.4: 152910043-152910043 on Build GRCh38
rs965051360 Chr.4: 152910207-152910207 on Build GRCh38
rs141134231 Chr.4: 152910059-152910059 on Build GRCh38
rs371767249 Chr.4: 152910253-152910253 on Build GRCh38
rs775296196 Chr.4: 152910098-152910098 on Build GRCh38
rs769842588 Chr.4: 152910078-152910078 on Build GRCh38
rs117074277 Chr.4: 152910250-152910250 on Build GRCh38
rs202078437 Chr.4: 152910206-152910206 on Build GRCh38
rs868221775 Chr.4: 152910292-152910292 on Build GRCh38
rs754778801 Chr.4: 152910112-152910112 on Build GRCh38
rs374308876 Chr.4: 152910201-152910201 on Build GRCh38
rs781586446 Chr.4: 152910175-152910175 on Build GRCh38
rs995528187 Chr.4: 152910394-152910394 on Build GRCh38
rs763463468 Chr.4: 152910128-152910128 on Build GRCh38
rs369437609 Chr.4: 152910073-152910073 on Build GRCh38
rs927646093 Chr.4: 152910012-152910012 on Build GRCh38
rs755307863 Chr.4: 152910144-152910144 on Build GRCh38
rs776586528 Chr.4: 152910210-152910210 on Build GRCh38
rs927744063 Chr.4: 152910436-152910436 on Build GRCh38
rs756362263 Chr.4: 152910181-152910181 on Build GRCh38
rs759355573 Chr.4: 152910148-152910149 on Build GRCh38
rs1014292443 Chr.4: 152910452-152910452 on Build GRCh38
rs758848450 Chr.4: 152910022-152910022 on Build GRCh38
rs746982072 Chr.4: 152910203-152910203 on Build GRCh38
rs891161426 Chr.4: 152910302-152910302 on Build GRCh38
rs749340196 Chr.4: 152910196-152910196 on Build GRCh38
rs781392108 Chr.4: 152910061-152910061 on Build GRCh38
rs375193580 Chr.4: 152910030-152910030 on Build GRCh38
rs760301712 Chr.4: 152910246-152910258 on Build GRCh38
rs1036961014 Chr.4: 152910336-152910339 on Build GRCh38
rs752868481 Chr.4: 152910177-152910177 on Build GRCh38
rs992407772 Chr.4: 152910097-152910097 on Build GRCh38
rs765957605 Chr.4: 152910249-152910249 on Build GRCh38
rs752179422 Chr.4: 152910139-152910139 on Build GRCh38
rs576478643 Chr.4: 152910291-152910291 on Build GRCh38
rs750915597 Chr.4: 152910021-152910021 on Build GRCh38
rs959910783 Chr.4: 152910086-152910086 on Build GRCh38
rs755760589 Chr.4: 152910451-152910451 on Build GRCh38
rs898600917 Chr.4: 152910342-152910342 on Build GRCh38
rs538400840 Chr.4: 152909993-152909993 on Build GRCh38
rs146931969 Chr.4: 152910101-152910101 on Build GRCh38
rs767338910 Chr.4: 152910153-152910154 on Build GRCh38
rs1046012836 Chr.4: 152910005-152910005 on Build GRCh38
rs780423224 Chr.4: 152910024-152910024 on Build GRCh38
rs775099378 Chr.4: 152910223-152910224 on Build GRCh38
rs777740272 Chr.4: 152910082-152910082 on Build GRCh38
rs370298612 Chr.4: 152910248-152910248 on Build GRCh38
rs761515589 Chr.4: 152910228-152910228 on Build GRCh38
rs760532325 Chr.4: 152910106-152910106 on Build GRCh38
rs752572761 Chr.4: 152910430-152910430 on Build GRCh38

View Full Product Details