Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 152910755-152911251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFIP1
Entrez Gene ID
27236
Gene Name
ADP ribosylation factor interacting protein 1
Gene Aliases
HSU52521
UniGene
Hs.416089
Gene Chromosome Location
Chr.4: 152779937-152911912 on Build GRCh38
Transcripts
NM_001025593.2 NM_001025595.2 NM_001287431.1 NM_001287432.1 NM_001287433.1 NM_014447.3 XM_011531868.2 XM_011531869.2 XM_011531870.2 XM_011531871.2 XM_011531872.1

SNP Detail

SNP ID Chromosome Location
rs949474678 Chr.4: 152910976-152910976 on Build GRCh38
rs555485755 Chr.4: 152911185-152911185 on Build GRCh38
rs142501451 Chr.4: 152911075-152911075 on Build GRCh38
rs983682806 Chr.4: 152910914-152910914 on Build GRCh38
rs537138906 Chr.4: 152911099-152911099 on Build GRCh38
rs775099943 Chr.4: 152911176-152911176 on Build GRCh38
rs184138885 Chr.4: 152910796-152910796 on Build GRCh38
rs1034864583 Chr.4: 152911103-152911103 on Build GRCh38
rs893154360 Chr.4: 152910978-152910978 on Build GRCh38
rs562063583 Chr.4: 152911144-152911144 on Build GRCh38
rs777951477 Chr.4: 152910963-152910963 on Build GRCh38
rs181285870 Chr.4: 152910916-152910916 on Build GRCh38
rs549709440 Chr.4: 152911092-152911092 on Build GRCh38
rs1027839019 Chr.4: 152911153-152911153 on Build GRCh38
rs974938991 Chr.4: 152910843-152910843 on Build GRCh38
rs557867008 Chr.4: 152910973-152910973 on Build GRCh38
rs959192962 Chr.4: 152910895-152910895 on Build GRCh38
rs531819541 Chr.4: 152910898-152910898 on Build GRCh38
rs768848734 Chr.4: 152910972-152910972 on Build GRCh38
rs529560157 Chr.4: 152910908-152910908 on Build GRCh38
rs371484060 Chr.4: 152910966-152910966 on Build GRCh38
rs374721241 Chr.4: 152911069-152911069 on Build GRCh38
rs1054836651 Chr.4: 152910967-152910967 on Build GRCh38
rs908152061 Chr.4: 152910928-152910928 on Build GRCh38
rs376259100 Chr.4: 152910906-152910906 on Build GRCh38
rs115166525 Chr.4: 152910957-152910957 on Build GRCh38
rs973283844 Chr.4: 152910951-152910951 on Build GRCh38
rs1010217620 Chr.4: 152911040-152911040 on Build GRCh38
rs983148973 Chr.4: 152911207-152911207 on Build GRCh38
rs376549732 Chr.4: 152910917-152910917 on Build GRCh38
rs776784040 Chr.4: 152911104-152911104 on Build GRCh38
rs995865244 Chr.4: 152911205-152911205 on Build GRCh38
rs894023928 Chr.4: 152911115-152911115 on Build GRCh38
rs188515130 Chr.4: 152910896-152910896 on Build GRCh38
rs76772112 Chr.4: 152910813-152910813 on Build GRCh38
rs1028745880 Chr.4: 152910866-152910866 on Build GRCh38
rs1020816664 Chr.4: 152911057-152911057 on Build GRCh38
rs944847849 Chr.4: 152910948-152910948 on Build GRCh38
rs11929781 Chr.4: 152911213-152911213 on Build GRCh38
rs7689343 Chr.4: 152910888-152910888 on Build GRCh38
rs543585620 Chr.4: 152911117-152911117 on Build GRCh38
rs966498627 Chr.4: 152910811-152910811 on Build GRCh38
rs747432595 Chr.4: 152911222-152911222 on Build GRCh38
rs878928082 Chr.4: 152910950-152910951 on Build GRCh38
rs1046583831 Chr.4: 152911019-152911026 on Build GRCh38

View Full Product Details