Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 158893508-158894012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf45
Entrez Gene ID
152940
Gene Name
chromosome 4 open reading frame 45
Gene Aliases
-
UniGene
Hs.415576
Gene Chromosome Location
Chr.4: 158893532-159082883 on Build GRCh38
Transcripts
NM_152543.2 XM_011531671.2 XM_011531672.2 XM_017007808.1 XM_017007809.1 XM_017007810.1 NM_001323916.1 NM_020840.2 XM_005263156.1 XM_005263158.1 XM_011532154.2 XM_017008487.1

Gene Symbol
FNIP2
Entrez Gene ID
57600
Gene Name
folliculin interacting protein 2
Gene Aliases
FNIPL, MAPO1
UniGene
Hs.652441
Gene Chromosome Location
Chr.4: 158768995-158908050 on Build GRCh38
Transcripts
NM_152543.2 XM_011531671.2 XM_011531672.2 XM_017007808.1 XM_017007809.1 XM_017007810.1 NM_001323916.1 NM_020840.2 XM_005263156.1 XM_005263158.1 XM_011532154.2 XM_017008487.1

SNP Detail

SNP ID Chromosome Location
rs954091901 Chr.4: 158893986-158893986 on Build GRCh38
rs915678857 Chr.4: 158893902-158893902 on Build GRCh38
rs1012533013 Chr.4: 158893852-158893852 on Build GRCh38
rs1004124631 Chr.4: 158893974-158893974 on Build GRCh38
rs1006186805 Chr.4: 158893667-158893667 on Build GRCh38
rs879674031 Chr.4: 158893875-158893875 on Build GRCh38
rs749959104 Chr.4: 158893662-158893662 on Build GRCh38
rs755504427 Chr.4: 158893722-158893722 on Build GRCh38
rs547149609 Chr.4: 158893614-158893614 on Build GRCh38
rs527358730 Chr.4: 158893572-158893572 on Build GRCh38
rs372847692 Chr.4: 158893652-158893652 on Build GRCh38
rs767801581 Chr.4: 158893926-158893927 on Build GRCh38
rs762284196 Chr.4: 158893895-158893895 on Build GRCh38
rs749486687 Chr.4: 158893760-158893760 on Build GRCh38
rs773524982 Chr.4: 158893747-158893747 on Build GRCh38
rs1048262276 Chr.4: 158893971-158893971 on Build GRCh38
rs902654088 Chr.4: 158893804-158893804 on Build GRCh38
rs986280901 Chr.4: 158893803-158893803 on Build GRCh38
rs764929726 Chr.4: 158893793-158893793 on Build GRCh38
rs1001051936 Chr.4: 158893808-158893808 on Build GRCh38
rs189547099 Chr.4: 158893729-158893729 on Build GRCh38
rs771999762 Chr.4: 158893764-158893764 on Build GRCh38
rs549411773 Chr.4: 158893759-158893759 on Build GRCh38
rs779508915 Chr.4: 158893684-158893684 on Build GRCh38
rs908346221 Chr.4: 158893551-158893551 on Build GRCh38
rs1048315347 Chr.4: 158893649-158893649 on Build GRCh38
rs764815657 Chr.4: 158893653-158893653 on Build GRCh38
rs757866128 Chr.4: 158893678-158893678 on Build GRCh38
rs369800736 Chr.4: 158893736-158893736 on Build GRCh38
rs767781768 Chr.4: 158893774-158893774 on Build GRCh38
rs888331514 Chr.4: 158893973-158893974 on Build GRCh38
rs188144435 Chr.4: 158893891-158893891 on Build GRCh38
rs894283327 Chr.4: 158893635-158893636 on Build GRCh38
rs961205125 Chr.4: 158893571-158893571 on Build GRCh38
rs1012786544 Chr.4: 158893645-158893645 on Build GRCh38
rs919856742 Chr.4: 158893591-158893591 on Build GRCh38
rs938420766 Chr.4: 158893545-158893545 on Build GRCh38
rs563856075 Chr.4: 158893631-158893631 on Build GRCh38
rs747519186 Chr.4: 158893695-158893695 on Build GRCh38
rs1004891675 Chr.4: 158893904-158893904 on Build GRCh38
rs748486644 Chr.4: 158893734-158893734 on Build GRCh38
rs961172663 Chr.4: 158893942-158893942 on Build GRCh38
rs932609974 Chr.4: 158893646-158893646 on Build GRCh38
rs1014976668 Chr.4: 158893938-158893938 on Build GRCh38
rs373687039 Chr.4: 158893800-158893800 on Build GRCh38
rs549137182 Chr.4: 158893974-158893974 on Build GRCh38
rs983542306 Chr.4: 158893533-158893533 on Build GRCh38
rs973860170 Chr.4: 158893970-158893970 on Build GRCh38
rs888385559 Chr.4: 158893659-158893659 on Build GRCh38
rs182999722 Chr.4: 158893813-158893813 on Build GRCh38
rs547814176 Chr.4: 158893859-158893859 on Build GRCh38
rs895372303 Chr.4: 158893846-158893846 on Build GRCh38
rs764550424 Chr.4: 158893814-158893814 on Build GRCh38
rs973933845 Chr.4: 158893574-158893574 on Build GRCh38
rs745968996 Chr.4: 158893704-158893706 on Build GRCh38

View Full Product Details