Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 165467605-165468116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPE
Entrez Gene ID
1363
Gene Name
carboxypeptidase E
Gene Aliases
CPH
UniGene
Hs.75360
Gene Chromosome Location
Chr.4: 165378942-165498547 on Build GRCh38
Transcripts
NM_001873.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3335542 c.544G>A p.A182T Chr.4: 165467727-165467727 on Build GRCh38
6241038 c.567G>T p.R189R Chr.4: 165467750-165467750 on Build GRCh38
5400766 c.536G>A p.R179Q Chr.4: 165467719-165467719 on Build GRCh38
274642 c.610G>A p.E204K Chr.4: 165467793-165467793 on Build GRCh38
6885657 c.522C>T p.D174D Chr.4: 165467705-165467705 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369463243 Chr.4: 165467874-165467874 on Build GRCh38
rs778955407 Chr.4: 165467773-165467773 on Build GRCh38
rs957030015 Chr.4: 165467941-165467941 on Build GRCh38
rs773137950 Chr.4: 165467868-165467868 on Build GRCh38
rs867760910 Chr.4: 165467718-165467718 on Build GRCh38
rs34171173 Chr.4: 165467649-165467650 on Build GRCh38
rs147923491 Chr.4: 165467781-165467781 on Build GRCh38
rs1014018933 Chr.4: 165467894-165467894 on Build GRCh38
rs116623425 Chr.4: 165468041-165468041 on Build GRCh38
rs757572105 Chr.4: 165467893-165467893 on Build GRCh38
rs760596400 Chr.4: 165467650-165467650 on Build GRCh38
rs781233088 Chr.4: 165467864-165467864 on Build GRCh38
rs116172145 Chr.4: 165467647-165467647 on Build GRCh38
rs764515112 Chr.4: 165467888-165467888 on Build GRCh38
rs778095440 Chr.4: 165467750-165467750 on Build GRCh38
rs375512739 Chr.4: 165467730-165467730 on Build GRCh38
rs748015585 Chr.4: 165467866-165467866 on Build GRCh38
rs373594119 Chr.4: 165467736-165467736 on Build GRCh38
rs758920925 Chr.4: 165467822-165467822 on Build GRCh38
rs751146124 Chr.4: 165467666-165467669 on Build GRCh38
rs149760126 Chr.4: 165467968-165467968 on Build GRCh38
rs932223386 Chr.4: 165468025-165468025 on Build GRCh38
rs898866656 Chr.4: 165467689-165467689 on Build GRCh38
rs757471198 Chr.4: 165467843-165467843 on Build GRCh38
rs966215766 Chr.4: 165467964-165467964 on Build GRCh38
rs988564288 Chr.4: 165467960-165467960 on Build GRCh38
rs568585778 Chr.4: 165467867-165467867 on Build GRCh38
rs201036265 Chr.4: 165467887-165467887 on Build GRCh38
rs753112662 Chr.4: 165467826-165467826 on Build GRCh38
rs931878903 Chr.4: 165467717-165467717 on Build GRCh38
rs922233265 Chr.4: 165468024-165468024 on Build GRCh38
rs779053382 Chr.4: 165467848-165467848 on Build GRCh38
rs768099130 Chr.4: 165467824-165467824 on Build GRCh38
rs764521091 Chr.4: 165467673-165467674 on Build GRCh38
rs778238027 Chr.4: 165467914-165467914 on Build GRCh38
rs747421668 Chr.4: 165468046-165468046 on Build GRCh38
rs768864865 Chr.4: 165467892-165467892 on Build GRCh38
rs779163656 Chr.4: 165467677-165467678 on Build GRCh38
rs750582097 Chr.4: 165467899-165467899 on Build GRCh38
rs949914338 Chr.4: 165468075-165468075 on Build GRCh38
rs527936136 Chr.4: 165467719-165467719 on Build GRCh38
rs755536693 Chr.4: 165467732-165467732 on Build GRCh38
rs765477706 Chr.4: 165467895-165467895 on Build GRCh38
rs761453876 Chr.4: 165467674-165467674 on Build GRCh38
rs776485567 Chr.4: 165467658-165467658 on Build GRCh38
rs144727363 Chr.4: 165467748-165467748 on Build GRCh38
rs748513122 Chr.4: 165468071-165468071 on Build GRCh38
rs766054842 Chr.4: 165467820-165467820 on Build GRCh38
rs756392535 Chr.4: 165467749-165467749 on Build GRCh38
rs751119399 Chr.4: 165467821-165467821 on Build GRCh38
rs768406124 Chr.4: 165467653-165467653 on Build GRCh38
rs773050049 Chr.4: 165467816-165467816 on Build GRCh38
rs750690232 Chr.4: 165467793-165467807 on Build GRCh38
rs373115142 Chr.4: 165467789-165467789 on Build GRCh38
rs761198746 Chr.4: 165467674-165467674 on Build GRCh38
rs999648510 Chr.4: 165467817-165467817 on Build GRCh38
rs1036422492 Chr.4: 165468065-165468065 on Build GRCh38
rs745972849 Chr.4: 165467850-165467850 on Build GRCh38
rs774106692 Chr.4: 165467875-165467875 on Build GRCh38
rs754122686 Chr.4: 165467840-165467840 on Build GRCh38
rs769446692 Chr.4: 165467815-165467815 on Build GRCh38
rs1008658045 Chr.4: 165467747-165467747 on Build GRCh38
rs754217127 Chr.4: 165467674-165467675 on Build GRCh38
rs757627968 Chr.4: 165467674-165467676 on Build GRCh38
rs778250104 Chr.4: 165468040-165468040 on Build GRCh38
rs531774367 Chr.4: 165467814-165467814 on Build GRCh38
rs368903815 Chr.4: 165467783-165467783 on Build GRCh38
rs749025729 Chr.4: 165467870-165467870 on Build GRCh38
rs140724941 Chr.4: 165467930-165467930 on Build GRCh38
rs573915071 Chr.4: 165468028-165468028 on Build GRCh38
rs563869512 Chr.4: 165468027-165468027 on Build GRCh38
rs539436772 Chr.4: 165467929-165467929 on Build GRCh38
rs59371445 Chr.4: 165467861-165467861 on Build GRCh38
rs766913324 Chr.4: 165467881-165467881 on Build GRCh38
rs749403366 Chr.4: 165467645-165467645 on Build GRCh38
rs770968886 Chr.4: 165467646-165467646 on Build GRCh38
rs372224022 Chr.4: 165467693-165467693 on Build GRCh38
rs922151049 Chr.4: 165467701-165467701 on Build GRCh38
rs762682435 Chr.4: 165467819-165467819 on Build GRCh38
rs771054364 Chr.4: 165467762-165467762 on Build GRCh38
rs776120236 Chr.4: 165467886-165467886 on Build GRCh38
rs764349458 Chr.4: 165467837-165467837 on Build GRCh38
rs1020023825 Chr.4: 165467961-165467961 on Build GRCh38
rs112084290 Chr.4: 165467982-165467983 on Build GRCh38
rs932030601 Chr.4: 165467780-165467780 on Build GRCh38
rs189226554 Chr.4: 165467933-165467933 on Build GRCh38
rs756549147 Chr.4: 165467827-165467827 on Build GRCh38
rs764962318 Chr.4: 165467676-165467676 on Build GRCh38
rs917970454 Chr.4: 165468072-165468072 on Build GRCh38
rs762590299 Chr.4: 165467698-165467698 on Build GRCh38
rs758557033 Chr.4: 165467904-165467904 on Build GRCh38

View Full Product Details