Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 168894444-168894947 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBR4
Entrez Gene ID
84869
Gene Name
carbonyl reductase 4
Gene Aliases
SDR45C1
UniGene
Hs.659311
Gene Chromosome Location
Chr.4: 168894484-169010317 on Build GRCh38
Transcripts
XM_005263315.2 XM_017008782.1 NM_001166108.1 NM_001166109.1 NM_001166110.1 NM_016081.3 XM_005262861.4 XM_005262866.2 XM_011531768.2 XM_011531769.2 XM_011531770.2 XM_011531771.2 XM_011531772.2 XM_011531773.1 XM_011531774.1 XM_011531775.1 XM_017007910.1

Gene Symbol
PALLD
Entrez Gene ID
23022
Gene Name
palladin, cytoskeletal associated protein
Gene Aliases
CGI-151, CGI151, MYN, PNCA1, SIH002
UniGene
Hs.151220
Gene Chromosome Location
Chr.4: 168497039-168928457 on Build GRCh38
Transcripts
XM_005263315.2 XM_017008782.1 NM_001166108.1 NM_001166109.1 NM_001166110.1 NM_016081.3 XM_005262861.4 XM_005262866.2 XM_011531768.2 XM_011531769.2 XM_011531770.2 XM_011531771.2 XM_011531772.2 XM_011531773.1 XM_011531774.1 XM_011531775.1 XM_017007910.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5471849 c.2101-9G>A p.? Chr.4: 168894570-168894570 on Build GRCh38
1053124 c.2112C>T p.Y704Y Chr.4: 168894590-168894590 on Build GRCh38
5471850 c.1627-9G>A p.? Chr.4: 168894570-168894570 on Build GRCh38
1695249 c.2132G>A p.R711Q Chr.4: 168894610-168894610 on Build GRCh38
6239616 c.1682C>A p.T561N Chr.4: 168894634-168894634 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs946641114 Chr.4: 168894491-168894491 on Build GRCh38
rs923842193 Chr.4: 168894747-168894747 on Build GRCh38
rs762267147 Chr.4: 168894653-168894653 on Build GRCh38
rs958807230 Chr.4: 168894714-168894714 on Build GRCh38
rs1039422295 Chr.4: 168894864-168894864 on Build GRCh38
rs529866121 Chr.4: 168894542-168894542 on Build GRCh38
rs768950992 Chr.4: 168894613-168894613 on Build GRCh38
rs999332535 Chr.4: 168894520-168894520 on Build GRCh38
rs768485147 Chr.4: 168894641-168894641 on Build GRCh38
rs564531182 Chr.4: 168894468-168894468 on Build GRCh38
rs754277865 Chr.4: 168894533-168894533 on Build GRCh38
rs763116219 Chr.4: 168894720-168894720 on Build GRCh38
rs902634226 Chr.4: 168894524-168894524 on Build GRCh38
rs762322026 Chr.4: 168894535-168894535 on Build GRCh38
rs754795126 Chr.4: 168894587-168894587 on Build GRCh38
rs569394160 Chr.4: 168894865-168894865 on Build GRCh38
rs767498519 Chr.4: 168894498-168894499 on Build GRCh38
rs769876532 Chr.4: 168894536-168894537 on Build GRCh38
rs1055504727 Chr.4: 168894478-168894478 on Build GRCh38
rs140360991 Chr.4: 168894675-168894675 on Build GRCh38
rs372414187 Chr.4: 168894590-168894590 on Build GRCh38
rs947361342 Chr.4: 168894740-168894740 on Build GRCh38
rs749980711 Chr.4: 168894554-168894554 on Build GRCh38
rs777923870 Chr.4: 168894554-168894555 on Build GRCh38
rs1050917985 Chr.4: 168894754-168894754 on Build GRCh38
rs374250410 Chr.4: 168894611-168894611 on Build GRCh38
rs760950843 Chr.4: 168894742-168894742 on Build GRCh38
rs751153163 Chr.4: 168894725-168894725 on Build GRCh38
rs11732454 Chr.4: 168894824-168894824 on Build GRCh38
rs769887762 Chr.4: 168894599-168894599 on Build GRCh38
rs981612426 Chr.4: 168894683-168894683 on Build GRCh38
rs748036704 Chr.4: 168894591-168894591 on Build GRCh38
rs902378006 Chr.4: 168894504-168894504 on Build GRCh38
rs878854269 Chr.4: 168894682-168894682 on Build GRCh38
rs1043644614 Chr.4: 168894500-168894500 on Build GRCh38
rs185234677 Chr.4: 168894546-168894546 on Build GRCh38
rs368890611 Chr.4: 168894570-168894570 on Build GRCh38
rs4293759 Chr.4: 168894610-168894610 on Build GRCh38
rs376654786 Chr.4: 168894681-168894681 on Build GRCh38
rs762509448 Chr.4: 168894717-168894717 on Build GRCh38
rs878854268 Chr.4: 168894622-168894622 on Build GRCh38
rs112659066 Chr.4: 168894577-168894577 on Build GRCh38
rs769639260 Chr.4: 168894516-168894516 on Build GRCh38
rs757885388 Chr.4: 168894569-168894569 on Build GRCh38
rs867266337 Chr.4: 168894570-168894572 on Build GRCh38
rs147151489 Chr.4: 168894669-168894669 on Build GRCh38
rs115275852 Chr.4: 168894750-168894750 on Build GRCh38
rs775439441 Chr.4: 168894603-168894603 on Build GRCh38
rs1006523205 Chr.4: 168894545-168894545 on Build GRCh38
rs200060953 Chr.4: 168894685-168894685 on Build GRCh38
rs745941762 Chr.4: 168894607-168894607 on Build GRCh38
rs28380014 Chr.4: 168894763-168894763 on Build GRCh38
rs754688204 Chr.4: 168894727-168894727 on Build GRCh38
rs749041581 Chr.4: 168894608-168894608 on Build GRCh38
rs1021022517 Chr.4: 168894522-168894522 on Build GRCh38
rs539985043 Chr.4: 168894866-168894866 on Build GRCh38
rs1034429265 Chr.4: 168894620-168894620 on Build GRCh38
rs1006617622 Chr.4: 168894838-168894838 on Build GRCh38
rs946730991 Chr.4: 168894852-168894852 on Build GRCh38
rs938281880 Chr.4: 168894471-168894471 on Build GRCh38

View Full Product Details