Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 185413594-185414105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf47
Entrez Gene ID
441054
Gene Name
chromosome 4 open reading frame 47
Gene Aliases
-
UniGene
Hs.549398
Gene Chromosome Location
Chr.4: 185405227-185449828 on Build GRCh38
Transcripts
XM_017008236.1 XM_017008237.1 XM_017008238.1 XM_017008239.1 XM_017008241.1 XM_017008242.1 NM_018359.3 NR_028085.1

Gene Symbol
UFSP2
Entrez Gene ID
55325
Gene Name
UFM1 specific peptidase 2
Gene Aliases
BHD, C4orf20
UniGene
Hs.713548
Gene Chromosome Location
Chr.4: 185399540-185425985 on Build GRCh38
Transcripts
XM_017008236.1 XM_017008237.1 XM_017008238.1 XM_017008239.1 XM_017008241.1 XM_017008242.1 NM_018359.3 NR_028085.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6329489 c.685G>T p.E229* Chr.4: 185413872-185413872 on Build GRCh38
4780326 c.713A>C p.H238P Chr.4: 185413844-185413844 on Build GRCh38
4780317 c.715G>T p.D239Y Chr.4: 185413842-185413842 on Build GRCh38
1664874 c.784A>G p.R262G Chr.4: 185413773-185413773 on Build GRCh38
3428382 c.715G>A p.D239N Chr.4: 185413842-185413842 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767535710 Chr.4: 185413896-185413896 on Build GRCh38
rs918158692 Chr.4: 185413879-185413879 on Build GRCh38
rs778055260 Chr.4: 185413749-185413749 on Build GRCh38
rs769742158 Chr.4: 185413840-185413840 on Build GRCh38
rs752847349 Chr.4: 185413854-185413854 on Build GRCh38
rs752915238 Chr.4: 185413816-185413816 on Build GRCh38
rs764169029 Chr.4: 185413911-185413911 on Build GRCh38
rs1020067991 Chr.4: 185413783-185413783 on Build GRCh38
rs201286860 Chr.4: 185413910-185413910 on Build GRCh38
rs1048434545 Chr.4: 185413917-185413917 on Build GRCh38
rs774520214 Chr.4: 185413880-185413884 on Build GRCh38
rs760111490 Chr.4: 185413676-185413676 on Build GRCh38
rs762659528 Chr.4: 185413845-185413845 on Build GRCh38
rs750230737 Chr.4: 185413712-185413712 on Build GRCh38
rs776156275 Chr.4: 185413826-185413826 on Build GRCh38
rs752755472 Chr.4: 185413733-185413733 on Build GRCh38
rs888615680 Chr.4: 185413895-185413895 on Build GRCh38
rs572891654 Chr.4: 185413979-185413979 on Build GRCh38
rs149105536 Chr.4: 185413730-185413730 on Build GRCh38
rs757889147 Chr.4: 185413768-185413768 on Build GRCh38
rs759741764 Chr.4: 185413883-185413884 on Build GRCh38
rs1009891619 Chr.4: 185413756-185413756 on Build GRCh38
rs775322292 Chr.4: 185413712-185413715 on Build GRCh38
rs542256425 Chr.4: 185413820-185413820 on Build GRCh38
rs762731792 Chr.4: 185413791-185413791 on Build GRCh38
rs959566033 Chr.4: 185413862-185413862 on Build GRCh38
rs768147237 Chr.4: 185413823-185413823 on Build GRCh38
rs911084082 Chr.4: 185413956-185413956 on Build GRCh38
rs868847285 Chr.4: 185414066-185414066 on Build GRCh38
rs73873445 Chr.4: 185413645-185413645 on Build GRCh38
rs537207355 Chr.4: 185413656-185413656 on Build GRCh38
rs779371750 Chr.4: 185413772-185413772 on Build GRCh38
rs923764808 Chr.4: 185413660-185413660 on Build GRCh38
rs779590111 Chr.4: 185413795-185413795 on Build GRCh38
rs1047288227 Chr.4: 185413867-185413867 on Build GRCh38
rs954586597 Chr.4: 185413631-185413631 on Build GRCh38
rs370137378 Chr.4: 185413811-185413811 on Build GRCh38
rs551272481 Chr.4: 185413748-185413748 on Build GRCh38
rs761802629 Chr.4: 185413704-185413704 on Build GRCh38
rs763165910 Chr.4: 185413717-185413718 on Build GRCh38
rs746253806 Chr.4: 185413779-185413779 on Build GRCh38
rs918559615 Chr.4: 185413909-185413909 on Build GRCh38
rs761712625 Chr.4: 185413839-185413839 on Build GRCh38
rs188710819 Chr.4: 185413853-185413853 on Build GRCh38
rs181515584 Chr.4: 185413902-185413902 on Build GRCh38
rs780537595 Chr.4: 185413923-185413923 on Build GRCh38
rs538793028 Chr.4: 185413986-185413986 on Build GRCh38
rs978504310 Chr.4: 185413832-185413832 on Build GRCh38
rs768525713 Chr.4: 185413678-185413678 on Build GRCh38
rs766735448 Chr.4: 185413867-185413868 on Build GRCh38
rs751589409 Chr.4: 185414067-185414067 on Build GRCh38
rs968472119 Chr.4: 185413785-185413785 on Build GRCh38
rs972727617 Chr.4: 185413908-185413908 on Build GRCh38
rs764248250 Chr.4: 185413869-185413869 on Build GRCh38
rs754367449 Chr.4: 185413761-185413761 on Build GRCh38
rs771693106 Chr.4: 185413792-185413792 on Build GRCh38
rs773020299 Chr.4: 185413843-185413843 on Build GRCh38
rs901069980 Chr.4: 185414042-185414042 on Build GRCh38
rs757799197 Chr.4: 185413889-185413889 on Build GRCh38
rs369673515 Chr.4: 185413693-185413693 on Build GRCh38
rs764982433 Chr.4: 185413705-185413705 on Build GRCh38
rs900185140 Chr.4: 185413951-185413951 on Build GRCh38
rs978027478 Chr.4: 185414078-185414078 on Build GRCh38
rs761020954 Chr.4: 185413863-185413863 on Build GRCh38
rs536525619 Chr.4: 185413900-185413900 on Build GRCh38
rs1030637384 Chr.4: 185413627-185413627 on Build GRCh38

View Full Product Details