Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 185414848-185415344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf47
Entrez Gene ID
441054
Gene Name
chromosome 4 open reading frame 47
Gene Aliases
-
UniGene
Hs.549398
Gene Chromosome Location
Chr.4: 185405227-185449828 on Build GRCh38
Transcripts
XM_017008236.1 XM_017008237.1 XM_017008238.1 XM_017008239.1 XM_017008241.1 XM_017008242.1 NM_018359.3 NR_028085.1

Gene Symbol
UFSP2
Entrez Gene ID
55325
Gene Name
UFM1 specific peptidase 2
Gene Aliases
BHD, C4orf20
UniGene
Hs.713548
Gene Chromosome Location
Chr.4: 185399540-185425985 on Build GRCh38
Transcripts
XM_017008236.1 XM_017008237.1 XM_017008238.1 XM_017008239.1 XM_017008241.1 XM_017008242.1 NM_018359.3 NR_028085.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4123739 c.616A>T p.K206* Chr.4: 185415223-185415223 on Build GRCh38
277991 c.620A>C p.K207T Chr.4: 185415219-185415219 on Build GRCh38
4798685 c.645T>G p.P215P Chr.4: 185415194-185415194 on Build GRCh38
7415432 c.519T>G p.I173M Chr.4: 185415320-185415320 on Build GRCh38
4739035 c.496C>T p.R166C Chr.4: 185415343-185415343 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760932642 Chr.4: 185415159-185415159 on Build GRCh38
rs762196328 Chr.4: 185415278-185415278 on Build GRCh38
rs368874211 Chr.4: 185415108-185415108 on Build GRCh38
rs756893960 Chr.4: 185415278-185415279 on Build GRCh38
rs753581590 Chr.4: 185415225-185415243 on Build GRCh38
rs767277480 Chr.4: 185415009-185415012 on Build GRCh38
rs745662711 Chr.4: 185415272-185415272 on Build GRCh38
rs994281698 Chr.4: 185414882-185414882 on Build GRCh38
rs982807105 Chr.4: 185414925-185414925 on Build GRCh38
rs544285954 Chr.4: 185415109-185415109 on Build GRCh38
rs747598189 Chr.4: 185415233-185415233 on Build GRCh38
rs758982123 Chr.4: 185415211-185415211 on Build GRCh38
rs751940089 Chr.4: 185415220-185415220 on Build GRCh38
rs1035083498 Chr.4: 185414922-185414922 on Build GRCh38
rs200801769 Chr.4: 185415221-185415221 on Build GRCh38
rs530553349 Chr.4: 185415044-185415044 on Build GRCh38
rs776788760 Chr.4: 185415197-185415197 on Build GRCh38
rs776983365 Chr.4: 185415277-185415277 on Build GRCh38
rs367904498 Chr.4: 185415266-185415266 on Build GRCh38
rs747683633 Chr.4: 185415116-185415116 on Build GRCh38
rs561192551 Chr.4: 185415134-185415134 on Build GRCh38
rs769652789 Chr.4: 185415129-185415129 on Build GRCh38
rs755851735 Chr.4: 185415114-185415115 on Build GRCh38
rs190859418 Chr.4: 185414936-185414936 on Build GRCh38
rs2289722 Chr.4: 185415311-185415311 on Build GRCh38
rs976869944 Chr.4: 185414972-185414972 on Build GRCh38
rs975660219 Chr.4: 185415083-185415083 on Build GRCh38
rs763893718 Chr.4: 185415177-185415180 on Build GRCh38
rs967230506 Chr.4: 185415112-185415112 on Build GRCh38
rs202022021 Chr.4: 185415300-185415300 on Build GRCh38
rs977273637 Chr.4: 185415123-185415123 on Build GRCh38
rs774253815 Chr.4: 185415143-185415143 on Build GRCh38
rs142237251 Chr.4: 185415184-185415184 on Build GRCh38
rs1025876570 Chr.4: 185414904-185414904 on Build GRCh38
rs78213743 Chr.4: 185414923-185414923 on Build GRCh38
rs773978794 Chr.4: 185415271-185415271 on Build GRCh38
rs770625826 Chr.4: 185415136-185415136 on Build GRCh38
rs909871970 Chr.4: 185415005-185415005 on Build GRCh38
rs564874831 Chr.4: 185415023-185415023 on Build GRCh38
rs935061504 Chr.4: 185415174-185415174 on Build GRCh38
rs962989807 Chr.4: 185415019-185415019 on Build GRCh38
rs765418421 Chr.4: 185415205-185415205 on Build GRCh38
rs182238194 Chr.4: 185414951-185414951 on Build GRCh38
rs544651670 Chr.4: 185414957-185414957 on Build GRCh38
rs749161129 Chr.4: 185415264-185415264 on Build GRCh38
rs368621371 Chr.4: 185414924-185414924 on Build GRCh38
rs780826514 Chr.4: 185415223-185415223 on Build GRCh38
rs775161244 Chr.4: 185414911-185414911 on Build GRCh38
rs750893352 Chr.4: 185415206-185415206 on Build GRCh38
rs763316965 Chr.4: 185415313-185415313 on Build GRCh38
rs755605765 Chr.4: 185415250-185415250 on Build GRCh38
rs777407096 Chr.4: 185415262-185415262 on Build GRCh38
rs752750800 Chr.4: 185415111-185415112 on Build GRCh38
rs532559381 Chr.4: 185415318-185415318 on Build GRCh38
rs546896521 Chr.4: 185415276-185415276 on Build GRCh38
rs935943346 Chr.4: 185415145-185415145 on Build GRCh38
rs762002036 Chr.4: 185415202-185415202 on Build GRCh38
rs374899013 Chr.4: 185415137-185415137 on Build GRCh38
rs1052076890 Chr.4: 185415181-185415181 on Build GRCh38

View Full Product Details