Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 864312-864805 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD9
Entrez Gene ID
65980
Gene Name
bromodomain containing 9
Gene Aliases
LAVS3040, PRO9856
UniGene
Hs.449278
Gene Chromosome Location
Chr.5: 863735-892824 on Build GRCh38
Transcripts
NM_001009877.2 NM_001317951.1 NM_023924.4 NR_134296.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1070997 c.1776G>A p.A592A Chr.5: 864486-864486 on Build GRCh38
3139994 c.1487C>T p.A496V Chr.5: 864487-864487 on Build GRCh38
6855319 c.1417C>A p.R473S Chr.5: 864557-864557 on Build GRCh38
1496169 c.1434G>A p.E478E Chr.5: 864540-864540 on Build GRCh38
5111718 c.1731C>T p.D577D Chr.5: 864531-864531 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780561168 Chr.5: 864571-864571 on Build GRCh38
rs1053855391 Chr.5: 864562-864562 on Build GRCh38
rs772275207 Chr.5: 864486-864486 on Build GRCh38
rs540370412 Chr.5: 864522-864522 on Build GRCh38
rs999363336 Chr.5: 864701-864701 on Build GRCh38
rs906882504 Chr.5: 864621-864621 on Build GRCh38
rs530843808 Chr.5: 864689-864689 on Build GRCh38
rs1039247800 Chr.5: 864342-864342 on Build GRCh38
rs73730994 Chr.5: 864708-864708 on Build GRCh38
rs777643182 Chr.5: 864720-864720 on Build GRCh38
rs906835836 Chr.5: 864371-864371 on Build GRCh38
rs557713899 Chr.5: 864412-864412 on Build GRCh38
rs751527206 Chr.5: 864772-864772 on Build GRCh38
rs764356349 Chr.5: 864502-864503 on Build GRCh38
rs890743798 Chr.5: 864662-864663 on Build GRCh38
rs1021754916 Chr.5: 864680-864680 on Build GRCh38
rs751819467 Chr.5: 864528-864528 on Build GRCh38
rs999594317 Chr.5: 864361-864361 on Build GRCh38
rs953269703 Chr.5: 864763-864763 on Build GRCh38
rs575914225 Chr.5: 864413-864413 on Build GRCh38
rs573206923 Chr.5: 864519-864519 on Build GRCh38
rs746628495 Chr.5: 864489-864489 on Build GRCh38
rs764239470 Chr.5: 864540-864540 on Build GRCh38
rs765027761 Chr.5: 864432-864432 on Build GRCh38
rs1048868470 Chr.5: 864349-864349 on Build GRCh38
rs940445572 Chr.5: 864596-864596 on Build GRCh38
rs984301757 Chr.5: 864726-864726 on Build GRCh38
rs975419541 Chr.5: 864751-864751 on Build GRCh38
rs887561679 Chr.5: 864594-864594 on Build GRCh38
rs563403530 Chr.5: 864642-864642 on Build GRCh38
rs777807470 Chr.5: 864514-864514 on Build GRCh38
rs750236621 Chr.5: 864451-864451 on Build GRCh38
rs781590586 Chr.5: 864666-864666 on Build GRCh38
rs921883099 Chr.5: 864755-864755 on Build GRCh38
rs757792104 Chr.5: 864467-864467 on Build GRCh38
rs780668199 Chr.5: 864612-864612 on Build GRCh38
rs752597145 Chr.5: 864350-864350 on Build GRCh38
rs746272978 Chr.5: 864483-864483 on Build GRCh38
rs372222979 Chr.5: 864497-864497 on Build GRCh38
rs761804915 Chr.5: 864431-864431 on Build GRCh38
rs368965233 Chr.5: 864581-864581 on Build GRCh38
rs1034593827 Chr.5: 864643-864643 on Build GRCh38
rs559109064 Chr.5: 864553-864553 on Build GRCh38
rs1003438013 Chr.5: 864683-864683 on Build GRCh38
rs753953007 Chr.5: 864545-864545 on Build GRCh38
rs371971169 Chr.5: 864546-864546 on Build GRCh38
rs750871144 Chr.5: 864552-864552 on Build GRCh38
rs768347792 Chr.5: 864500-864500 on Build GRCh38
rs965786483 Chr.5: 864749-864749 on Build GRCh38
rs779900379 Chr.5: 864487-864487 on Build GRCh38
rs779217610 Chr.5: 864475-864475 on Build GRCh38
rs984674736 Chr.5: 864771-864771 on Build GRCh38
rs144044545 Chr.5: 864457-864457 on Build GRCh38
rs548363568 Chr.5: 864622-864622 on Build GRCh38
rs758871899 Chr.5: 864557-864557 on Build GRCh38
rs375279506 Chr.5: 864550-864550 on Build GRCh38
rs753071813 Chr.5: 864434-864438 on Build GRCh38
rs761161687 Chr.5: 864516-864516 on Build GRCh38
rs918385161 Chr.5: 864426-864426 on Build GRCh38
rs141705577 Chr.5: 864531-864531 on Build GRCh38
rs200771795 Chr.5: 864459-864459 on Build GRCh38
rs368064725 Chr.5: 864416-864416 on Build GRCh38
rs536139559 Chr.5: 864362-864362 on Build GRCh38
rs894713036 Chr.5: 864662-864662 on Build GRCh38
rs1016636329 Chr.5: 864750-864750 on Build GRCh38
rs762859429 Chr.5: 864526-864526 on Build GRCh38
rs893854314 Chr.5: 864587-864587 on Build GRCh38
rs900703377 Chr.5: 864347-864347 on Build GRCh38
rs367592539 Chr.5: 864605-864605 on Build GRCh38
rs767750037 Chr.5: 864453-864453 on Build GRCh38
rs757461208 Chr.5: 864634-864634 on Build GRCh38
rs758090084 Chr.5: 864452-864452 on Build GRCh38
rs374226502 Chr.5: 864530-864530 on Build GRCh38
rs749822409 Chr.5: 864465-864465 on Build GRCh38
rs529868667 Chr.5: 864613-864613 on Build GRCh38
rs942297772 Chr.5: 864338-864338 on Build GRCh38
rs970584202 Chr.5: 864415-864415 on Build GRCh38
rs72708957 Chr.5: 864351-864351 on Build GRCh38
rs1023609146 Chr.5: 864657-864657 on Build GRCh38
rs758454620 Chr.5: 864395-864395 on Build GRCh38
rs1034584599 Chr.5: 864393-864393 on Build GRCh38
rs756506634 Chr.5: 864444-864445 on Build GRCh38
rs868738713 Chr.5: 864373-864373 on Build GRCh38
rs115833503 Chr.5: 864464-864464 on Build GRCh38

View Full Product Details